Whole Allergen
f320 Crayfish
f320 Crayfish Scientific Information
Type:
Whole Allergen
Display Name:
Crayfish
Route of Exposure:
Ingestion
Family:
Cambaridae
Species:
Astacus astacus, Procambarus clarkii, Cherax quadricarinatus, Cherax destructor, Pontastacus leptodactylus, Pontastacus leptodactylus, Potamobius leptodactylus
Latin Name:
Astacus astacus
Other Names:
Crawfish, freshwater lobster, red swamp crayfish, narrow-clawed crayfish, red-claw crayfish
Summary
Crayfish is a popular food source as it has an appealing taste and good balance of nutrition, with high levels of protein, low levels of fat and a low number of calories. With increasing consumption of crayfish, allergic reactions have become more common. Typically, allergic symptoms of shellfish allergy appear within minutes, but may occur within 1–2 hours of ingestion, including oral allergy syndrome, urticaria and/or angioedema, gastrointestinal symptoms such as nausea, vomiting, abdominal pains and diarrhea, or respiratory symptoms such as tightness in the chest and shortness of breath. Anaphylaxis may occur in rare cases. Cross-reactivity has been identified between shrimp, crab, lobster and crayfish allergens to a high level.
Allergen
Nature
Crayfish is a popular food source as it has an appealing taste and good balance of nutrition, with high levels of protein, low levels of fat and a low number of calories; therefore, it has become the most widely cultured freshwater shellfish species (1, 2) . The largest consumers of seafood are China, Japan, and North America (2).
The family Cambaridae includes the species Procambarus clarkii, or red swamp crayfish, which is native to northern Mexico and southern United States. The main sources of fresh, frozen or processed foodstuffs containing red swamp crayfish, which are consumed either raw or cooked, are from China and the United States (3, 4). Over 2 million tons of crayfish were produced in 2019, equal to an economic value of more than $18 billion US dollars (4).
The family Parastacidae includes another species of crayfish: the Cherax quadricarinatus, also known as the red claw crayfish, which is indigenous to Australia. Primarily produced in Australia and Malaysia, C. quadricarinatus is exported across the world, particularly to Asian countries. In 2019, its production reached 0.17 million tons, equal to an economic value of $2.1 million US dollars (4).
Taxonomy
Crustaceans fall within the classification of arthropods, which includes over 50,000 species of shrimps, prawns, lobsters, crayfish and barnacles (3).
Taxonomic tree of crayfish (Astacus astacus) (5)
Domain Eukaryota
Kingdom Animalia
Phylum Arthropoda
Subphylum Crustacea
Class Malacostraca
Order Decapoda
Family Astacidae
Genus Astacus
Species Astacus astacus
Epidemiology
Worldwide distribution
With increasing consumption of crayfish, allergic reactions have become more common (2)..
Route of Exposure
Main
Most allergic reactions are caused by ingestion of crayfish and are mediated by IgE (2).
Other topics
A case of occupational asthma caused by lobster or shrimp has been reported in a fishmonger sensitized to crayfish (6). 
Detection
Real-time polymerase chain reaction has been used as a method for identification of crustaceans in food products, offering high sensitivity and specificity, even in cases where the allergenic food has been processed with aggressive treatments of temperature or pressure (7).
Clinical Relevance
Allergic reactions to crayfish, and other shellfish, can result in a range of clinical symptoms (1). Typically, allergic symptoms of shellfish allergy appear within minutes, but may occur within 1–2 hours of ingestion, including oral allergy syndrome, urticaria and/or angioedema, gastrointestinal symptoms such as nausea, vomiting, abdominal pains and diarrhea, or respiratory symptoms such as tightness in the chest and shortness of breath (2, 3, 8). Infrequently, anaphylactic shock may occur (3)
Molecular Aspects
Table adapted (3, 9).
Allergen name
Protein group (if known)
Source
Pan j 1
Tropomyosin
Panulirus japonicus
Che de 1
Tropomyosin
Cherax destructor
Che de 1.0101
Tropomyosin
Cherax destructor
Pon l 4
SCP
Pontastacus leptodactylus, Potamobius leptodactylus
Pon l 4.0101
SCP
Astacus leptodactylus, Pontastacus leptodactylus, Potamobius leptodactylus
Pon l 7
Troponin
Astacus leptodactylus, Pontastacus leptodactylus, Potamobius leptodactylus
Pon l 7.0101
Troponin
Astacus leptodactylus, Pontastacus leptodactylus, Potamobius leptodactylus
Pro c 1
Tropomyosin
Procambarus clarkii
Pro c 1.0101
Tropomyosin
Procambarus clarkii
Pro c 2
Arginine kinase
Procambarus clarkii
Pro c 2.0101
Arginine kinase
Procambarus clarkii
Pro c 21kD
Ferritin
Procambarus clarkii
Pro c 4
SCP
Procambarus clarkii
Pro c 5
Myosin
Procambarus clarkii
Pro c 5.0101
Myosin
Procambarus clarkii
Pro c 8
Triosephosphate isomerase
Procambarus clarkii
Pro c 8.0101
Triosephosphate isomerase
Procambarus clarkii
Pro c HC
Hemocyanins
Procambarus clarkii
Over 20 allergens have been identified in crayfish (9)
Three key crayfish allergens have been thoroughly characterized; tropomyosin (TM), arginine kinase (AK), and sarcoplasmic calcium-binding protein (SCP). TM allergen occurs in many different types of seafood, including shrimp, lobster, crab and mollusks, and has been identified as a major allergen of crayfish; TM is a heat stable myofibrillar protein (2). TM has been identified in raw and boiled samples of C. quadricarinatus (4). TM may undergo a change in secondary structures, which could increase antibody recognition, as a result of high temperature treatment (4). AK is another major allergen of crayfish, and is one of the main cross-reactive allergens in crustaceans; it is a 40 kDa myosinogen protein(2). AK has been identified in raw samples of P. clarkii (4). The third allergen, SCP, is also a shrimp allergen found in white leg Pacific shrimp (Litopenaeus vannamei) and the black tiger prawn (Penaeus monodon); SCP has three subunits and three 22 kDa isoforms, all of which are allergy inducing (2).
Further allergens have also been identified, including myosin light chain 1, or MLC1, and triosephosphate isomerase (2, 10). MLC1 is more resistant to gastric digestion than intestinal digestion, enabling it to preserve immunological activity (2).
Cross-reactivity
Cross-reactivity has been identified between shrimp, crab, lobster and crayfish allergens to a high level (6). Also, reports have emerged regarding cross-reactivity between shellfish and insects; this is likely due to both animals belonging to the Arthropoda phylum (1).
Tropomyosin is regarded as a cross-reactive allergen in shellfish (1) TM from C. quadricarinatus has demonstrated high levels of sequence homology of 95.4–99.3% with other species of freshwater crustaceans, including Procambarus clarkii, Macrobrachium rosenbergii and Macrobrachium lanchesteri (4).
Cross-reactivity has been identified in MLC from crayfish by investigating reactivity with rabbit IgG. Furthermore, crayfish myofibrillar protein was shown to inhibit IgE binding by MLC from crayfish (1)
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed: December  2021
References
  1. Yang Y, Yan HF, Zhang YX, Chen HL, Cao MJ, Li MS, et al. Expression and epitope identification of myosin light chain isoform 1, an allergen in Procambarus clarkii. Food Chem. 2020;317:126422.
  2. Zhang YX, Chen HL, Maleki SJ, Cao MJ, Zhang LJ, Su WJ, et al. Purification, Characterization, and Analysis of the Allergenic Properties of Myosin Light Chain in Procambarus clarkii. J Agric Food Chem. 2015;63(27):6271-82.
  3. Fernandes TJR, Costa J, Oliveira MBPP, Mafra I. An overview on fish and shellfish allergens and current methods of detection. Food and Agricultural Immunology. 2015;26(6):848-69.
  4. Laurchan P, T EK, Srisapoome P, Unajak S, Sinthuvanich C. Molecular Characterization and Cross-Allergenicity of Tropomyosin from Freshwater Crustaceans. J Agric Food Chem. 2021;69(29):8247-56.
  5. ITIS. Crayfish 2021 [cited 2021 5.11.21]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=97491#null.
  6. Jeebhay MF, Robins TG, Lehrer SB, Lopata AL. Occupational seafood allergy: a review. Occup Environ Med. 2001;58(9):553-62.
  7. Santaclara FJ, Espineira M. Fast Real-Time PCR for the Detection of Crustacean Allergens in Foods. Methods Mol Biol. 2017;1620:163-71.
  8. Daul CB MJ, Waring NP, McCants ML, Hughes J, Lehrer SB. Immunologic evaluation of shrimp-allergic individuals. J Allergy Clin Immunol. 1987(80(5)):716-22.
  9. www.allergome.org. <Allergome.org.pdf>.
  10. Yang Y, Zhang YX, Liu M, Maleki SJ, Zhang ML, Liu QM, et al. Triosephosphate Isomerase and Filamin C Share Common Epitopes as Novel Allergens of Procambarus clarkii. J Agric Food Chem. 2017;65(4):950-63.