Whole Allergen
m12 Aureobasidium pullulans
m12 Aureobasidium pullulans Scientific Information
Type:
Whole Allergen
Display Name:
Aureobasidium pullulans
Route of Exposure:
Inhalation
Family:
Dothioraceae
Species:
pullulans
Latin Name:
Aureobasidium pullulans
Summary
Aureobasidium pullulans (also known as Pullularia pullulans) is a ubiquitous mold of the Order Dothideales, found in a large variety of environments, indoors and outdoors, including extremes of temperature. It is a widely used organism in the biotechnology and medical industry. The main particles that induce sensitization are the conidia (a type of spore).
Hypersensitivity pneumonitis (HP), allergic rhinitis and worsening of asthma symptoms are the clinical presentations most commonly associated with A. pullulans allergy. This fungus is also an opportunistic pathogen and it is able to cause a wide range of diseases, especially in immunocompromised patients and those with underlying conditions. Cross reactivity with other molds, especially if taxonomically close, may occur.
Allergen
Nature
Aureobasidium pullulans (also known as Pullularia pullulans, black yeast) is a yeast-like ubiquitous saprophyte mold (1, 2). This fungal species is characterized by highly variable phenotype and physiology; three sub varieties have been identified: A. pullulans var. pullulans, A. pullulans var. melanogenum and A. pullulans var. aubasidani (1, 2), some of which were later re-classified as separate species following genomic analyses (3, 4). The hyphae are hyaline (transparent), smooth and single celled, variable in size and shape, form chlamydospores in places and the colony color can turn dark due to melanin production. Budding of conidia (a type of spore) is frequent. A. pullulans is a fast-growing mold, able to withstand extreme osmotic environments (2, 5) and freezing temperatures (6). The variations observed in the types of colony depend on factors such as the type of carbon substrate used in the medium, age of the colony, temperature and light conditions (7).
In a US study, the researchers found that hyaline conidia presence peaked during periods of rainfall and high winds in the fall and winter (8). However, certain fungi are able to produce spores all year round. There is considerable overlap between peak fungal activity season and other allergens such as grass and weed pollens, often masking the pathology caused by fungal spores in people multi-sensitized (9).
A. pullulans can be found as epiphyte on different fruits and crops (apples, grapes, strawberries, citrus fruits, cabbages, wheat, barley, nuts) and other foods (meat, shrimp, flour, frozen foods); it affects the esthetic appearance of food but is not considered a common cause of spoilage (6). Other sources are plant leaves, wood, damp indoor environments (including cement walls, window frames, glass and latex-coated surfaces), animal feed and even rocks (1, 2, 6, 10). Strains that formed part of a genotyping study were also isolated from human sputum and peritoneal dialysis (1).
A. pullulans is important in the field of microbiology and industrial processes because of its ability to produce pullulan (a biodegradable polysaccharide used for coatings and wrappings). Other metabolites and enzymes of Aureobasidium spp. are widely used in the food and medical industry (11).
Taxonomy 
Aureobasidium pullulans (Pullularia pullulans) belongs to the Family Dothioraceae, Order Dothideales, Class Dothideomycetes, Phylum Ascomycota (12, 13).
Taxonomic tree of Aureobasidium genus (12, 13)
Domain
Eukaryota
Kingdom
Fungi
Phylum
Ascomycota
Subphylum
Pezizomycotina
Class
Dothideomycetes
Order
Dothideales
Family
Dothioraceae
Genus
Aureobasidium
Tissue 
Hyaline unicellular conidia (an asexual, colorless microscopic component released in the environment in lieu of sexual reproduction), which can reach high concentrations in the air and represent the main fungal allergen detected (90%; (8)). 
Epidemiology
Worldwide distribution 
Aureobasidium pullulans causes allergic disease in multiple locations worldwide; including in Poland (14), Mexico (15), Norway (16), North America (8, 17).
Prevalence of hypersensitivity pneumonitis, which may be caused by A. pullulans can vary considerably and if the diagnostic protocols do not agree, it may be underdiagnosed (16).
A study of Skin Prick Tests (SPTs) in asthmatic patients that included A. pullulans in the testing panel resulted in positive reactions in 98% of asthmatic patients and 66% of non-asthmatic controls (14).
Risk factors 
Being male was associated with poorer control of asthma symptoms in patients exposed to indoor molds, including A. pullulans (15).
Certain occupations (e.g. farmers) appear to be at higher risk of developing hypersensitivity pneumonitis following exposure to this mold (16).
Night-time, rainy and windy weather were associated with peaks of hyaline conidia concentrations (8).
Immunocompromised patients are more likely to develop opportunistic A. pullulans infections (18).
Environmental Characteristics
Worldwide distribution 
A. pullulans is commonly found in cold and temperate climates, but it can also be detected in tropical areas such as Thailand (2, 10). It is thus able to survive and replicate in a large variety of habitats and conditions, without significant changes to its genomic makeup (3).
Route of Exposure
Main
Inhalation of mold components, mostly conidia (8)
Detection
Environmental source 
Air
Main methods 
Molecular assay using probes that target the 18S ribosomal RNA gene showed satisfactory performance for the early detection of A. pullulans infection (18). Controlled emission chambers have also been used to determine the source of fungal aerosols (8)
Clinical Relevance
Aureobasidium pullulans is a recognized cause of hypersensitivity pneumonitis (HP) in certain professions (farmers;(16)), but also in children (17). The symptoms of HP include fatigue, fever, cough and dyspnea, other flu-like symptoms, headache and weight loss (16). “Humidifier lung” is a type of HP that has been associated with A. pullulans infection (19). In a study of 405 children born to allergic/asthmatic parents, high fungal concentrations within the home in the first 3 months of life were showed as predictors of allergic rhinitis diagnosed in the first 5 years (20).
Patients sensitized this mold are at higher risk of severe asthma (14), and pre-existing asthma symptoms can be exacerbated by A. pullulans (15). The increased presence of spores of Ascomycota fungi was associated with thunderstorm-related asthma (9).
Aureobasidium pullulans was described as the cause of subcutaneous mycosis in a 66-year old immunocompromised patient who was undergoing chemotherapy (21). Fungemia and pulmonary embolism caused by A. pullulans was diagnosed and successfully treated in another 66-year old patient (22).
Diagnostics Sensitization
The presence of allergen-specific antibodies is determined by skin prick tests (SPTs) and serology for IgE and IgG specificities (8, 9, 16).
Prevention and Therapy
Allergen immunotherapy
Immunotherapy is currently not recommended for patients allergic to molds, due to complexities of the allergens and patient co-allergies (9).
Prevention strategies       
Complete avoidance is often not possible as A. pullulans is a very common mold. For certain professions (such as farmers) where some jobs carry higher risk of exposure, use of protective equipment such as filtered masks and respirators must be considered (16).
Molecular Aspects
Allergenic molecules
IgE from atopic patients bind to several allergens in A. pullulans extracts (8). In a later study, a 64 kDa and a 100 kDa allergen were identified by IgG immunoblot using serum from a farmer with hypersensitivity pneumonitis (16).
Cross-reactivity
Immunoblotting showed that A. pullulans extracts contain antigens that cross react with antibodies specific to certain antigens of the related mold Alternaria alternata (23). In a study carried out on 668 serum samples from patients who had previously recorded at least one IgE positivity to fungal antigens, associations were observed between the patterns of IgE sensitization and fungal phylogenetic relationships. Using a panel of 17 fungal extracts including A. pullulans, some samples were only positive to one fungal species, whereas many were multi-sensitized. The results suggest that the associations are likely due to antigen cross-reactivity between fungal species (24).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed: December  2022
References
  1. Yurlova N, De Hoog G, Van den Ende A. Taxonomy of Aureobasidizum and allied genera. Studies in Mycology. 1999:63-9.
  2. Zalar P, Gostincar C, de Hoog GS, Ursic V, Sudhadham M, Gunde-Cimerman N. Redefinition of Aureobasidium pullulans and its varieties. Studies in mycology. 2008;61:21-38.
  3. Gostinčar C, Turk M, Zajc J, Gunde-Cimerman N. Fifty Aureobasidium pullulans genomes reveal a recombining polyextremotolerant generalist. Environ Microbiol. 2019;21(10):3638-52.
  4. Gostinčar C, Ohm RA, Kogej T, Sonjak S, Turk M, Zajc J, et al. Genome sequencing of four Aureobasidium pullulans varieties: biotechnological potential, stress tolerance, and description of new species. BMC Genomics. 2014;15:549.
  5. Davis U. Aureobasidium spp. Davis, California2021 [cited 2021 23.11.21]. Available from: https://wineserver.ucdavis.edu/industry-info/enology/wine-microbiology/yeast-mold/aureobasidium-spp.
  6. Roukas T. AUREOBASIDIUM. In: Robinson RK, editor. Encyclopedia of Food Microbiology. Oxford: Elsevier; 1999. p. 109-12.
  7. Slepecky RA, Starmer WT. Phenotypic plasticity in fungi: a review with observations on Aureobasidium pullulans. Mycologia. 2009;101(6):823-32.
  8. Taylor PE, Esch R, Flagan RC, House J, Tran L, Glovsky MM. Identification and possible disease mechanisms of an under-recognized fungus, Aureobasidium pullulans. Int Arch Allergy Immunol. 2006;139(1):45-52.
  9. Twaroch TE, Curin M, Valenta R, Swoboda I. Mold allergens in respiratory allergy: from structure to therapy. Allergy Asthma Immunol Res. 2015;7(3):205-20.
  10. Prasongsuk S, Sullivan R, Kuhirun M, Eveleigh D, Punnapayak H. Thailand habitats as sources of pullulan-producing strains of Aureobasidium pullulans. World Journal of Microbiology and Biotechnology. 2005;21:393-8.
  11. van Nieuwenhuijzen EJ. Aureobasidium. In: Batt CA, Tortorello ML, editors. Encyclopedia of Food Microbiology (Second Edition). Oxford: Academic Press; 2014. p. 105-9.
  12. Suzuki T, Kusano K, Kondo N, Nishikawa K, Kuge T, Ohno N. Biological Activity of High-Purity β-1,3-1,6-Glucan Derived from the Black Yeast Aureobasidium pullulans: A Literature Review. Nutrients. 2021;13(1).
  13. Uniprot.org. Taxonomy - Aureobasidium pullulans (Black yeast) (Pullularia pullulans) 2021 [cited 2021 23.11.21]. Available from: https://www.uniprot.org/taxonomy/5580.
  14. Niedoszytko M, Chełmińska M, Jassem E, Czestochowska E. Association between sensitization to Aureobasidium pullulans (Pullularia sp) and severity of asthma. Ann Allergy Asthma Immunol. 2007;98(2):153-6.
  15. Segura-Medina P, Vargas MH, Aguilar-Romero JM, Arreola-Ramírez JL, Miguel-Reyes JL, Salas-Hernández J. Mold burden in house dust and its relationship with asthma control. Respir Med. 2019;150:74-80.
  16. Tjalvin G, Mikkelsen KE, Apelseth TO, Hollund BE, Svanes C, Van Do T. Hypersensitivity Pneumonitis in Farmers: Improving Etiologic Diagnosis to Optimize Counselling. J Agromedicine. 2020;25(1):65-72.
  17. Temprano J, Becker BA, Hutcheson PS, Knutsen AP, Dixit A, Slavin RG. Hypersensitivity pneumonitis secondary to residential exposure to Aureobasidium pullulans in 2 siblings. Ann Allergy Asthma Immunol. 2007;99(6):562-6.
  18. Chan GF, Puad MS, Chin CF, Rashid NA. Emergence of Aureobasidium pullulans as human fungal pathogen and molecular assay for future medical diagnosis. Folia Microbiol (Praha). 2011;56(5):459-67.
  19. Shimoda M, Morimoto K, Tanaka Y, Ken O, Yoshimori K. Humidifier lung induced by endotoxin and various pathogens: Characteristic differences from other phenotypes of hypersensitivity pneumonitis. Respiratory Medicine Case Reports. 2020;31:101267.
  20. Stark PC, Celedón JC, Chew GL, Ryan LM, Burge HA, Muilenberg ML, et al. Fungal levels in the home and allergic rhinitis by 5 years of age. Environmental health perspectives. 2005;113(10):1405-9.
  21. Oliveira LR, Moraes-Souza H, Maltos AL, Santos KC, Molina RJ, Barata CH. Aureobasidium pullulans infection in a patient with chronic lymphocytic leukemia. Rev Soc Bras Med Trop. 2013;46(5):660-2.
  22. Mehta SR, Johns S, Stark P, Fierer J. Successful treatment of Aureobasidium pullulans central catheter-related fungemia and septic pulmonary emboli. IDCases. 2017;10:65-7.
  23. Twaroch TE, Curin M, Sterflinger K, Focke-Tejkl M, Swoboda I, Valenta R. Specific Antibodies for the Detection of Alternaria Allergens and the Identification of Cross-Reactive Antigens in Other Fungi. Int Arch Allergy Immunol. 2016;170(4):269-78.
  24. Soeria-Atmadja D, Onell A, Borgå A. IgE sensitization to fungi mirrors fungal phylogenetic systematics. J Allergy Clin Immunol. 2010;125(6):1379-86.e1.:729-32.