press-releases
Press Releases
Rapid mycoplasma testing method now sccepted by Regulators for QA/QC and Lot Release
MycoSEQ Mycoplasma Detection Method can accelerate production timelines for manufacturers of cell and gene therapies, biotherapeutics, vaccines and other cell-culture-based modalities
BEDFORD, Mass., April 3, 2018 /PRNewswire/ -- Manufacturers of cell-culture-based therapeutics can now rely on a faster method for detecting mycoplasma contamination. The Applied Biosystems MycoSEQ Mycoplasma Detection Kit is a fully integrated solution for real-time PCR-based mycoplasma detection. Used throughout the bioproduction workflow, the MycoSEQ method is an alternative to costly, time-consuming culture-based tests often done externally by contract labs, which can take up to 28 days.
Following validation, regulatory review and acceptance, the MycoSEQ assay and method can be used for lot release by manufacturers in different therapeutic modalities including biotherapeutics, cell and gene therapies, vaccines and other cell-culture-based therapeutics.
To date, nine global manufacturers of therapies subject to U.S. Food and Drug Administration, European Medicines Agency and/or local regulatory agency review have received regulatory acceptance for accelerated lot release protocols that specify the Thermo Fisher Scientific MycoSEQ method.
"Rapid microbiological detection methods for product testing and release are critical for cell culture-based therapies with short shelf lives," said John Duguid, senior director of research and development at Vericel Corporation. "It also makes sense to adopt new, proven methods during product development and licensure to avoid the time and expense of validation after a therapy is actively being marketed to patients."
In late 2017, Thermo Fisher received two patents from The U.S. Patent and Trademark Office for its MycoSEQ real-time PCR-based mycoplasma detection assay. These patents cover the assay's proprietary multiplexed PCR primer approach and the discriminatory positive/extraction control, which are new innovations to real-time PCR that enable this rapid test for mycoplasma to meet the rigorous requirements of regulatory authorities and manufacturers.
"Patient safety is the highest priority for manufacturers of cell-culture-based therapies, and they go to considerable lengths to ensure that," said Michael Brewer, head of pharma analytics at Thermo Fisher. "With the MycoSEQ method, manufacturers can continue that vigilance without the extra time and expense associated with conventional culture-based methods that were, until recently, the only approved method for lot release. Now they have a new solution that brings patented innovations to their processes."
Thermo Fisher Scientific MycoSEQ System
Apr 3, 2018
MycoSEQ System
MycoSEQ System (PRNewsfoto/Thermo Fisher Scientific)
With its combination of optimized sample preparation and multiplexed real-time PCR based detection, the MycoSEQ assay can detect more than 90 mycoplasma species in under five hours, with well-documented sensitivity and specificity. This enables manufacturers to use it at multiple points in their process, from raw material acceptance to lot release, helping to ensure product quality and patient safety.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue exceeding $30 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 80,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media contact information:
Christine Williamson
Greenough (for Thermo Fisher)
617.275.6528
cwilliamson@greenough.biz
SOURCE Thermo Fisher Scientific
If you have any questions regarding this Press Release, please email us at media.relations@thermofisher.com.