New Trace-level Online Silica Analyzer Improves Reliability of Process Water Monitoring
Press Releases
New Trace-level Online Silica Analyzer Improves Reliability of Process Water Monitoring
Thermo Scientific Orion 8030cX Silica Analyzer enables real-time monitoring of ultrapure water, steam and water chemistry across demanding environments in the power generation, oil and gas, chemical, pulp and paper, and semiconductors industries
A new trace-level silica analyzer for real-time monitoring of ultrapure water, steam and water chemistry can more accurately protectexpensive components, including turbines and heat exchangers. The analyzer ensures sustained, energy efficient operations and maximum uptime across the power generation, oil and gas, chemical, pulp and paper, and semiconductor industries.
Thermo Scientific Orion 8030cX Silica Analyzer enables real-time monitoring of ultrapure water, steam and water chemistry across demanding environments in the power generation, oil and gas, chemical, pulp and paper, and semiconductors industries. (Photo: Business Wire)
Thermo Scientific Orion 8030cX Silica Analyzer enables real-time monitoring of ultrapure water, steam and water chemistry across demanding environments in the power generation, oil and gas, chemical, pulp and paper, and semiconductors industries. (Photo: Business Wire)
The new Thermo Scientific Orion 8030cX Silica Analyzer combines a patented fluid delivery system with optimized reagent chemistry to reliably improve silica detection limits to sub-parts-per-billion levels. It delivers accurate and precise results across a wide analysis range, from 0-5000 µg/L. The analyzer demonstrates a 10% improvement in reagent longevity over its competitors - consuming about 200 µL of reagent per test. It can continuously monitor unattended for up to 100 days while simultaneously reducing chemical waste. The improved reagent chemistry minimizes common interferences from phosphate and can be used without performance degradation for over six months (under intermittent operation).
"Impurities in process water can quickly become disruptive and costly without constant surveillance," said Denzil Vaughn, vice president and general manager for growth protection and separation at Thermo Fisher Scientific. "The Orion 8030cX Silica Analyzer answers that need with direct, accurate online monitoring of silica in real time to prevent built-up, improve energy efficiency and maximize uptime."
The Orion 8030cX Silica Analyzer features state-of-the-art automation with advanced intuitive functions such as automated calibration, validation and cleaning, intelligent system diagnostics and alert notifications that reduce manual intervention and maintenance. The IP65-rated rugged analyzer provides superior analytical performance in demanding industrial environments.
For more information about the Thermo Scientific Orion 8030cX Silica Analyzer, please visit www.thermofisher.com/orion8030.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue of approximately $40 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, increasing productivity in their laboratories, improving patient health through diagnostics or the development and manufacture of life-changing therapies, we are here to support them. Our global team delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services, Patheon and PPD. For more information, please visit www.thermofisher.com.
Media contact information:
Mauricio Minotta
Director, Public Relations
Thermo Fisher Scientific
+1 760-929-2456
Mauricio.minotta@thermofisher.com
Source: Thermo Fisher
If you have any questions regarding this Press Release, please email us at media.relations@thermofisher.com