press-releases
Press Releases
Thermo Fisher Scientific to Support National Program to Accelerate SARS-CoV-2 Variant Identification
CARLSBAD, Calif., (March 23, 2022) Thermo Fisher Scientific Inc., the world leader in serving science, today announced its collaboration with the National Institutes of Health (NIH) Rapid Acceleration of Diagnostics (RADx) Initiative, Helix, and Rosalind aimed at developing a new genotyping method for SARS-CoV-2 that could speed up the identification of variants as they emerge.
Utilizing a Polymerase Chain Reaction (PCR)-based genotyping approach that can be implemented in any testing lab performing real-time PCR, this program allows for a significant scale up of SARS-CoV-2 surveillance efforts in the United States. Data generated from this program will be available to the public through the Rosalind Tracker website. This effort was funded by the National Institute of Biomedical Imaging and Bioengineering (NIBIB) as part of the NIH RADx Initiative to increase testing capacity and accessibility for the virus that causes COVID-19.
The rapid emergence of new SARS-CoV-2 variants raises several important public health questions, such as their ability to cause disease and their impact on vaccine efficacy. This makes it critical to track variants to better understand transmission patterns and possible consequences on health care resources. Next-Generation Sequencing (NGS) is the conventional method for detecting and tracing new variants, with about 5 percent of randomly selected SARS-CoV-2 positive samples in the United States being sent for variant identification. NGS, however, can possibly take up to 21 days from a positive result before variant data is available in public repositories.
With the addition of genotyping, labs can classify a high percentage of positive samples and provide information within two to three days. They can also reduce costs for strain subtyping as compared to NGS. Samples that cannot be assigned to a known variant through the genotyping approach become prime candidates to detect new or emerging variants more rapidly. 
“As COVID-19 evolves with new variants constantly emerging, surveillance will play a critical role as we learn to live with the virus,” said Dr. Manoj Gandhi, senior medical director of genetic testing solutions, Thermo Fisher.
“Our approach makes it possible to conduct surveillance testing on a significantly larger number of samples and quickly determine which variant is present in a sample,” said Dr. Eric Lai, co-lead investigator of the NIH RADx initiative. “If we start seeing a trend where we are unable to assign lineages using known markers in the genotyping assays, it is an early warning that a new variant may be emerging and helps determine which samples should be sent for sequencing. This is significant because emerging variants have the potential to evade vaccines, cause diagnostic test performance issues, or cause more severe disease.”
A recent paper outlines the performance of this method for identifying markers and assigning lineages to several SARS-CoV-2 variants, including previously characterized Variants of Concern (VOC) such as Alpha, Beta, Gamma and Delta. In addition, the study describes how this approach can be rapidly adapted to develop a targeted panel for currently circulating strains, such as Delta and Omicron, to allow for large-scale surveillance in routine testing laboratories.
The rapid selection and validation of highly sensitive and specific biomarkers for SARS-CoV-2 variants was enabled with the ROSALIND platform during the collaborative initiative. This dynamic, cloud-based data analytics platform was able to act as a centralized tool for labs by aggregating and analyzing the data in a real-time dashboard format. Thermo Fisher has updated the TaqMan SARS-CoV-2 Mutation Panel* to include the four markers that can detect the Omicron and Delta variants with high precision. 
“Our challenge has been staying in front of this pandemic at the rate the virus is evolving. We’ve demonstrated an approach for maintaining highly accurate markers, rapidly implementing marker updates in manufacturing, and quickly deploying these assays to nationwide testing labs for validating COVID-19 positivity with lineage assignment,” said Tim Wesselman, CEO of Rosalind. “With a public tracking dashboard, this automated analysis, classification, and real-time genotyping reporting provides a snapshot of the most current strains circulating the country and provide us with valuable information to respond more quickly.”
“A robust early warning surveillance capability will eventually incorporate multiple layers of novel technologies,” said Dr. David Becker, vice president of quality at Helix and co-investigator of the program. “We were excited to explore, understand, and validate this technology that enables rapid reporting and increased surveillance of the SARS-CoV-2 variants in circulation."
*For research use only. Not for use in diagnostic procedures.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue of approximately $40 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, increasing productivity in their laboratories, improving patient health through diagnostics or the development and manufacture of life-changing therapies, we are here to support them. Our global team delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services, Patheon and PPD. For more information, please visit www.thermofisher.com.
Media Contact Information:
Mauricio Minotta 
760-929-2456 
Mauricio.minotta@thermofisher.com
SOURCE Thermo Fisher Scientific
If you have any questions regarding this Press Release, please email us at media.relations@thermofisher.com.