Whole Allergen
w15 Scale/Lenscale
w15 Scale/Lenscale Scientific Information
Type:
Whole Allergen
Display Name:
Scale/Lenscale
Family:
Amaranthaceae (Chenopodiaceae)
Latin Name:
Atriplex lentiformis
Other Names:
Lenscale, Scale, Salt Bush, Saltbrush, Quail-Brush, Quailbush
Route of Exposure
Other topics
Lenscale is native to the temperate and tropical regions of North America, but a few species are grown as ornamentals throughout the world because of their attractive greyish foliage. The different Atriplex species are closely related to each other and consist of annual and perennial weeds and shrubs.
Lenscale is a fast-growing, compact, woody perennial shrub usually growing between 0.15 and 1.52m in height, but sometimes reaching 3m. The plant is deciduous in arid areas but tends to be evergreen elsewhere. Numerous slender and wide-spreading branches contain grey-green, thick leaves 1.0 to 5.0cm long and 0.5 to 4.0cm wide, with a fine, scaly surface.
Lenscale produces inconspicuous yellow flowers from May to August. It is not yet clear whether the flowers are primarily dioecious (plants are male or female) or monoecious (individual flowers are either male or female, but both sexes can be found on the same plant). The plant is wind-pollinated and greatly contributes to the pollen loads of arid regions. These plants have the ability to alter their sexual state from one season to the next in response to environmental conditions. Many seeds are borne on bracts and wind dispersed. The fruits and seeds mature from September to October.
Lenscale often occurs along seashores and in other saline soils, especially in arid regions, and has been used for windbreaks, borders, and range management.
All parts of the plant are edible. Native Americans ground this plant’s seeds into meal for piñole or porridge, and for an emetic.
Clinical Relevance
IgE mediated reactions
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis are common following exposure to pollen from Lenscale; however, no specific studies have been reported to date. (1)
Molecular Aspects
No allergens from this plant have yet been characterized.
Cross-reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected, as well as to a certain degree among members of the subfamily Chenopodiaceae. (2)
Atriplex latifolia, Beta vulgaris, Salsola kali and Amaranthus retroflexus were compared with an extract from Chenopodium album by both in vivo and in vitro methods. The presence of common allergenic determinants was suggested. This implied that as A. latifolia is cross-reactive with A. lentiformis, cross-reactivity might occur between this plant and other members of the Amaranthaceae. (3)
Compiled By
Last Reviewed-  April 2022
References
  1. Larenas Linnemann D ACA, Guidos Fogelbach GA, Cid del Prado ML. Allergens used in skin tests in Mexico. [Spanish]. Rev Alerg Mex. 2009;56(2):41-7.
  2. L. Y. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. Pharmacia Diagnostics AB Uppsala Sweden. 1982.
  3. Lombardero M, Duffort O, Sellés JG, Hernández J, Carreira J. Cross-reactivity among Chenopodiaceae and Amaranthaceae. Annals of allergy. 1985;54(5):430-6.