Type:
Whole Allergen
Whole Allergen
Whole Allergen
Octopus
Octopodidae
Octopus vulgaris
Octopus
IgE antibodies have been measured in sera from Japanese patients with atopic dermatitis (1).
Octopus vulgaris is a cephalopod mollusk of the family Octopodidae, an octopus with eight suckered arms and a round mouth. IgE antibodies have been measured in sera from Japanese patients with atopic dermatitis (1). Also measured in children with allergy (2). Sensitivity to octopus is also common in southern Europe (3). In a study using Pharmacia CAP System, octopus and squid showed a low degree of correlation with each other (4).
Octopus and squid showed a low degree of correlation with each other (4). IgE antibodies to octopus were also measured in children with allergy (2).
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.