Type:
Whole Allergen
Whole Allergen
Whole Allergen
Redtop, Bentgrass
Inhalation
Poaceae
Agrostis stolonifera
Agrostis stolonifera
Creeping bentgrass
Agrostis stolonifera is a member of the Poaceae family that produces grass pollens. Poaceae family pollen is a leading airborne biological pollutant and the main cause of pollen allergy worldwide. Cross-reactivity between grass pollen protein and fresh fruit pan-allergens is associated with allergies to foods such as beans, peas, cereals, peanut, melon, watermelon, carrot, and celery.
A. stolonifera is a self-pollinating cool season grass species (1).
Poaceae family pollen is a leading airborne biological pollutant and the main cause of pollen allergy worldwide (Garcıa-Mozo, 2017). In Europe, 20% of the population are affected by grass pollen allergy, while 15% of US individuals ate sensitized to grass pollen (2). Marked regional variations of grass pollen sensitization are seen between and within individual countries (2).
Taxonomic tree of Agrostis stolonifera (3) |
|
Domain |
Eukaryota |
Kingdom |
Plantae |
Phylum |
Spermatophyta |
Subphylum |
Angiospermae |
Class |
Monocotyledonae |
Family |
Poaceae |
Genus |
Agrostis |
Grass-induced pollinosis is the most common pollen allergy in Europe and worldwide (4). Marked regional variations of grass pollen sensitization are seen between and within individual countries (2); depending on climate and region, global sensitization rates range from 1–30%. In Germany, 14.8% of adults are affected by hay fever (4).
Inhalation (1).
Symptoms of grass pollen sensitization include allergic rhinoconjunctivitis and bronchial asthma.
Table adapted from Allergome.org (5).
Allergen |
Type |
Mass (kDa) |
Expansin |
30 |
|
Berberine bridge enzyme |
60 |
|
Ribonuclease |
- |
Grass group 1 and 5 allergens are considered immunodominant major Pooideae pollen allergens (6). Group 1 allergens are the most clinically relevant, followed by group 5 allergens (7).
Cross-reactivity between grass pollen protein and fresh fruit pan-allergens is associated with food allergies (2); foods including beans, peas, cereals, peanut, melon, watermelon, carrot, and celery have been shown to cross react with Poaceae pollen (7).
In addition to cross reactivity among Poaceae species (7), high cross-reactivity is seen between grass and cereal pollen allergens (1).
Author: RubyDuke Communications
Reviewer: Dr. Christian Fischer
Last reviewed:April 2022