Whole Allergen
g2 Bermuda grass
g2 Bermuda grass Scientific Information
Type:
Whole Allergen
Display Name:
Bermuda grass
Allergen code:
g2
Family:
Poaceae (Chloridoideae)
Species:
dactylon
Route of Exposure:
Inhalation
Source Material:
Pollen
Latin Name:
Cynodon dactylon
Other Names:
Couch Grass, Scutch grass, Wire grass, Star grass, Bahama grass, Devil grass
Synonyms:
Panicum dactylon
WHO/ICD-11 code:
XM5ZN2
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Bermuda grass is a perennial grass that grows in the warm-season and used globally as lawn grass, forage grass, and for soil protection. It has scaly rhizomes and flat stolon, cultivated mainly in the tropical and subtropical regions of the world. It can grow on any type of soil such as alkaline soils, in drought conditions, high temperature, intensive sunlight. It belongs to the genus Cynodon, family Poaceae, and subfamily Chloridoideae. The pollen grain is round to oval.  
It can be found throughout the world. A Filipino study reported Bermuda grass to be one of the most common outdoor allergens.
A study in Kuwait reported a prevalence rate of 55% sensitization to Bermuda grass by allergic rhinitis patients aged 6-64 years.
The Bermuda grass allergen is associated with asthma, allergic rhinitis, atopic dermatitis, sinusitis and conjunctivitis. A study conducted at Saudi University reported the prevalence of physician-diagnosed allergic diseases to be asthma (27%), atopic dermatitis (13.1%), and allergic rhinitis (5%).
Bermuda grass pollen has seven allergens and the major allergen is Cyn d 1. Mutual sensitization to profilin and/or polcalcin is linked with a longer duration of allergic disease and to co-sensitization to a larger number of species-specific allergen molecules. Partial cross reactivity with Bermuda grass can be seen with Timothy grass and maize species. 
Allergen
 Nature
Bermuda grass is a perennial grass having horizontal shoots and the leaves coming off the stem, interchangeably. The bud contains 3-7 purple colored flowering spikes. It is planted in lawns, but the runners are very robust and are hard to manage and invade flowerbeds (1). The flowering period is usually between May and September (2).
Habitat
Bermuda grass grows in warmer regions. Although, it is immune to very high temperatures, yet is prone to hard or persistent frost. It is exclusively prevalent as a weed in subtropical conditions in both annual and perennial crops and in pastures, fallows and waste areas. It occurs in a wide range of soil types under semi-arid and irrigated conditions with varying pH and salinity. It can be easily found in waste places, agriculture fields, irrigated ditches, streambeds and also on the sides of the roads (1, 3).
Taxonomy
Epidemiology
Worldwide distribution
Bermuda grass is a native perennial turfgrass of the Mediterranean region but can be found throughout the world. A recent study conducted on adult Filipinos with allergic rhinitis (AR) stated Bermuda grass (67%) to be the most common allergen present in the environment (7). Similarly, Botswana also has the highest aeroallergen sensitization of Bermuda grass (41%) and it was found to be significantly associated with conjunctivitis (p< 0.001) and rhinitis (p< 0.01) (8).
It is practically suitable for every ecosystem due to its eco-physiological and genetic traits coupled with both forms of propagation. Ontario had also reported high sensitization rates for Bermuda grass (9). It has also contributed to pollen rain in the semiarid environment of Tucson, Arizona and increases the risk of AR and asthma (10). A survey conducted by the Orange County Society of Allergy and Clinical Immunology in California also reported the highest (85%) positive rate for Bermuda pollens (11). 
Risk factors
Pollens from flowers of Bermuda grass are hardly visible to the naked eye but can lead to major allergenic blow (12). Grass pollen sensitization imposes a threat of allergic respiratory diseases across the world due to its airborne nature. This pollen accounts for almost half of immunoglobulin E (IgE) related to allergies (13). Primarily, it leads to high production of allergen-specific IgE and on secondary sensitization, it leads to cross-reactivity (14). 
Pediatric issues
Taiwan reported Bermuda grass to be highly sensitized pollen among asthmatic children (8%) (15). Bermuda grass was also considered to be the most prevalent allergen among children with AR (16).
Environmental Characteristics
Worldwide distribution
It is found in all of Europe except Scandinavia, in the USA, South Africa, Australia, India, and Japan. The origin is Africa but it is found throughout the tropical and subtropical regions including Asia, North, Central and South America, the Caribbean, and islands in the Pacific Ocean (5). It is the most common tropical lawn grass and is sensitive to extended frost. It is mostly used as fodder in Brazil. In the Southern US, it is considered as an imperative pasture grass. Commercial cultivations are done for several genotypes and hybrids of Bermuda grass and over one thousand accessions are now offered at the U. S. National Plant Germplasm System (3). 
Route of Exposure
Main
Exposure to Bermuda grass pollens may occur due to inhalation or skin contact. Changes in climatic conditions, pollen-derived nanovesicles and other small components have an explicit role in the development of allergic diseases (17).
Clinical Relevance
Allergic rhinitis
Allergic rhinitis (AR) is a heterogeneous disease. A 19-year retrospective study conducted between 1998 and 2017, reported a statistically significant increase in Bermuda pollen sensitization from 13.64% in 1998 to 30.9% in 2017 (18).
A study conducted on 706 AR patients aged 6-64 years in Kuwait, reported a prevalence rate of 55% sensitization to Bermuda grass (19). Similarly, a study conducted among the Thai population (n=100) suffering from AR, reported 17% of the patients sensitive to Bermuda pollens (20).
In northern New South Wales, Australia, an association between Bermuda grass and AR in adults was reported (21). 
Asthma
Pollens of different subtropical grasses, including Bermuda grass causes AR and worsens the condition of asthma. AR and asthma patients show higher allergic sensitivity and subfamily-specific immunoreactivity with pollen of subtropical grasses, compared with temperate grass pollens. This can be found across subtropical regions including Australia, Africa, India, and the USA (22).
In Southern China, positive tests for Bermuda grass specific IgE (sIgE) was reported for 21.8% of patients with AR, 22.8% for patients with asthma, and 22.7% for patients with both AR and asthma (23)
In Spain from 101 patients with asthma, 70% were found to be highly sensitive to Bermuda grass (24). 
Atopic Dermatitis
Students at Saudi University reported the prevalence of physician-diagnosed allergic diseases to be 27% for asthma, 13.1% for atopic dermatitis, and 5% for AR. The skin prick test was found to be positive for 20.8% of students for Bermuda grass (25). 
Other diseases
Bermuda pollens that come along with sandstorm dust also triggers various airway illnesses. Reports also stated that Bermuda pollens are associated with sinusitis (26). 
Prevention and Therapy
Allergen immunotherapy
Allergen immunotherapy may recover the patient from symptoms, reduce medication requirements, and improve quality of life. It can be of two types- Subcutaneous injection immunotherapy (SCIT) and Sublingual Immunotherapy (SLIT) (27). 
Prevention strategies
Avoidance
Avoidance from allergen is highly effective in seasonal allergic rhino-conjunctivitis patients. Aggravators such as smoke, traffic pollution should also be avoided as it may worsen AR. Other preventive strategies include wearing sunglasses, nasal filters, usage of balms and ointments for the nose. Minimizing outdoor activity when pollen is highest, avoiding going out during/after thunderstorms, pollen season, shutting windows and showering after high exposure (27). 
Molecular Aspects
Allergenic molecules
Bermuda pollens contain seven allergens. It also contains species-specific allergen. Among the following, Cyn d 1 is the major allergen as its reactivity with IgE is the highest (22, 28).
  • Cyn d 1 (a major Group 1 allergen, consists of 11 isoforms; Beta-expansin; 32 kDa),
  • Cyn d 7 (Polcalcin, calcium binding protein; 12 kDa),
  • Cyn d 12 (Profilin; 14.5 kDa),
  • Cyn d 15, (9 kDa),
  • Cyn d 22 (enolase),
  • Cyn d 23 (9 kDa),
  • Cyn d 24 (Pathogenesis-related protein PR-1; 21 kDa),
The major allergen found in Bermuda pollen is Cyn d 1. It can be found in 76-100% of patients allergic to grass pollens. Increased levels of IgE to Cyn d 1 can be found at primary sensitization (17).
Cyn d 1 allergen may occur as a single 34 kDa protein or as a mixture of 34 and 29 kDa proteins. It contains two isoallergens known as Cyn d 1a and b. Cyn d 1a is a 32 kDa protein, and Cyn d 1b is a 31 kDa protein. Around 50% of Bermuda sIgE shows reactivity to Cyn d 1 whereas only 10% shows to Cyn d 7 (22, 29).
Mutual sensitization to profilin and/or polcalcin is linked with a longer duration of allergic disease and co-sensitization to a larger number of species-specific allergen molecules (30). 
Biomarkers of severity
Serum specific IgE antibodies to nCyn d 1 is a molecular specific biomarker of genuine sensitization to Chloridoideae grass pollen i.e. Bermuda grass (30). Cyn d 1 is the most abundant pollen in Bermuda grass, concluding to 15% of the whole-pollen extract. The frequency of sensitization is from 76-100% (30). As it is immunologically different from Phl p 1 of Timothy grass, it is therefore a suitable marker for sensitization to Cynodon dactylon (30). Moderately higher levels of IgE specific to nCyn d 1 is considered as a marker of sensitization to Bermuda grass (30).
Cross-reactivity
Bermuda grass is comprised of widely distributed and genetically diverse nine species. The strongest genetic similarities were found between species C. aethiopicus, C. arcuatus, C. transvaalensis, C. plectostachyus, C. incompletus and C. nlemfuensis. Cross-reactivity had been reported between giant Bermuda grass extract and Bermuda grass (31).
There is a  cross-reactivity between Phl p 1 of timothy grass and Cyn d 1 of Bermuda grass that may obstruct the identification of the allergen source causing sensitization (30).
The close allergenic relationship is reported between Salt and Grama to Bermuda grass. Johnson grass also shares some allergenicity with Bermuda grass (32).
A study conducted to determine the degree of antigenic and IgE-binding similarities found the lowest similarity between Maize (23%) and Bermuda (10%) (33).
Bermuda grass has a low degree of cross-reactivity with Kikuyu grass (34). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: October  2020
References
  1. S.K. Nitu, S.M.S. Islam, Tarique MH. Morphological characteristics of different accessions of Cynodon dactylon (L.) Pers. and physico-chemical properties of soil of their growing region in Bangladesh. Int J Biosci. 2019;15(5):350-69.
  2. Roseler STM, Baron JM, Hoflich C, Merk HF, Bas M, Bier H, et al. "New" inhalant plant allergens. Allergol Select. 2020;4:1-10.
  3. Huang S, Jiang S, Liang J, Chen M, Shi Y. Current knowledge of bermudagrass responses to abiotic stresses. Breed Sci. 2019;69(2):215-26.
  4. Fang T, Dong H, Yu S, Moss JQ, Fontanier CH, Martin DL, et al. Sequence-based genetic mapping of Cynodon dactylon Pers. reveals new insights into genome evolution in Poaceae. Commun Biol. 2020;3(1):358.
  5. Miaoli Wang, Jingxue Zhang, Zhipeng Guo, Yongzhuo Guan, Gen Qu, Jianyu Liu, et al. Morphological variation in Cynodon dactylon (L.) Pers., and its relationship with the environment along a longitudinal gradient. BMC. 2020;157(4):1-11.
  6. Davies JM, Li H, Green M, Towers M, Upham JW. Subtropical grass pollen allergens are important for allergic respiratory diseases in subtropical regions. Clin Transl Allergy. 2012;2(1):4.
  7. Navarro-Locsin CG, Lim-Jurado M. Aeroallergen sensitization and associated comorbid diseases of an adult Filipino population with allergic rhinitis. Asia Pac Allergy. 2018;8(3):e25.
  8. Shiang-Ju Kung, Loeto Mazhani, Steenhoff AP. Allergy in Botswana. J Allergy Clin Immunol. 2013;26(4):202-9.
  9. Rawls M, Thiele J, Adams DE, Steacy LM, Ellis AK. Clinical symptoms and biomarkers of Bermuda grass-induced allergic rhinitis using the nasal allergen challenge model. Ann Allergy Asthma Immunol. 2020;124(6):608-15 e2.
  10. Sneller MR, Hayes HD, Pinnas JL. Pollen changes during five decades of urbanization in Tucson, Arizona. Ann Allergy. 1993;71(6):519-24.
  11. Chiu JT, Nelson B, Sokol W, Ellis M, Galant S, Novey HS. Extended evaluation of diagnostic skin testing practices in Orange County, California. Allergy Proc. 1993;14(4):283-6.
  12. Schmidt CW. Pollen Overload: Seasonal Allergies in a Changing Climate. Environ Health Perspect. 2016;124(4):A70-5.
  13. Luo W, Pan G, Huang H, Zheng P, Wei N, Zhang Y, et al. A Component-resolved Diagnostic Approach for a Study on Grass Pollen Allergens in Chinese Southerners with Allergic Rhinitis and/or Asthma. J Vis Exp. 2017(124).
  14. Babak Aberumand, Matthew Rawls, Daniel E. Adams, Lisa M. Steacy, Ellis AK. Possible sources of Bermuda grass sensitization in Timothy grass allergic subjects. Ann Allergy Asthma Immunol. 2020.
  15. Chou TY, Wu KY, Shieh CC, Wang JY. The clinical efficacy of in vitro allergen-specific IgE antibody test in the diagnosis of allergic children with asthma. Acta Paediatr Taiwan. 2002;43(1):35-9.
  16. Halonen M, Stern DA, Wright AL, Taussig LM, Martinez FD. Alternaria as a major allergen for asthma in children raised in a desert environment. Am J Respir Crit Care Med. 1997;155(4):1356-61.
  17. Pablos I, Wildner S, Asam C, Wallner M, Gadermaier G. Pollen Allergens for Molecular Diagnosis. Curr Allergy Asthma Rep. 2016;16(4):31.
  18. Pongsakorn Tantilipikorn, Bannapuch Pinkaew, Kanokporn Talek, Paraya Assanasen, Triphoon Suwanwech, Bunnag C. Pattern of allergic sensitization in chronic rhinitis: A 19-year retrospective study. Asian Pac J Allergy. 2020.
  19. Dowaisan A, Al-Ali S, Khan M, Hijazi Z, Thomson MS, Ezeamuzie CI. Sensitization to aeroallergens among patients with allergic rhinitis in a desert environment. Ann Allergy Asthma Immunol. 2000;84(4):433-8.
  20. Pumhirun P, Towiwat P, Mahakit P. Aeroallergen sensitivity of Thai patients with allergic rhinitis. Asian Pac J Allergy Immunol. 1997;15(4):183-5.
  21. Bass DJ, Delpech V, Beard J, Bass P, Walls RS. Late summer and fall (March-May) pollen allergy and respiratory disease in Northern New South Wales, Australia. Ann Allergy Asthma Immunol. 2000;85(5):374-81.
  22. Davies JM. Grass pollen allergens globally: the contribution of subtropical grasses to burden of allergic respiratory diseases. Clin Exp Allergy. 2014;44(6):790-801.
  23. Luo W, Huang H, Zheng P, Wei N, Luo J, Sun B, et al. Major grass pollen allergens and components detected in a southern Chinese cohort of patients with allergic rhinitis and/or asthma. Mol Immunol. 2016;78:105-12.
  24. Martinez Ordaz VA, Rincon Castaneda CB, Lopez Campos C, Velasco Rodriguez VM. [Cutaneous hypersensitivity in patients with bronchial asthma in La Comarca Lagunera]. Rev Alerg Mex. 1997;44(6):142-5.
  25. Alqahtani JM. Atopy and allergic diseases among Saudi young adults: A cross-sectional study. J Int Med Res. 2020;48(1):300060519899760.
  26. Lombardi E, Stein RT, Wright AL, Morgan WJ, Martinez FD. The relation between physician-diagnosed sinusitis, asthma, and skin test reactivity to allergens in 8-year-old children. Pediatr Pulmonol. 1996;22(3):141-6.
  27. Scadding GK, Kariyawasam HH, Scadding G, Mirakian R, Buckley RJ, Dixon T, et al. BSACI guideline for the diagnosis and management of allergic and non-allergic rhinitis (Revised Edition 2017; First edition 2007). Clin Exp Allergy. 2017;47(7):856-89.
  28. WHO/IUIS. Allergen Nomenclature. Allergen Nomenclature Sub-Committee 2020 [15-10-2020]. Available from: http://www.allergen.org/search.php?allergenname=&allergensource=Cynodon+dactylon&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
  29. Smith PM, Avjioglu A, Ward LR, Simpson RJ, Knox RB, Singh MB. Isolation and characterization of group-I isoallergens from Bermuda grass pollen. Int Arch Allergy Immunol. 1994;104(1):57-64.
  30. Popescu F-D. Molecular biomarkers for grass pollen immunotherapy. World J Methodol. 2014.
  31. Assefa S, Taliaferro CM, Anderson MP, de los Reyes BG, Edwards RM. Diversity among Cynodon accessions and taxa based on DNA amplification fingerprinting. Genome. 1999;42(3):465-74.
  32. Martin BG, Mansfield LE, Nelson HS. Cross-allergenicity among the grasses. Ann Allergy. 1985;54(2):99-104.
  33. Suphioglu C, Singh MB, Knox RB. Peptide mapping analysis of group I allergens of grass pollens. Int Arch Allergy Immunol. 1993;102(2):144-51.
  34. Potter PC, Mather S, Lockey P, Ainslie G, Cadman A. IgE specific immune responses to an African grass (Kikuyu, Pennisetum clandestinum). Clin Exp Allergy. 1993;23(7):581-6.