Recent searches Clear History
Fale conosco
Phadia Prime Operator Software
Enhanced productivity through optimized workflows.
Phadia™ instruments are powered by Phadia™ Prime software. Phadia Prime makes it easy to manage assay runs, test requests, results, and management of large databases. It also provides automated, real-time management of reagent inventory. Phadia Prime provides connectivity to Laboratory Information System (LIS), Laboratory Automation System (LAS), and Phadia™ LabCommunity that allows you to optimize laboratory and information workflows.
 Result reporting
  • Intuitively monitor results and quickly interpret data. 
  • Manual or auto-verification of results.
  • Generate customized reports to aid in the interpretation of ImmunoCAP sIgE test results. 
  • Extended event log reporting. 
  • Fast and secure transfer of test result. 
 Test management
  • Centralized control and monitoring of requests. 
  • Manage outsourced tests.
  • Test prioritization by adult/pediatric samples. 
  • Enhanced sample management, including quantity-not-sufficient (QNS) handling. 
  • User-configurable reflex testing.
 Stock management 
  • Real-time management of reagent inventory and automated shelf-life control.
  • Possibilities for ERP (Enterprise Resource Planning) integration.
  • Calculate required consumables.
 System integration
  • Manage several connected Phadia instruments with one Phadia Prime computer. 
  • Manage configurations and settings. 
  • Access event log and quality information. 
  • Automatic database backup. 
 Connectivity
  • Easy integration with Laboratory Information Systems (LIS).
  • Connect to Phadia LabCommunity to experience the enhanced support and troubleshooting. 
  • Manage LAS connectivity of capable Phadia instruments
 Help function
  • Integrated electronic manuals for easy access. 
View the video
To learn more about Phadia Prime, contact your local representative.
Contact Us
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.