Whole Allergen
e74 Rat urine proteins
e74 Rat urine proteins Scientific Information
Type:
Whole Allergen
Display Name:
Rat urine proteins
Route of Exposure:
Inhalation
Family:
Muridae
Species:
Rattus spp.
Latin Name:
Rattus
Other Names:
Rat
Summary
Urine proteins of rodents, including Rattus spp. (rat), are inhalant allergens that cause respiratory symptoms in sensitized individuals. In addition to being common pets and ubiquitous pests, rats are commonly used for scientific research. As such rat urine protein allergens are an important cause of allergic respiratory disease, particularly occupational disease in laboratory workers.
Allergen
Nature
In addition to being common urban and rural pests, rats are kept in homes as pets and in laboratories as animal models in scientific research (1); in 2014, rats accounted for 7% of the 3.8 million animals used for scientific research in Great Britain (2).
The sensitizing agents in rats are proteins found in their urine and saliva (2). These proteins contaminate their fur and fomites and are readily aerosolized by husbandry practices (2).
Urine is such a potent source of rat allergens because rats have permanent proteinuria (males have particularly high protein levels in their urine) and perform behavioural enurination. When the urine dries, proteins associate with airborne dust particles and can be inhaled, leading to sensitization. (3).
Reported prevalence rates of allergies in laboratory technicians working with laboratory animals range from 11% to 30% (4).
Taxonomy 
Taxonomic tree of Rattus (5)
Domain
Eukaryota
Kingdom
Metazoa
Phylum
Chordata
Subphylum
Vertebrata
Class
Mammalia
Order
Rodentia
Family
Muridae
Genus
Rattus
Epidemiology
Worldwide distribution 
Exposure to laboratory animal allergens is a significant cause of IgE-mediated occupational allergy and asthma in the pharmaceutical sector, contract toxicology laboratories, research institutes and universities (2); reported prevalence rates range from 11% and 30% (4).
In a study of 101 laboratory animal technicians, 30% had symptoms (100% had rhinitis; 10% had concurrent bronchial asthma), and 19% had confirmed allergy (4). In the same study, 18% of laboratory workers exposed to rats, had confirmed allergy (4). All the rat-sensitized individuals had been working with mature male animals (4).
In a study of occupational sensitization to rat and/or mouse urinary allergens, out of 498 employees exposed to both rat and mouse at work and 220 exposed to rat only (6), 14% were sensitized to rats. 
Risk factors 
Exposure to rat urine in the workplace, especially research laboratories (2), or in the home (1).
Some handling and husbandry activities are associated with high exposures (2). For example, filtered cages may reduce exposure, while filter changing and/or cleaning can lead to high exposures (2). Equally, continued contact with animals may raise sensitivity; mild symptoms may serve as a precursor to a warning for the risk of developing bronchial symptoms (4).
A history of atopic disease; in one study, atopy was overrepresented in technicians with positive RAST and skin test reactions to the animal allergens compared with technicians without positive tests (4).
Route of Exposure
Main 
Inhalation (3).
Secondary 
Contact (4).
Clinical Relevance
Symptoms of sensitization to rat urine proteins include rhinorrhea, nasal congestion, sneezing, conjunctival itching, cough, dyspnea, wheezing (4) and, rarely, anaphylaxis (2). Itching and skin weals may also occur at contact sites (4).
Molecular Aspects
Allergenic molecules
The following allergens have been identified from rat urine and serum (table adapted from Allergome.org (7)).
Allergen
Type
Mass (kDa)
Rat n 1
Lipocalin (α2u-globulin)
21
Rat n 4
Albumin
69
Rat n 7
Immunoglobulin; IgG
94
Rat n Transferrin
Serotransferrin
76
Cross-reactivity
In a study of occupational sensitization to rat and/or mouse urinary allergens, 62% of rat-sensitized individuals were also mouse-sensitized, while 91% of mouse-sensitized individuals were also rat-sensitized (6).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr.Christian Fischer
Last reviewed: June 2022
References
  1. Munera M, Contreras N, Sánchez A, Sánchez J, Emiliani Y. In silico analysis of a major allergen from Rattus norvegicus, Rat n 1, and cross-reactivity with domestic pets. F1000Res. 2019;8:1707.
  2. Mason H, Willerton L. Airborne exposure to laboratory animal allergens. AIMS Allergy and Immunology. 2017;1(2):78-88.
  3. Schulten V, Westernberg L, Birrueta G, Sidney J, Paul S, Busse P, et al. Allergen and Epitope Targets of Mouse-Specific T Cell Responses in Allergy and Asthma. Front Immunol. 2018;9.
  4. Agrup G, Belin L, Sjöstedt L, Skerfving S. Allergy to laboratory animals in laboratory technicians and animal keepers. British Journal of Industrial Medicine. 1986;43(3):192-8.
  5. CABI. Rattus (rats) Wallingford, UK 2019. Available from: https://www.cabi.org/isc/datasheet/46827.
  6. Jeal H, Harris J, Draper A, Taylor AN, Cullinan P, Jones M. Dual sensitization to rat and mouse urinary allergens reflects cross-reactive molecules rather than atopy. Allergy. 2009;64(6):855-61.
  7. Allergome. Rat urine proteins. 2022.