These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Pernicious anemia (PA) is the final stage of autoimmune gastritis and characterized by vitamin B12 deficiency.1 Its definitive diagnosis is important because the patients need lifelong parenteral vitamin B12 treatment.2
PA is associated with two autoantibodies that can be used for diagnostic testing:1
The simultaneous determination of antibodies to parietal cells and those to intrinsic factor create a higher probability in the diagnosis of PA.1,2
With an exceptionally high clinical specificity of 99.4% and 89.9%, respectively, combined with a clinical sensitivity of 51.9% and 81.0%, respectively, EliA™ Intrinsic Factor test and EliA™ Parietal Cell test form the best-in-class, fully automated portfolio to aid in the diagnosis of PA.3
Combining the outstanding sensitivity and specificity of the EliA Intrinsic Factor and EliA Parietal Cell test increases the clinical sensitivity to 91.1% while maintaining specificity at 87%, helping you put the patient first by detecting more patients suffering from PA, sooner.3
EliA Intrinsic Factor test and EliA Parietal Cell test can be measured from one sample in one run and on the same fully automated Phadia™ Laboratory Systems.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.