Table of Contents
Component
f440 Cor a 9
f440 Cor a 9 Scientific Information
Type:
Component
Name; WHO/IUIS:
Cor a 9
Biological function:
11S seed storage globulin – legumin-like protein
Allergen code:
f440
Source Material:
Seed storage proteins Native sourced from the extracts of Corylus avellana
Other Names :
Corylin
Summary
Cor a 9 is a thermoresistant storage protein of the 11S globulin family, found in hazelnut (Corylus avellana). The allergenic acidic polypeptide chain of Cor a 9 possesses a molecular weight of 40 kDa. Cor a 9 sensitization among hazelnut-sensitized individuals has been described in Central, Southern and North-Western Europe as well as in the United States. Age-related variations in prevalence, with greater IgE-reactivity towards Cor a 9 in children than in adults, are reported. Cor a 9 possesses two important features: its ability to induce direct (primary) sensitization to hazelnut, as opposed to pollen-related sensitization, and its association with more severe clinical reactions (skin, respiratory, gastrointestinal, or even anaphylaxis). Determination of specific immunoglobulin E (sIgE) for Cor a 9 can serve as a predictive marker for clinical allergy to hazelnut with severe symptoms and could differentiate between sensitization and clinical allergy. However, the predictive ability of Cor a 9-sIgE was found to be lower than that of Cor a 14-sIgE. Combined assessment of Cor a 9 and Cor a 14 sensitization results in improved diagnostic accuracy for hazelnut allergy. Even though high sequence homology of Cor a 9 exists with several other 11S globulins, such as walnut, mustard, soybean, peanut, Brazil nut, almond, cashew nut, and pistachio, clinical cross-reactivity may not be present. Co-sensitization of Cor a 9 has been reported with 11S globulins like Jug r 4 (walnut) and Sin a 2 (mustard) as well as with 2S albumins like Jug r 1 and Ana o 2 (cashew nut). 
Epidemiology
Worldwide distribution
Hazelnut allergy (Corylus avellana) is the most common tree nut allergy in Europe (1, 2). According to a systematic review, this allergy accounts for about 17%-100% of the total tree nut allergies in Europe (3). Hazelnut allergy as well as sensitization are also frequent outside Europe, in countries such as India, Russia, Israel, Australia, Mexico and South Africa (4-9).
Cor a 9 is a seed storage protein of the 11S albumin family (10). In the EuroPrevall study conducted from 2005 to 2010 among 731 hazelnut-sensitized individuals in 12 European centers (central, eastern, north-western and southern countries), sensitization to Cor a 9 exhibited an overall prevalence of 9.9%. The highest prevalence was noted (18.2%-27.3%) in some cities of Central Europe (Prague [Czech Republic], Utrecht [The Netherlands], and Manchester [United Kingdom], Southern Europe (Madrid [Spain]) and North-western Europe (Reykjavik [Iceland]), while the prevalence in other cities of Central and Southern Europe was found to be <7% (11).
In the Netherlands, sensitization to Cor a 9 was found to be 35% in hazelnut-sensitized patients (81 children and 80 adults) (12), while in Sweden among 220 hazelnut-allergic and asthmatic children (10-≤18 years) and young adults (>18-35 years), sensitization to Cor a 9 was 14.1% (31 out of 220) (13). On the other hand, a study conducted in Turkey reported 72.5% (37 out of 51) prevalence of Cor a 9 sensitization, among hazelnut-sensitized children having a median age of 3.4 years (14).
Similarly, the prevalence of Cor a 9 among hazelnut-sensitized patients ranged from 48% to 59% in different regions of the United States (US). It was further noted that the prevalence rates decreased with increasing age (15).
Age-related variations in prevalence, with greater IgE-reactivity towards Cor a 9 in children than in adults, were reported in a few studies. In Belgium, sensitization to Cor a 9 was reported to be 80% in preschool children, 70% in school-age children and 43% in adults. Furthermore, 28% of infants with atopic dermatitis were also found to be sensitized to Cor a 9 (16). A similar pattern was reported in a study conducted in Dutch children and adults, where sensitization to Cor a 9 was reported to be highest in pre-school children (65%), followed by school-going children (50%) and the adults (17%), showing severe reactions to hazelnut (17).
Environmental Characteristics
Source and tissue
Cor a 9 is present in the hazelnut, the seed of hazel tree (Corylus avellana). It is a thermo resistant storage protein of the 11S globulin family (10).
Ion chromatography and hydrophobic chromatography (reverse-phase) were used for purification of natural Cor a 9 from defatted hazelnut extract (17).
Clinical Relevance
Disease severity
Similar to other nut storage proteins, Cor a 9 possesses two important features: its ability to induce direct (primary) sensitization to hazelnut, as opposed to pollen-related sensitization, and its association with more severe clinical reactions (skin, respiratory, gastrointestinal [GI], or even anaphylaxis) (10, 18-20). As an example, the Europrevall study conducted among 731 hazelnut-sensitized individuals demonstrated a statistically significant association of Cor a 9 sensitization (p=0.025) with the prevalence of moderate-to-severe clinical presentations as stated above. In this study, Cor a 9 sensitization was present in 18.3% of patients with severe symptoms, 10.2% with moderate symptoms, and 7.2% with OAS (18). Also, the association of Cor a 9 sensitization with severe allergic reactions or anaphylaxis was found to be 71.4% in a study conducted among 227 children with severe allergic reactions or anaphylaxis (21). Similar data were collected in a study conducted in Belgium among 119 hazelnut-sensitized patients, with Cor a 9 sensitization significantly more prevalent in patients with generalized reactions (erythema, urticaria, angioedema, GI symptoms, and respiratory symptoms) as compared to patients with OAS (70% vs. 11%, p<0.05) (16). In another study involving 220 hazelnut-allergic asthmatic patients, individuals displaying positive specific immunoglobulin E (sIgE) to Cor a 9 or Cor a 14 were found to report more generalized reactions, like GI symptoms or anaphylaxis as compared to those without Cor a 9 or Cor a 14 sensitization (13).
Further, sensitization to Cor a 9 was found to be associated with the presence of clinical allergy to hazelnut. This could be seen from few studies conducted in the Netherlands (n= 38), Denmark (n=155) and the US (n=42) that showed higher Cor a 9 sensitization in hazelnut challenge (double-blind placebo-controlled food challenge [DBPCFC])-positive patients than in challenge negative individuals (17, 22, 23). However, in a Turkish pediatric cohort, Cor a 9 sensitization was similarly frequent in hazelnut-sensitized children passing (19/26, 73.1%) or failing (18/25, 72%) an oral challenge (14).
Cross-reactive molecules
Clinical cross-reactivity may be expected among homologous seed storage proteins, such as 2S albumins, 7S globulins, 11S globulins or oleosins as they may share common IgE epitopes or due to sequence or structural similarity (24). Co-sensitization of Cor a 9 with the 2S albumins Jug r 1 (walnut) or Ana o 3 (cashew nut) may be present as observed in a study conducted among 10,503 hazelnut-sensitized individuals in the US. Moderate co-sensitization was reported between Cor a 9 and Jug r 1 of walnut (correlation coefficient=0.5) and Ana o 3 of cashew nut (correlation coefficient=0.5) (15).
A study conducted on 13 walnut and/or hazelnut-allergic patients revealed that walnut allergy or serological reactivity to walnut may place the patients at potential risk of developing hazelnut allergy due to cross-reactivity between their seed storage proteins i.e., Cor a 9, Cor a 14 and Jug r 1 (25). Further, co-sensitization of Cor a 9 with the 11S globulin of walnut (Jug r 4) was also observed in another study among 10 Jug r 4-sensitized subjects with walnut allergy (26).
Cor a 9 and peanut 11S globulin Ara h 3 are not found to be significantly cross-reactive, as demonstrated in a study investigating peanut and hazelnut molecular profiles in 114 hazelnut-allergic patients, of whom 48% were also confirmed as peanut-allergic. Co-sensitization between Cor a 9 and Ara h 3 was virtually absent in hazelnut-allergic patients without peanut allergy, however, reached up to ~10% in peanut-allergic patients with no hazelnut allergy, and to even 30% in patients with both hazelnut- and peanut-allergy (12).
Furthermore, 11S globulins of mustard (Sin a 2) and hazelnut (Cor a 9) were reported to be cross-reactive in a study conducted among 34 mustard-allergic patients (27).
Molecular Aspects
Biochemistry
Cor a 9, referred to as corylin, belongs to the 11S globulin family of seed storage proteins, a member of the cupin superfamily of proteins, that possesses a molecular weight of 40 kDa (acidic subunit). This superfamily of proteins is divided into monocupins and dicupins. The 11S globulin is a dicupin and consists of acidic as well as basic polypeptide chains that are bridged by disulfide linkages. They, together with 7S albumins, constitute 50% of the total seed storage proteins that are required for the germination of a plant (10).
The allergen has demonstrated high thermal and digestive resistance (21). The allergenicity of Cor a 9 was found stable after roasting (hot air or infrared), till a temperature of 140℃, however was lost by roasting at 170℃ after infra-red processing  (28). Further, the allergenicity was also found to be affected by autoclaving at 121℃ for 15 mins or 138℃ for 30 mins  (10).
Isoforms, epitopes, antibodies
As of 10th March 2021, only one isoallergen of Cor a 9 i.e., Cor a 9.0101, has been identified and officially published by the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature (10, 29).
Cross-reactivity
Clinical cross-reactivity or co-sensitization may be expected among different 11S globulins owing to high sequence identity or similarity. However, such structural or sequence homology may not necessarily translate into clinical cross-reactivity (24).
Cor a 9 shares significant sequence homology with other 11S globulins such as Jug r 4 of walnut (63%) (26), Ara h 3 of peanut (67%) (12), Sin a 2 of mustard (66%) (27) as well as Gly m 6 of soybean, Ana o 2 of cashew nut, Ber e 2 of Brazil nut, Pru du 6 of almond and Pis v 2 of pistachio (46-70%) (10). 
Diagnostic Relevance
Disease Severity
Determination of sIgE for Cor a 9 can serve as a prediction marker for clinical allergy to hazelnut with severe symptoms (20, 30) which holds true even in regions with less birch pollen-sensitization as reported by a study conducted on 82 individuals in Finland (31).
The predictive capacity of Cor a 9 determination was confirmed by DBPCFC test conducted on 8 children. The specificity and sensitivity of IgE estimation to differentiate between sensitization and clinical allergy were found to be 72% and 100%, respectively (32). Another study in Denmark reported the specificity and sensitivity of Cor a 9-sIgE to be 82.7% and 62.9%, respectively. However, Cor a 9-sIgE was also found in some hazelnut-tolerant individuals (22). Thus, it was suggested that Cor a 9-sIgE may be less specific than Cor a 14-sIgE (14, 22, 32). Furthermore, the predictive ability of Cor a 9-sIgE was best observed at a cut-off value of 1.0 kUA/L as reported in a meta-analysis involving 8 studies. However, it was suggested that rather than an independent test, the predictive ability of Cor a 9-sIgE could be enhanced by combining it with Cor a 14-sIgE and thus alleviate the risk of misclassifying patients (33).
Exposure
The main route of exposure is through ingestion (18).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: March 2021
References
  1. Barni S, Mori F, Piccorossi A, Sarti L, Pucci N, Maresca M, et al. Low-Dose Oral Food Challenge with Hazelnut: Efficacy and Tolerability in Children. International archives of allergy and immunology. 2019;178(1):97-100.
  2. Calamelli E, Trozzo A, Di Blasi E, Serra L, Bottau P. Hazelnut Allergy. Medicina (Kaunas, Lithuania). 2021;57(1).
  3. McWilliam V, Koplin J, Lodge C, Tang M, Dharmage S, Allen K. The Prevalence of Tree Nut Allergy: A Systematic Review. Current Allergy and Asthma Reports. 2015;15(9):54.
  4. Botha M, Basera W, Facey-Thomas HE, Gaunt B, Gray CL, Ramjith J, et al. Rural and urban food allergy prevalence from the South African Food Allergy (SAFFA) study. The Journal of allergy and clinical immunology. 2019;143(2):662-8.e2.
  5. Elizur A, Appel MY, Nachshon L, Levy MB, Epstein-Rigbi N, Golobov K, et al. NUT Co Reactivity - ACquiring Knowledge for Elimination Recommendations (NUT CRACKER) study. Allergy. 2018;73(3):593-601.
  6. Fedorova OS, Ogorodova LM, Fedotova MM, Evdokimova TA. [The prevalence of food allergy to peanut and hazelnut in children in Tomsk Region]. Voprosy pitaniia. 2014;83(1):48-54.
  7. Mahesh PA, Wong GW, Ogorodova L, Potts J, Leung TF, Fedorova O, et al. Prevalence of food sensitization and probable food allergy among adults in India: the EuroPrevall INCO study. Allergy. 2016;71(7):1010-9.
  8. McWilliam V, Peters R, Tang MLK, Dharmage S, Ponsonby AL, Gurrin L, et al. Patterns of tree nut sensitization and allergy in the first 6 years of life in a population-based cohort. The Journal of allergy and clinical immunology. 2019;143(2):644-50.e5.
  9. Ruiz Segura LT, Figueroa Pérez E, Nowak-Wegrzyn A, Siepmann T, Larenas-Linnemann D. Food allergen sensitization patterns in a large allergic population in Mexico. Allergologia et immunopathologia. 2020:In press.
  10. Costa J, Mafra I, Carrapatoso I, Oliveira MB. Hazelnut Allergens: Molecular Characterization, Detection, and Clinical Relevance. Critical reviews in food science and nutrition. 2016;56(15):2579-605.
  11. Datema MR, Zuidmeer-Jongejan L, Asero R, Barreales L, Belohlavkova S, de Blay F, et al. Hazelnut allergy across Europe dissected molecularly: A EuroPrevall outpatient clinic survey. The Journal of allergy and clinical immunology. 2015;136(2):382-91.
  12. Masthoff LJ, van Hoffen E, Mattsson L, Lidholm J, Andersson K, Zuidmeer-Jongejan L, et al. Peanut allergy is common among hazelnut-sensitized subjects but is not primarily the result of IgE cross-reactivity. Allergy. 2015;70(3):265-74.
  13. Johnson J, Malinovschi A, Lidholm J, Petersson CJ, Nordvall L, Janson C, et al. Sensitization to storage proteins in peanut and hazelnut is associated with higher levels of inflammatory markers in asthma. Clin Mol Allergy. 2020;18:11.
  14. Buyuktiryaki B, Cavkaytar O, Sahiner UM, Yilmaz EA, Yavuz ST, Soyer O, et al. Cor a 14, Hazelnut-Specific IgE, and SPT as a Reliable Tool in Hazelnut Allergy Diagnosis in Eastern Mediterranean Children. J Allergy Clin Immunol Pract. 2016;4(2):265-72.e3.
  15. Valcour A, Lidholm J, Borres MP, Hamilton RG. Sensitization profiles to hazelnut allergens across the United States. Ann Allergy Asthma Immunol. 2019;122(1):111-6.e1.
  16. Faber MA, De Graag M, Van Der Heijden C, Sabato V, Hagendorens MM, Bridts CH, et al. Cor a 14: missing link in the molecular diagnosis of hazelnut allergy? International archives of allergy and immunology. 2014;164(3):200-6.
  17. Masthoff LJ, Mattsson L, Zuidmeer-Jongejan L, Lidholm J, Andersson K, Akkerdaas JH, et al. Sensitization to Cor a 9 and Cor a 14 is highly specific for a hazelnut allergy with objective symptoms in Dutch children and adults. The Journal of allergy and clinical immunology. 2013;132(2):393-9.
  18. Datema MR, van Ree R, Asero R, Barreales L, Belohlavkova S, de Blay F, et al. Component-resolved diagnosis and beyond: Multivariable regression models to predict severity of hazelnut allergy. Allergy. 2018;73(3):549-59.
  19. van Ree R, Aalberse RC. Allergens and the Allergenic Composition of Source Materials. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Ollert M, editors. EAACI Molecular Allergology User's Guide: Using Molecular Allergology in the Clinical Practice. Zutich, Switzerland: The European Academy of Allergy and Clinical Immunology (EAACI); 2016. p. 11-20.
  20. Masthoff LJN, Blom WM, Rubingh CM, Klemans RJB, Remington BC, Bruijnzeel-Koomen C, et al. Sensitization to Cor a 9 or Cor a 14 has a strong impact on the distribution of thresholds to hazelnut. J Allergy Clin Immunol Pract. 2018;6(6):2112-4.e1.
  21. Blazowski L, Majak P, Kurzawa R, Kuna P, Jerzynska J. Food allergy endotype with high risk of severe anaphylaxis in children-Monosensitization to cashew 2S albumin Ana o 3. Allergy. 2019;74(10):1945-55.
  22. Eller E, Mortz CG, Bindslev-Jensen C. Cor a 14 is the superior serological marker for hazelnut allergy in children, independent of concomitant peanut allergy. Allergy. 2016;71(4):556-62.
  23. Kattan JD, Sicherer SH, Sampson HA. Clinical reactivity to hazelnut may be better identified by component testing than traditional testing methods. J Allergy Clin Immunol Pract. 2014;2(5):633-4 e1.
  24. Knol EF, Wickman M. Tree Nut and Seed Allergy. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Ollert M, editors. EAACI Molecular Allergology User's Guide: Using Molecular Allergology in the Clinical Practice. Zutich, Switzerland: The European Academy of Allergy and Clinical Immunology (EAACI); 2016. p. 245-54.
  25. Villalta D, Scala E, Mistrello G, Amato S, Asero R. Evidence of Cross-Reactivity between Different Seed Storage Proteins from Hazelnut (Corylus avellana) and Walnut (Juglans regia) Using Recombinant Allergen Proteins. International archives of allergy and immunology. 2019;178(1):89-92.
  26. Blankestijn MA, den Hartog Jager CF, Blom WM, Otten HG, de Jong GAH, Gaspari M, et al. A subset of walnut allergic adults is sensitized to walnut 11S globulin Jug r 4. Clin Exp Allergy. 2018;48(9):1206-13.
  27. Sirvent S, Akotenou M, Cuesta-Herranz J, Vereda A, Rodriguez R, Villalba M, et al. The 11S globulin Sin a 2 from yellow mustard seeds shows IgE cross-reactivity with homologous counterparts from tree nuts and peanut. Clin Transl Allergy. 2012;2(1):23.
  28. Lamberti C, Nebbia S, Antoniazzi S, Cirrincione S, Marengo E, Manfredi M, et al. Effect of hot air and infrared roasting on hazelnut allergenicity. Food Chem. 2020:128174.
  29. WHO/IUIS. Cor a 9 - Corylus avellana: WHO/IUIS Allergen Nomenclature Sub-Committee; 2021 [updated August 1, 2019March 10, 2021]. Available from: http://www.allergen.org/viewallergen.php?aid=236.
  30. Inoue Y, Sato S, Takahashi K, Yanagida N, Yamamoto H, Shimizu N, et al. Component-resolved diagnostics can be useful for identifying hazelnut allergy in Japanese children. Allergology international : official journal of the Japanese Society of Allergology. 2020;69(2):239-45.
  31. Uotila R, Röntynen P, Pelkonen AS, Voutilainen H, Kaarina Kukkonen A, Mäkelä MJ. For hazelnut allergy, component testing of Cor a 9 and Cor a 14 is relevant also in birch-endemic areas. Allergy. 2020;75(11):2977-80.
  32. Brandström J, Nopp A, Johansson SG, Lilja G, Sundqvist AC, Borres MP, et al. Basophil allergen threshold sensitivity and component-resolved diagnostics improve hazelnut allergy diagnosis. Clin Exp Allergy. 2015;45(9):1412-8.
  33. Nilsson C, Berthold M, Mascialino B, Orme M, Sjölander S, Hamilton R. Allergen components in diagnosing childhood hazelnut allergy: Systematic literature review and meta-analysis. Pediatr Allergy Immunol. 2020;31(2):186-96.