Whole Allergen
f222 Tea
f222 Tea Scientific Information
Type:
Whole Allergen
Display Name:
Tea
Allergen code:
f222
Family:
Theaceae
Species:
Camellia sinensis
Route of Exposure:
Ingestion
Source Material:
Tea leaves
Latin Name:
Camellia sinensis
Other Names:
Black Tea, Green Tea, Oolong tea
Summary
Tea comes from the Camellia sinensis plant and is regarded as the second most frequently consumed therapeutic, non-alcoholic beverage globally. Overall, 78% of total global tea production and consumption is black tea, 20% is green tea, and <2% is yellow or oolong tea. Despite its frequent consumption, allergy due to consumption of tea is extremely rare and is limited to individual case reports. Most frequently, food allergy to green tea is seen in patients with a history of occupational asthma, although it has occasionally been reported without prior sensitization. Symptoms include throat swelling, shortness of breath, nasal congestion, cough, dyspnea and, in severe cases, loss of consciousness. Contact sensitization to tea has also been reported in occupational settings. Epigallocatechin gallate has been indicated as the main causative agent in tea for both occupational and food allergy. 
Allergen
Nature
The tea plant (Camellia sinensis) belongs to the Theaceae family and originates from the tropical and temperate regions of Asia, South America and Africa. In recent times, the majority of tea has been produced and consumed in China, Sri Lanka, Japan, and India. Currently, tea is regarded as the second most frequently consumed therapeutic, non-alcoholic beverage globally, with only water being consumed more often. Overall, 78% of total global tea production and consumption is black tea, 20% is green tea, and <2% is yellow or oolong tea (1). All 3 types come from the Camellia sinensis plant, with differences arising only from their processing: green tea is unfermented, oolong is semifermented and black is fermented (2, 3). Tea consumption is associated with many health benefits, including its antioxidant and anti-inflammatory effects, as well as its positive effects on cardiovascular health, diabetes, gastrointestinal cancers, memory and bone health. Tea is also predicted to have anti-allergenic properties, due to its antihistaminic activity (1).
Taxonomy
Taxonomic tree of Tea (4)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Tracheophyta
Subphylum
Spermatophytina
Class
Magnoliopsida
Family
Theaceae
Genus
Camellia
Clinical Relevance
Despite its frequent consumption, allergy due to consumption of tea (Camellia sinensis) is extremely rare and is limited to individual case reports. Most frequently, food allergy to green tea is seen in patients with a history of occupational asthma, although it has been reported occasionally without prior sensitization (2, 3).
A 20-year-old woman with no history of asthma, anxiety, or occupational exposure to green tea presented with allergy symptoms after consuming brewed green tea. Approximately 15 minutes after consuming green tea, she began to experience throat swelling, itching and soreness, hoarse voice, shortness of breath, and nasal congestion, all lasting up to 3 hours. She had imbibed green tea in the past without any reaction; however, she then had a second episode more severe than the first. A skin prick test (SPT) to green tea elicited a 6-mm wheal with a 61-mm flare, whereas green tea SPT were negative in the 5 control subjects. Further SPT performed to food allergens based on her food history were non-reactive, including to chocolate and coffee. IgE testing to fermented black tea or a blend of several black teas were negative (3).
Occupational asthma caused by inhalation of green dust has been reported in 11 patients, 5 of which also developed food allergy to green tea. These patients had worked for several years at different green tea factories in Japan. They developed food allergy to green tea simultaneously with the onset of occupational asthma or subsequently. Allergic symptoms, including cough, dyspnea and, in a severe case, loss of consciousness, occurred soon after ingestion of black, green and oolong teas, as well as after ingestion of cakes and noodles containing green tea powder. The diagnosis of green tea–induced asthma was confirmed with SPT and inhalation challenges using green tea dust (2).
Asthma
The catechin epigallocatechin gallate (EGCg) from green tea has been shown to inhibit mast cell degranulation and suppress allergen- and non-allergen-specific IgE production in asthmatic patients. This suggests that green tea may have beneficial immunoregulatory effects on IgE responses in asthmatics (5).
Atopic Dermatitis
Cases of immediate and delayed contact sensitization to Camellia sinensis have been described in occupational settings. A 49-year-old female wine and tea merchant with no personal history of atopy or skin disease experienced symptoms of contact sensitization within 5 minutes of exposure to tea dust. The patient tolerated drinking tea infusions both before and after this incident. Symptoms included itching of the face, neck and mouth, periorbital edema and rhinitis. The rash lasted for 3–4 days, with antihistamine treatment having some effect. Histamine release (HR) testing showed sensitivity to 3 out of the 5 tea blends tested. Skin prick tests to standard inhalant allergens were positive to birch, grass, and mugwort pollen as well as to house dust mites and, to a lesser degree, dog and Alternaria alternata. The report concluded that the history, clinical tests and HR tests were suggestive of concomitant immediate and delayed hypersensitivity reactions to tea (6).
Prevention and Therapy
Allergen immunotherapy
Green tea has shown to possess antioxidative, antimutagenic and anti-inflammatory properties. Immunoreregulatory effects on human IgE responses in-vitro have also been demonstrated, suggesting a possible role for green tea in the treatment of atopic disease (7). 
Molecular Aspects
Allergenic molecules
The following allergenic molecules have been characterized from Camellia sinensis leaves (8):
Name
Type
Mass (kDa)
Cam s
Unknown function
-
EGCg, a major low-molecular-weight polyphenol in green tea leaves, has been indicated as the causative agent in occupational green tea allergy. The specific IgE antibody to EGCg is also suspected to be a key factor for the subsequent development of food allergy to green tea (2).
Cross-reactivity
There is no indication for cross-reactivity with other allergens
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. Naveed M, BiBi J, Kamboh AA, Suheryani I, Kakar I, Fazlani SA, et al. Pharmacological values and therapeutic properties of black tea (Camellia sinensis): A comprehensive overview. Biomed Pharmacother. 2018;100:521-31.
  2. Shirai T, Hayakawa H, Akiyama J, Iwata M, Chida K, Nakamura H, et al. Food allergy to green tea. J Allergy Clin Immunol. 2003;112(4):805-6.
  3. Wu SS, Johnson JA, Peppers B, Tcheurekdjian H, Hostoffer R. A case of green tea (Camellia sinensis) imbibement causing possible anaphylaxis. Ann Allergy Asthma Immunol. 2017;118(6):747-8.
  4. ITIS. Camellia sinensis 2021 [cited 2022 24.01.22]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=506801#null.
  5. Wu SY, Silverberg JI, Joks R, Durkin HG, Smith-Norowitz TA. Green tea (Camelia sinensis) mediated suppression of IgE production by peripheral blood mononuclear cells of allergic asthmatic humans. Scand J Immunol. 2012;76(3):306-10.
  6. Paulsen E, Hvid L, Andersen F. Immediate and delayed contact reactions to white and green tea blends. Contact Dermatitis. 2022;86(2):134-6.
  7. Hassanain E, Silverberg JI, Norowitz KB, Chice S, Bluth MH, Brody N, et al. Green tea (Camelia sinensis) suppresses B cell production of IgE without inducing apoptosis. Ann Clin Lab Sci. 2010;40(2):135-43.
  8. allergome.org. Bromus inermis 2021 [cited 2022 19.01.22]. Available from: http://www.allergome.org/script/search_step2.php.