Whole Allergen
f91 Mango
f91 Mango Scientific Information
Type:
Whole Allergen
Display Name:
Mango
Allergen code:
f91
Family:
Anacardiaceae
Species:
indica
Route of Exposure:
Ingestion, contact
Source Material:
Flesh of the fruit, peel
Latin Name:
Mangifera indica
Other Names:
Edible mango; Indian mango
WHO/ICD-11 code:
XM6AW1
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Mango (Mangifera indica) is part of the Anacardiaceae family and grown worldwide. There are many different cultivars, which vary in appearance, flavor and uses. Peel and flesh of the fruit are the main sources of hypersensitivity and allergic reactions, though pollen may also be implicated in allergies. A large number of mango allergens have been described.
Immediate hypersensitivity is a common clinical presentation of mango allergy, with symptoms ranging from localized to systemic, including anaphylaxis. Pollen-food allergy syndrome (PFAS) and one of its common presentations, oral allergy syndrome (OAS) may be experienced. Contact dermatitis is also a known clinical presentation of mango allergy. Cross reactivity with a number of other plant food and pollen allergens can occur.
Allergen
Nature
Mango (Mangifera indica) is the fruit of the mango tree, which can be found worldwide. Due to the many cultivars, phenotype of both tree and fruit varies greatly. The fruit is edible, variable in shape and size (up to and over 30 cm long); the skin is thick and can be variably colored, with green, red and yellow areas. The flesh varies in color and flavor; a tough, fibrous seed is present within. The mango tree is a large evergreen plant, reaching upwards of 40 m; the leaves have variable morphology, usually elliptic to lanceolate, leathery with visible nerves. The tree produces large inflorescences (up to 30 cm long), branched and heavily flowered (300–6000 flowers); the flowers colors vary depending on cultivar (1).
The mango plant and fruit are versatile; from stem bark extracts and mangiferin as medicinal compounds (2), to food. The fruit is commonly consumed raw or preserved in the form of juice, canned slices and chutneys (3).
Taxonomy
Taxonomic ree of mango Mangifera indica (1)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Dicotyledonae
Order
Sapindales
Family
Anacardiaceae
Genus
Mangifera
Tissue
Peel and flesh of the fruit (4-7).
Epidemiology
Worldwide distribution 
Mango allergy has been reported in many countries, from where mango is grown and also from other countries that import the fruit (8).
Risk factors 
Male patients and adults (19–60 years old) might be at higher risk of mango sensitization (7). Preschool age children may also be at higher risk of mango hypersensitivity than younger children (9). 
Exposure during the spring season and suffering from eczema increased the risk of anaphylaxis to certain fruits, including mango (6).
Previous sensitization to urushiol (found in poison ivy and poison oak) increases the risk of hypersensitivity contact reactions to mango peel (4, 10). In addition, working as mango pickers (if having a history of contact with, or living in areas where poison ivy and poison oak are endemic) may be a risk factor for developing mango contact dermatitis (10).
Environmental Characteristics
Worldwide distribution
The mango tree is found worldwide, preferring warmer climates (1).
Route of Exposure
Main
Ingestion of the fruit (5-7), contact with the peel (4).
Clinical Relevance
Immediate hypersensitivity is a common clinical presentation of mango allergy. This can be localized or show systemic symptoms, including anaphylaxis. Patients may present with a combination of the following: erythema and urticaria, angioedema of the face or extremities, rhinorrhea and rhino-conjunctivitis, dyspnea/respiratory distress, cough, gastrointestinal symptoms. Anaphylactic shock may present within few minutes of mango ingestion (8). In a study of 250 patients referred to hospital with fruit-induced anaphylaxis, 9.2% of cases were caused by mango. Approximately 10% of patients experienced severe reactions, 15% had mild reactions and the remaining presented with moderately severe reactions (6).
Testing of specific IgE in a 30-year old woman with history of seasonal rhinitis showed that this patient is multi-sensitized to a number of plant allergens; however, the only severe anaphylactic reaction experienced until then was following mango ingestion. It was suspected that the symptoms may be due to cross-reactivity with mugwort antigens (11).
Allergic rhinitis
Although most patients experience mango allergy after ingestion of the fruit, mango tree pollen was shown to be a sensitizing agent in 16% of patients with allergic rhinitis (12). In another study, asthma/allergic rhinitis (AR) patients showed hypersensitivity to mango allergens, as shown by IgE specificities (13).
Other diseases
Contact dermatitis (type IV delayed hypersensitivity response) to mango peel was the cause of the diffuse, macular, blanching, erythematous and pruritic rash presented by a 41-year old patient two days after handling mangos. He reported a remote history of contact dermatitis to poison ivy (4). Periorbital edema, eczematous rash and blisters around the mouth, alongside contact dermatitis, have also been described following exposure to mango (8).
Allergic conjunctivitis (AC) was the topic of a study which showed preschool children with this condition were significantly more likely to present with mango (and other foods)-specific serum IgE than younger age groups (9).
OAS (considered a manifestation of pollen-food allergy syndrome PFAS), consisting of IgE-mediated allergy symptoms limited to the oral cavity, is another presentation of mango allergy (8).
Diagnostics Sensitization
Testing for food allergies involves measuring serum specific IgE responses, skin prick tests (SPTs) and oral challenges (5, 13, 14). In a study of 309 patients with known history of asthma AR, specific allergen IgE testing showed that 14.5% of serum samples reacted to mango allergens (13).
In another study of a 100 patients suffering from AR, 16% has positive SPT to mango tree pollen (12). Other researchers analyzing food-specific IgE responses in 1795 patients of all age groups showed that mango elicited moderate to high positivity (42.8% and 28.6% respectively) (15). A large study, in China (Henan Province) recruited 15,534 patients after analysis of their medical records. A patient would be included if they had experienced symptoms suggestive of allergies such as skin rash, hives, eczema, runny nose, red or itchy eyes, sneezing and shortness of breath. The results showed mango sensitization in 4.94% of samples, as determined by the presence of specific IgE (7).
Prevention and Therapy
Allergen immunotherapy
Mangiferin (a polyphenol present in different tissues of the mango tree) showed antioxidant, anti-inflammatory and anti-allergic effects in a murine model of AR, by reducing nasal inflammation. This suggests that mangiferin might be a suitable candidate for the treatment of human AR (16).
Prevention strategies 
Avoidance consists of eliminating mango from the diet, once accurately diagnosed to avoid unnecessary diet restrictions (17).
Molecular Aspects
Allergenic molecules
A number of mango-allergenic antigens have been identified. The main ones are Man i 1 (40kDa; GAPDH), Man i 2 (30kDa) and Man i 3 (a profilin, with similar structure to mugwort Art v 1)(11, 18).
Three known banana allergens (Mus a 1, Mus a 2 and Mus a 5; a profilin, chitinase and β-1,3-glucanase respectively) were found to be shared by mango by using proteomics libraries (19). 5-resorcinol is an allergen found in mango peel and appears to be related to the poison ivy/poison oak antigen urushiol (4). Other antigens found in the skin, bark and external flesh of the fruit are limonene, cardol and β-pinene (8). Chitinases and β-1,3-glucanases from the mango protein fraction were recognized by IgE of patients with known allergies; however, the clinical relevance of these remained unclear as the same patients did not show symptoms when consuming mango (14).
Table adapted from Allergome.org (20)
Allergen
Type
Mass (kDa)
Man i 1
GAPDH
40
Man i 2
Unknown
30
Man i 3
Profilin
14
Man i 14kDa
PR proteins
14
Man i Chitinase
Chitinase
46
Cross-reactivity
Cross-reactivity between members of the Anacardiaceae family has been reported.
A study of 18 children with pistachio and/or cashew nut allergies (both Anacardiaceae) showed that mango was well tolerated in food challenges, suggesting elimination diets may be unnecessary (5).
In a larger study, initial results suggested that 21% of cashew allergic patients (up to 52% with a history of eczema, asthma or AR) had mango hypersensitivity; the follow-up study carried out open food challenges with mango which did not elicit any clinical reactions (21). In another study, 50% of patients with specific IgE to cashew nut showed cross sensitivity to pistachio, mango, pink peppercorn or sumac (14).
Cross reactivities with other plants pollens have also been discussed in the context of pollen-food allergy syndrome (PFAS), especially birch-mango (6, 22). Mugwort pollen, celery and carrot have also been associated with mango cross-reactivity, due to allergens related to Bet v 1 (birch) and Art v 1 (mugwort)(11, 23)  and birch profilin Bet v 2 (22, 24). The pan-allergen hevein (present in latex) is known to cause cross reactions with certain allergens present in a number of common foods such as banana, avocado, kiwi, tomato, wheat, mango, chestnut, peach, orange and passion fruit (8, 24).
Cross-reactivity between mango peel and poison ivy/poison oak (also of the family Anacardiaceae) has been reported (4).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:March 2022
References
  1. CABI. Mangifera indica (mango) 2021 [cited 2022 13.01.22]. Available from: https://www.cabi.org/isc/datasheet/34505.
  2. Bhat H, Sampath P, Pai RJ, Bollor R, Baliga MS, Fayad R. Chapter 15 - Indian Medicinal Plants as Immunomodulators: Scientific Validation of the Ethnomedicinal Beliefs. In: Watson RR, Preedy VR, editors. Bioactive Food as Dietary Interventions for Arthritis and Related Inflammatory Diseases. San Diego: Academic Press; 2013. p. 215-24.
  3. Bello-Pérez LA, Agama-Acevedo E, Osorio-Díaz P, Utrilla-Coello RG, García-Suárez FJ. Chapter 22 - Banana and Mango Flours. In: Preedy VR, Watson RR, Patel VB, editors. Flour and Breads and their Fortification in Health and Disease Prevention. San Diego: Academic Press; 2011. p. 235-45.
  4. Yoo MJ, Carius BM. Mango Dermatitis After Urushiol Sensitization. Clin Pract Cases Emerg Med. 2019;3(4):361-3.
  5. Azcona OM, Romero L, Bartolome B, Balboa V, Vila L. Clinical cross-reactivity among mango, pistachio nut, and cashew nut in allergic children. The Journal of Allergy and Clinical Immunology: In Practice. 2020;8(9):3234-6.e2.
  6. Gabrielli S, Clarke AE, Morris J, Gravel J, Lim R, Chan ES, et al. Fruit-Induced Anaphylaxis: Clinical Presentation and Management. J Allergy Clin Immunol Pract. 2021;9(7):2825-30.e2.
  7. Sun X, Zhao J, Wang Q, Shi G, Yang J, Ming L. Prevalence of allergen sensitization among 15,534 patients with suspected allergic diseases in Henan Province, China. Asian Pac J Allergy Immunol. 2019;37(2):57-64.
  8. Sareen R, Shah A. Hypersensitivity manifestations to the fruit mango. Asia Pac Allergy. 2011;1(1):43-9.
  9. Wang W, Du X, Ye L, Wang X, Zhang G. Distribution of serum specific IgE in children with allergic conjunctivitis and analysis of its concomitant allergic diseases. Transl Pediatr. 2020;9(5):636-44.
  10. Hershko K, Weinberg I, Ingber A. Exploring the mango – poison ivy connection: the riddle of discriminative plant dermatitis. Contact Dermatitis. 2005;52(1):3-5.
  11. Ukleja-Sokołowska N, Gawrońska-Ukleja E, Lis K, Żbikowska-Gotz M, Sokołowski Ł, Bartuzi Z. Anaphylactic reaction in patient allergic to mango. Allergy, Asthma & Clinical Immunology. 2018;14(1):78.
  12. Pumhirun P, Towiwat P, Mahakit P. Aeroallergen sensitivity of Thai patients with allergic rhinitis. Asian Pac J Allergy Immunol. 1997;15(4):183-5.
  13. Di LL, Wang LX, Ma X, Wen WL, Gao XP. Allergic sensitization in patients with rhinitis and bronchial asthma in Ningxia region of China. J Eval Clin Pract. 2020;26(3):1001-4.
  14. Bastiaan-Net S, Reitsma M, Cordewener JHG, van der Valk JPM, America TAHP, Dubois AEJ, et al. IgE Cross-Reactivity of Cashew Nut Allergens. International Archives of Allergy and Immunology. 2019;178(1):19-32.
  15. Segura-Medina P, Vargas MH, Aguilar-Romero JM, Arreola-Ramírez JL, Miguel-Reyes JL, Salas-Hernández J. Mold burden in house dust and its relationship with asthma control. Respir Med. 2019;150:74-80.
  16. Piao CH, Fan YJ, Nguyen TV, Song CH, Chai OH. Mangiferin Alleviates Ovalbumin-Induced Allergic Rhinitis via Nrf2/HO-1/NF-κB Signaling Pathways. Int J Mol Sci. 2020;21(10).
  17. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  18. Tsai WC, Wu TC, Chiang BL, Wen HW. Cloning, expression, and purification of recombinant major mango allergen Man i 1 in Escherichia coli. Protein Expr Purif. 2017;130:35-43.
  19. Cardona EEG, Heathcote K, Teran LM, Righetti PG, Boschetti E, D'Amato A. Novel low-abundance allergens from mango via combinatorial peptide libraries treatment: A proteomics study. Food Chem. 2018;269:652-60.
  20. Allergome. Mango. 2022.
  21. van der Valk JPM, Bouche Re, Gerth van Wijk R, de Groot H, Wichers HJ, Dubois AEJ, et al. Low percentage of clinically relevant pistachio nut and mango co-sensitisation in cashew nut sensitised children. Clinical and Translational Allergy. 2017;7(1):8.
  22. Song J, Zhang H, Liu Z, Ran P. Mango profilin: cloning, expression and cross-reactivity with birch pollen profilin Bet v 2. Molecular Biology Reports. 2007;35(2):231.
  23. Paschke A, Kinder H, Zunker K, Wigotzki M, Steinhart H, Weßbecher R, et al. Characterization of cross-reacting allergens in mango fruit. Allergy. 2001;56(3):237-42.
  24. Hassan AK, Venkatesh YP. An overview of fruit allergy and the causative allergens. Eur Ann Allergy Clin Immunol. 2015;47(6):180-7.