Whole Allergen
t222 Arizona cypress pollen
t222 Arizona cypress pollen Scientific Information
Type:
Whole Allergen
Display Name:
Arizona cypress pollen
Allergen code:
t222
Family:
Cupressaceae
Species:
Hesperocyparis arizonica (formerly: Cupressus arizonica)
Route of Exposure:
inhalation
Source Material:
Pollen
Latin Name:
Hesperocyparis arizonica
Other Names:
Cypress, Arizona rough cypress, Cedro blanco, C. glabra, C. arizonica var. glabra
WHO/ICD-11 code:
XM5TH6
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Arizona cypress, indigenous to North America, has been extensively planted in the Mediterranean area, the Middle East, and Australia. It bears considerable resemblance with other members of the Cupressaceae family regarding pollen characteristics (small and anemophilous), pollination season (winter), symptoms (rhinitis and rhinoconjunctivitis with virtually no asthma), diagnosis (interchangeable whole allergens and molecular allergens), treatment, and prevention.
Allergen
Nature
Arizona cypress (Hesperocyparis arizonica, formerly known as Cupressus arizonica) is typically used as a windbreak, privacy screen, ornamental tree, or for soil erosion prevention. Arizona cypress is a steeple-shaped, coniferous evergreen that can grow over 25 m tall. Its leaves are pale green to grey-blue. The small, inconspicuous yellow flowers are monoecious (individual flowers are either male or female, but both sexes can be found on the same plant) and are pollinated by wind (anemophilous pollen) (1).
Taxonomy
The Cupressaceae family contains five genera: Cupressus, Hesperocyparis, Juniperus, Cryptomeria, and Chamaecyparis. Arizona cypress, currently classified as Hesperocyparis arizonica, was formerly known as Cupressus arizonica (1-4).
Taxonomic tree of Arizona cypress
Kingdom Plantae
Clade Tracheophytes
Division Pinophyta
Class Pinopsida
Subclass Pinidae
Clade Conifers II
Family Cupressaceae
Genus Hesperocyparis
Species H. arizonica
Tissue 
Pollen grains from Arizona cypress are spherical, with a diameter of 24 µm (5). Due to morphological resemblance and extensive cross-reactivity (6), pollens of the Cupressaceae family are counted together. They make up an important part of global airborne pollen counts, e.g. 25% to 40% in Mediterranean areas, 30% in Mexico, up to 10% on the Eastern coast of the US, 19% in Yunnan, China, and up to 60% in Japan (1). Cupressaceae pollen is anemophilous, meaning that it becomes airborne; its small size favors its dispersion over long distances, outside the actual region where the trees grow (1). Arizona cypress pollinates in January and February and partially overlaps with C. sempervirens, which pollinates from February to the end of March (7). Cupressaceae pollens can persist during all seasons and their allergenicity may be accentuated by air pollution (1,5).
Epidemiology
Worldwide distribution
Pollen grains from Arizona cypress contain allergenic molecules able to induce sensitization and allergic diseases. The allergenicity of Arizona cypress pollen was reported to be higher than that of C. sempervirens (8,9). Arizona cypress pollinosis occurs mainly in Central and North America, where the tree is indigenous, and in the Mediterranean area, Iran, South Africa, and Australia (1).
Risk factors 
The individual risk factors for being diagnosed with Cupressaceae allergy were addressed in 110 Cupressaceae-allergic consecutive outpatients from Southern France, as compared with grass pollen allergic patients from the same clinic (10). Significantly different features between the two patient groups were: equal gender distribution (vs a male/female ratio of 2 for grass pollinosis, p = 0.001), later age of symptom onset (32 vs 18, p = 0.0002), and relatively high prevalence of dry cough (16.5% vs 0%, p = 0.008) for Cupressaceae pollinosis. The later age of onset of Cupressaceae pollinosis, especially in monosensitized patients, is consistently retrieved in other studies (11-13).
Environmental Characteristics
Living environment
Arizona cypress shares with other trees from the Cupressaceae family the ability to grow in arid and hot areas, explaining their widespread use as ornamental trees in the 20th century (1,4,5).
Worldwide distribution (of the species)
Arizona cypress is the only cypress species native to the south-west of North America. It can be found in coniferous woodlands on rough, very dry and very rocky soils. It has been widely exported, especially to Europe, where it is now the second most common cypress species following the Italian (funeral) cypress (Cupressus sempervirens), but also to Australia and the Middle East (1-4). The wide use of Arizona cypress for reforestation, wind and noise barriers, and ornamentally in gardens and parks, especially in the Mediterranean area and in North America, led to an increase in annual pollen counts (14-17).
Route of Exposure
Main 
The main route of exposure to Arizona cypress allergens is inhalation of allergenic pollen, which occurs mainly during the pollination season in January and February.
Secondary
Direct skin contact with Cupressaceae pollen or resin may result in contact dermatitis (18).
Clinical Relevance
Cupressaceae pollinosis, including Arizona cypress, manifests during wintertime, mainly as rhinitis or rhinoconjunctivitis, less often as asthma or cutaneous symptoms, and exceptionally as bronchitis (1,19,20). Its prevalence increases with exposure to Cupressaceae pollens, which has plummeted during the second half of last century, due to the extended use of Cupressaceae as ornamental trees next to housing units (1,9). In fact, Cupressaceae pollinosis was underestimated until the end of the 20th century, because of the lack of physician and patient awareness and lack of performant Cupressaceae pollen extracts for diagnostic testing (7,11,12).  Due to the similar morphology of Cupressaceae pollens, to their allergenic cross-reactivity and to the interchangeable use of pollen extracts for diagnosis, such as Juniperus ashei, Arizona cypress, C. sempervirens, and more recently the molecular allergen Cup a 1 (21,22), Arizona cypress pollinosis is not distinguished from pollinosis induced by other Cupressaceae genera or species (1,10).
For further details on the clinical relevance of Cupressaceae pollinosis, the reader is referred to the Cupressus sempervirens (t23) section.
Diagnostics Sensitization
Arizona cypress extracts have been consistently found more potent than C. sempervirens for the diagnosis of Cupressaceae sensitization and allergy with skin prick tests and in vitro assays (6,9,23). Following the characterization of the major allergen Cup a 1 and the demonstration of its sequence identity and cross-reactivity with other Cupressaceae pectate lyases Cup s 1 (C. sempervirens), Jun a 1 (Juniperus ashei), Cry j 1 (Cryptomeria japonica), and Cha o 1 (Chamaecyparis obtusa) (9,24-26), this allergen has become widely used as a marker of genuine sensitization to Cupressaceae pollen (26).
Prevention and Therapy
Allergen immunotherapy
Allergen immunotherapy for Cupressaceae pollinosis proved efficient in several studies using Arizona cypress or other Cupressaceae extracts, but the number of patients included in each of these studies was low (reviewed in ref 1).
Prevention strategies
Avoidance
During the pollination season, exposure to Cupressaceae pollen can be reduced by using a few precautionary measures, such as avoiding outdoor exercise, keeping doors and windows closed, washing hair before sleeping, using protective sunglasses and a mask for outdoor activities, and avoiding drying of linen clothes outside (1). If the patient’s health condition deteriorates, moving to an area with lower pollen counts may be advised (27).
Molecular Aspects
Allergenic molecules
Cup a 1 is the only allergen from Arizona cypress included in the IUIS database as of July 2021. Cup a 1, used either a recombinant or a a purified molecule, is widely used as a marker of genuine sensitization to Cupressaceae (26). Further allergens from Arizona cypress pollen have been reported and characterized in the literature.
Name
Biochemical activity
Molecular weight (kDa)
Glycosylation
Major/minor allergen
Marker allergen
IUIS
Reference
Cup a 1
Pectate lyase
43
Yes
Major
Yes
Yes
24
Cup a 2
Polygalacturonase
41 (fragment)
Yes
Major
No
No
1
Cup a 3
Thaumatin-like protein
21
No
Major
No
No
28
Cup a 4
Polcalcin
18
No
Minor
No
No
29
Although currently not characterized, an allergenic gibberellin-regulated protein is presumably expressed in Arizona cypress pollen, similarly to C. sempervirens (Cup s 7), J. ashei (Jun a 7), and Cryptomeria japonica (Cry j 7) (30,31).
Biomarkers of severity
Cup a 1 is the marker allergen for sensitization to Cupressaceae family including Arizona cypress and C. sempervirens (22,25,26). Polcalcin sensitization can be assayed using Phl p 7 or Bet v 4 as surrogates. In patients with suspected pollen – food cross-reactivity due to gibberellin-regulated proteins, Pru p 7 is the currently available marker (32).
Cross-reactivity
There is extensive cross-reactivity between Arizona cypress pollen and other members of the Cupressaceae family, due to high sequence identity and similarity between molecular allergens. Cross-reactivity with pollens outside the family is possible, mainly through polcalcin sensitization, but relatively infrequent for this minor allergen (9.6% Cup a 4 sensitization in cypress-allergic patients, ref 29). The carbohydrate moieties present on pectate lyase Cup a 1 and other major allergens of Cupressaceae family contribute to IgE binding, but their pathogenic role is not established. Pollen-food syndrome due to Cupressaceae primary sensitization has been suspected for a long time. Although multiple allergens could be involved, e.g. polygalacturonase, thaumatin-like protein, and gibberellin-regulated protein, formal demonstration has only been provided for the latter (31,32).
Compiled By
Author:Joana Vitte
Reviewer: Dr. Christian Fischer
Last reviewed: March  2022
References
  1. Charpin D, Pichot C, Belmonte J, Sutra JP, Zidkova J, Chanez P et al.  Cypress Pollinosis: from Tree to Clinic. Clin Rev Allergy Immunol 2019;56(2):174-95.
  2. Adams RP, Bartel JA, Price RA. A new genus, Hesperocyparis, for the cypresses of the Western Hemisphere (Cupressaceae). Phytologia 2009;91(1):160-85.
  3. Mao K, Ruhsam M, Ma Y, Graham SW, Liu J, Thomas P et al. A transcriptome-based resolution for a key taxonomic controversy in Cupressaceae. Ann Bot 2019;123(1):153-67.
  4. Osada T, Okano M. Japanese cedar and cypress pollinosis updated: New allergens, cross-reactivity, and treatment. Allergol Int 2021;70(3):281-90.
  5. Shahali Y, Pourpak Z, Moin M, Mari A, Majd A. Instability of the structure and allergenic protein content in Arizona cypress pollen. Allergy 2009;64(12):1773-9.
  6. Barletta B, Afferni C, Tinghino R, Mari A, Di Felice G, Pini C. Cross-reactivity between Cupressus arizonica and Cupressus sempervirens pollen extracts. J Allergy Clin Immunol 1996;98(4):797-804.
  7. D’Amato G, Spieksma FT, Liccardi G, Jäger S, Russo M, Kontou-Fili K et al. Pollen-related allergy in Europe. Allergy 1998;53(6):567-78.
  8. Mistrello G, Roncarolo D, Zanoni D, Zanotta S, Amato S, Falagiani P, Ariano R. Allergenic relevance of Cupressus arizonica pollen extract and biological characterization of the allergoid. Int Arch Allergy Immunol 2002;129(4):296-304.
  9. Barberini S, Della Rocca G, Danti R, Zanoni D, Mori B, Ariano R et al. Different allergenicity of pollen extracts of three Mediterranean cypress species accounted for cytological observations. Eur Ann Allergy Clin Immunol 2015;47(5):149-55.
  10. Boutin-Forzano S, Gouitaa M, Hammou Y, Ramadour M, Charpin D. Personal risk factors for cypress pollen allergy. Allergy 2005;60(4):533-5.
  11. Bousquet J, Knani J, Hejjaoui A, Ferrando R, Cour P, Dhivert H, Michel FB. Heterogeneity of atopy. I. Clinical and immunologic characteristics of patients allergic to cypress pollen. Allergy 1993;48(3):183-8.
  12. Mari A, Di Felice G, Afferni C, Barletta B, Tinghino R, Pini C. Cypress allergy: an underestimated pollinosis. Allergy 1997;52(3):355-6.
  13. Agea E, Bistoni O, Russano A, Corazzi L, Minelli L, Bassotti G et al. The biology of cypress allergy. Allergy 2002;57(10):959-60.
  14. Emberlin JC. Grass, tree and weed pollen. In: Kay B, editor. Allergy and allergic diseases. Oxford: Blackwell Scientific, 1997:845-57.
  15. Caiaffa MF, Macchia L, Strada S, Bariletto G, Scarpelli F, Tursi A. Airborne Cupressaceae pollen in southern Italy. Ann Allergy 1993;71(1):45-50.
  16. Waisel Y, Epstein V. Cypress pollen counts in Israel. Allergie et Immunologie 2000;31(3):121.
  17. Sneller MR, Hayes HD, Pinnas JL. Pollen changes during five decades of urbanization in Tucson, Arizona. Ann Allergy 1993;71(6):519-24.
  18. Samaran Q, Clark E, Dereure O, Raison‐Peyron N. Allergic contact dermatitis to Cupressus sempervirens resin and cross‐reaction with colophonium. Contact Dermatitis 2020;83(1):33-35.
  19. Italian Association of Aerobiology. An epidemiological survey of Cupressaceae pollenosis in Italy. J Investig Allergol Clin Immunol 2002;12(4):287-92.
  20. Bobolea I, Barranco P, Sastre B, Fernández-Nieto M, del Pozo V, Quirce S. Seasonal eosinophilic bronchitis due to allergy to Cupressus arizonica pollen. Ann Allergy Asthma Immunol 2011;106(5):448-9.
  21. Mari A, Di Felice G, Afferni C, Barletta B, Tinghino R, Sallusto F et al. Assessment of skin prick test and serum specific IgE detection in the diagnosis of Cupressaceae pollinosis. J Allergy Clin Immunol 1996;98(1):21-31.
  22. Klingebiel C, Charpin D, Mège JL, Vitte J. Laboratory diagnosis of respiratory allergy to Cupressaceae: Better performance using juniper pollen extract rather than cypress pollen extract. Rev Fr Allergol 2016;56:452-61.
  23. Di Felice G, Caiaffa MF, Bariletto G, Afferni C, Di Paola R, Mari A et al. Allergens of Arizona cypress (Cupressus arizonica) pollen: characterization of the pollen extract and identification of the allergenic components. J Allergy Clin Immunol 1994;94(3 Pt 1):547-55.
  24. Aceituno E, Del Pozo V, Mínguez A, Arrieta I, Cortegano I, Cárdaba B et al. Molecular cloning of major allergen from Cupressus arizonica pollen: Cup a 1. Clin Exp Allergy 2000;30(12):1750-8.
  25. Pichler U, Hauser M, Wolf M, Bernardi ML, Gadermaier G, Weiss R et al. Pectate lyase pollen allergens: sensitization profiles and cross-reactivity pattern. PLoS One 2015;10(5):e0120038.
  26. Scala E, Asero R, Niederberger V. Tree Pollen Allergy, pp 45-52. In Molecular Allergology User’s Guide, Matricardi et al, Pediatr Allergy Clin Immunol 2016;27 Suppl 23:1-250.
  27. Xie Z-J, Guan K, Yin J. Advances in the clinical and mechanism research of pollen induced seasonal allergic Asthma. Am J Clin Exp Immunol 2019;8(1):1-8.
  28. Cortegano I, Civantos E, Aceituno E, del Moral A, López E, Lombardero M et al. Cloning and expression of a major allergen from Cupressus arizonica pollen, Cup a 3, a PR-5 protein expressed under polluted environment. Allergy 2004;59(5):485-90.
  29. Pico de Coaña Y, Parody N, Fuertes MÁ, Carnés J, Roncarolo D, Ariano R et al. Molecular cloning and characterization of Cup a 4, a new allergen from Cupressus arizonica. Biochem Biophys Res Commun 2010;401(3):451-7.
  30. Tuppo L, Alessandri C, Giangrieco I, Ciancamerla M, Rafaiani C, Tamburrini M et al. Isolation of cypress gibberellin-regulated protein: Analysis of its structural features and IgE binding competition with homologous allergens. Mol Immunol 2019;114:189-195.
  31. Ehrenberg AE, Klingebiel C, Östling J, Larsson H, Mattsson L, Vitte J et al. Characterization of a 7 kDa pollen allergen belonging to the gibberellin-regulated protein family from three Cupressaceae species. Clin Exp Allergy 2020;50(8):964-972.
  32. Klingebiel C, Chantran Y, Arif-Lusson R, Ehrenberg AE, Östling J, Poisson A et al. Pru p 7 sensitization is a predominant cause of severe, cypress pollen-associated peach allergy. Clin Exp Allergy 2019;49(4):526-536.