Whole Allergen
k214 Bougainvillea
k214 Bougainvillea Scientific Information
Type:
Whole Allergen
Display Name:
Bougainvillea
Family:
Nyctaginaceae
Latin Name:
Bougainvillea spp.
Other Names:
Bougainvillea
Environmental Characteristics
Other topics
Bougainvilleas are beautiful plants indigenous to tropical South America but now found widely in tropical and temperate regions, and even in colder climates in greenhouses. These members of the Nyctaginaceae (Four-O'Clock) family are very vigorous vines, deciduous but long-flowering. Their thin, woody stems are clothed with small, dark-green leaves and sharp thorns. The small, unnoticeable flowers are surrounded by large, papery bracts in a variety of brilliant colours.
Bougainvilleas grow and bloom in cycles. The bloom cycle is usually about 5 to 6 weeks long, and after it all the bracts fall off. The plant then goes into a 6- to 8-week period when no bracts are produced. During this cycle, new leaves and stems grow
Bougainvilleas are immensely popular for their toughness and adaptability, qualities that place them among the world’s most familiar garden plants. They are insect-resistant and do well with minimal care in most soils, as long as they have direct sunlight. The plant can take many forms, from Bonsais, through potted plants of varying sizes, to small flowering trees, to climbing forms to clipped hedges. In a temperate climate, blooming starts early in spring and lasts until the first cold spells. Late in autumn, after blooming has ceased, the foliage turns yellow and drops.
Clinical Relevance
IgE-mediated reactions
Bougainvillea pollen does not appear to be a major component in the atmosphere, as shown by aerobiological monitoring (2), but may nevertheless uncommonly induce symptoms of asthma, rhinitis and conjunctivitis in sensitised individuals
Other topics
A plant species producing pollen, which may infrequently induce hayfever, asthma and conjunctivitis in sensitised individuals.
Molecular Aspects
No allergens from this plant have yet been characterised.
Cross-reactivity
An extensive cross-reactivity among the different individual species of the Nyctaginaceae family could be expected but has not been documented to date (1).
Compiled By
Last reviewed:April 2022
References
  1. Yman L. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. 1982: ISBN 91-970475-09
  2. Mishra RP, Singh B, Oommachan M. Airborne pollen flora of Jabalur - the central India. Aerobiologia 2002;18:73-81