Phadia 200 | Phadia 250 | Phadia 1000 | Phadia 2500+ series | Phadia 5000+ series | |
ImmunoCAP | Allergy | Allergy | Allergy | Allergy | Allergy |
EliA | Autoimmunity |
Autoimmunity | N/A | Autoimmunity | Autoimmunity |
System Description | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system |
Processing | Batch | Continuous | Continuous | Continuous | Continuous |
Maximum Throughput | Up to 96 test results per batch | Up to 60 results/hour | Up to 240 results/hour | Up to 480 results/hour | Up to 960 results/hour |
Sample Types | Serum, Plasma | Serum, plasma | Serum, plasma | Serum, plasma | Serum, plasma |
Sample Tubes for Sample Rack w/ 10 positions |
Height 42-100mm Diam. 10-16mm |
Height 50-110mm Diam. 10-17mm |
Height 50-105mm Diam. 10-17mm |
N/A | N/A |
Sample Tubes for Sample Rack w/ 5 positions |
N/A | N/A | N/A | Height 70-105mm Diam. 10-17mm |
Height 70-105mm Diam. 10-17mm |
Sample Barcode Types | Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Sample Dilution | Allergy Autoimmunity |
Allergy Autoimmunity |
N/A | Autoimmunity | Autoimmunity |
Level and Clot Detection | Yes | Yes | Yes | Yes | Yes |
Reflex Testing | Yes | Yes | Yes | Yes | Yes |
STAT Samples | No | Yes | No | Yes | Yes |
Reagents | General ImmunoCAP and EliA reagents available on board. Loaded batch wise with ImmunoCAP or EliA reagents | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared |
Type of calibration | One Isotype master curve per assay group (IgG, IgE, IgM, IgA) | One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
Calibration stability | 28 days | 28 days | 28 days | 28 days | 28 days |
Water Requirements for Rinse Solution | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* |
Remote Troubleshooting and Support | Phadia LabCommunity |
Phadia LabCommunity | Phadia LabCommunity | Phadia LabCommunity | Phadia LabCommunity |
Ambient Operating Temperature | 18–32 °C |
18–32 °C | 18–32 °C | ImmunoCap 18–32 °C EliA 18-29 °C |
ImmunoCap 18–32 °C EliA 18-29 °C |
Electrical Requirements | 110-240V 50/60Hz |
100 V, 120 V, 220 V, 230 V, 240 V ±10% 50/60 Hz |
200 V, 220 V, 230 V, 240 V±10% 50/60 Hz |
200 V, 208 V, 230 V, 240 V±10% 50/60 Hz |
200 V, 208 V, 230 V, 240 V±10% 50/60 Hz |
Dimensions (HxWxD) | 55 x 80 x 60 cm (22 x 32 x 24 in) | 186x127x75 cm w/ table 73x50x30 in |
215x183x105 cm 85x72x42 in |
195x411x135 cm 77x162x53 in |
195x619x135 cm 77x244x53 in |
Weight (empty) | 47 kg/ 104 lb. | 220 kg / 485 lb | 688 kg / 1516 lb | 2000 kg / 4409 lb | 3342 kg / 7367 lb |
*Clinical laboratory reagent water (CLRW) may be used in place of Type II Laboratory Grade water; Approved Guideline-Fourth Edition.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
With four parallel processing lines, it doubles the capacity of the Phadia 2500+ series of instruments** and provides even greater flexibility. Our flagship solution is for extremely high throughput laboratories that deliver 25,000 to 45,000 results per week.
The three configuration options give you flexibility to acquire the most appropriate instrument to fit your laboratory needs, whether for ImmunoCAP™ only (Phadia 5000), or for both ImmunoCAP and EliA™ (Phadia 5000E and Phadia 5000E+E). This offers your laboratory the ability to customize a high-throughput automated solution that aligns with your testing needs and schedules.
*The Phadia 5000+ series comprises the Phadia 5000, Phadia 5000E, and Phadia 5000E+E instruments.
** The Phadia 2500+ series comprises the Phadia 2500, Phadia 2500E, and Phadia 2500EE instruments.
*** Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.