Make a difference aiding in the diagnosis of connective tissue disease and order EliATM CTD assays for laboratory testing today.
Offered to support a wide variety of diagnostic needs.
Autoantibodies/antigens |
EliA assay name |
Assays within EliA SymphonyS test |
---|---|---|
U1-RNP (A, C, 70 kDa) |
EliA U1RNP |
X |
SmD3 |
EliA SmDP-S |
X |
SS-A/Ro (52 kDa, 60 kDa) |
EliA Ro |
X |
SS-B/La |
EliA La |
X |
Centromere |
EliA CENP |
X |
Topoisomerase I/Scl-70 |
EliA Scl-70S |
X |
Histidyl-tRNA-synthetase/Jo-1 |
EliA Jo-1 |
X |
dsDNA |
EliA dsDNA |
|
RNA Polymerase III |
EliA RNA Pol III |
|
Ribosomal P-protein/Rib-P |
EliA Rib-P |
|
RNP70 |
EliA RNP70 |
|
SS-A/Ro (52 kDa) |
EliA Ro52 |
|
SS-A/Ro (60 kDa) |
EliA Ro60 |
Autoantibodies/antigens |
EliA assay name |
Assays within EliA SymphonyS test |
---|
Less false positives
High specificity due to the selection and use of recombinant antigens in this portfolio means laboratories can help reduce the number of false positives, aiding clinicians in prescribing appropriate therapies.1
Laboratory efficient
On PhadiaTM Laboratory Systems, the EliA portfolio may help to reduce labor costs and hands-on time with onboard dilution, random access, and calibration curves stored for every isotype (IgG, IgA, IgM).2
High sensitivity and specificity1,3,4
Even for difficult-to-detect antibodies, such as anti-SS-A/Ro or anti-Jo-1, high quality EliA antigens can help identify important antibodies associated with CTD.5,6
Many antigens used in EliA CTD assays are manufactured using innovative human recombinant protein technology which allows for optimized accessibility and purity, increasing the clinical utility of these assays. 1,3,4
EliA CTD assays are fully automated solid-phase fluorescence enzyme immunoassays (FEIA) performed on Phadia Laboratory Systems, optimizing laboratory workflow efficiency and throughput while minimizing turnaround time.
View EliA assays in our product catalog
Using tests like the EliA SymphonyS and other tests within the EliA CTD portfolio may help in differentiating between a wide range of connective tissue diseases.
View all EliA assays for connective tissue disease in our product catalog
For additional information on EliA CTD assays, autoimmune diseases, and educational tools and resources, contact our experts.
Whether you’re a healthcare provider or laboratorian, Thermo Fisher Scientific offers valuable resources geared to your specific needs.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.