Type:
Whole Allergen
Whole Allergen
Whole Allergen
Pholcodine
Oral, intravenous
Pholcodine is an antitussive morphine derivative with a structure similar to codeine and to the quaternary ammonium ions found in neuromuscular blocking agents (NMBAs). It has been shown that pholcodine significantly increases levels of IgE antibodies to NMBA in sensitized patients. Cross-inhibition studies indicate that some patients have IgE not only against the ammonium ion, but also against the hydrophobic part of the pholcodine molecule.(1) Cross-reactivity between different NMBA is common since they all share the quaternary ammonium ion allergenic epitope.(2) NMBAs represent a significant cause of anesthesia-related anaphylaxis(3), which is rare but can be severe.(4) Diagnostic approach of anesthesia-related IgE-mediated immediate hypersensitivity reactions (IHR) generally starts with patient’s history, thorough review of the anesthetic/surgical notes complemented with skin prick testing and/or in vitro quantification of specific IgE antibodies.(3)
Pholcodine (3-(2-morpholinoethyl)morphine) (PHO) is a morphine analogue molecule with a morpholino side chain. There is no depression of respiration, pain relief, and CNS excitation, and it is void of euphorizing properties and risk of addiction. PHO is a mild cough suppressant acting directly on the cough center of the CNS, and it has become a widely used cough medicine in several formulations. Pharmacokinetically, it is much more slowly eliminated from the body than opioids like codeine, and the concentration in saliva becomes 3 to 4 times higher than in plasma, properties that might add to its immunogenicity.(5)
Many opioids are potent histamine releasers producing a variety of hemodynamic changes and anaphylactoid reactions, but the relationship of the appearance of these effects to the histamine plasma concentration is complex and there is no direct and invariable relationship between the two.(6)
Usual route of administration for pholcodine is oral.
NMBA are mainly administered intravenously but may also be given intramuscularly.(1)
Opioids provoke non-specific wheals in the skin by causing direct degranulation of mast cells without the involvement of either opioid receptors or opioid-specific IgE antibodies.(6) Genuine IgE-mediated allergies to opiates (morphine, codeine) remain rare notwithstanding their frequent and universal use. Moreover, correct diagnosis is not straightforward, mainly because of uncertainties associated with measurement of drug-specific IgE antibodies and skin testing.(3)
In recent years, increasing evidence for a connection between the consumption of pholcodine and IgE-mediated anaphylactic reactions to neuromuscular blocking agents (NMBA) have been presented.
Anaphylaxis during general anesthesia is rare but can be severe, as it is often complicated by significant morbidity. Identification of the cause of anaphylaxis may pose a significant dilemma to the allergist and anesthetist. Results from a study in the UK showed that when the drug culprit of anaphylaxis was detected NMBA represented 38.1% of the cases.(4)
In many countries, curarizing NMBAs represent a significant cause of anesthesia-related anaphylaxis.(3) Allergy to neuromuscular blocking agents (NMBA) constitutes a major cause of potentially life-threatening perioperative anaphylaxis. In a study with 344 patients, NMBA accounted for 40% of all patients with an IgE-mediated perioperative allergy.(7) However other studies showed even higher values (up to 60%).(1)
A study in Scandinavian countries indicated that there was a large difference in rates of anaphylaxis to NMBAs in Norway (high) and Sweden (low). The predominant difference identified was in the availability and use of pholcodine (PHO was not available in Sweden but was widely used in Norway). It was postulated that the substituted ammonium ion structure of PHO was resulting in sensitization and an increased reaction rate to NMBAs observed in Norway relative to Sweden (this led to the voluntary withdrawal of the single PHO product on the market in Norway).(8) As a result, six years after PHO withdrawal, the Norwegian population has become significantly less IgE-sensitized and clinically more tolerant to NMBAs.(9)
Another survey showed that the mortality rate from an immediate hypersensitivity reaction to NMBA administration was relatively high (4.1%) in France over the period 2000–2011.(10)
The major IgE-binding epitope of the NMBA contains the quaternary ammonium ion (QAI) or its tertiary variety. In their well-known studies Florvaag and Johansson have shown that pholcodine (and its parent molecule morphine -MOR-) contains the QAI epitope, which is shared with NMBAs.
The direct effects of pholcodine on IgE synthesis results in a remarkable polyclonal increase in IgE. However, in addition, another epitope of unknown structure on PHO and morphine, not present on NMBAs, was found based on inhibition studies where IgE antibody binding to QAI (suxamethonium), is completely inhibited with PHO and MOR, but, the binding to PHO and MOR is only partially inhibited with QAI. As a consequence, PHO can in principle not initiate an allergic reaction toward itself through cross-binding of IgE antibodies. Therefore, tests dependent on such cross-binding will come out negative with PHO in individuals who are IgE sensitized to PHO. On the other hand, the NMBAs like suxamethonium and rocuronium are structurally bivalent for the QAI epitope. As such, they appear functionally as allergens that through the cross-binding of IgE antibodies on the surfaces of effector cells can induce immediate allergic inflammatory responses and enhanced by way of administration, eventually leading to anaphylaxis.(5)
At present, the mechanisms underlying why exposure and sensitization to a substituted ammonium ion in pholcodine can prime NMBA anaphylaxis, but not result in allergy to all compounds containing similar substituted ammonium ions, remain unresolved.(11)
One of the major uncertainties in evaluating diagnostic test results in suspected cases of anaphylaxis to anesthetic drugs is the possibility that a negative result to the suspected drug(s) may occur in a subject who experienced some or all of the signs associated with an anaphylactic or anaphylactoid reaction due to another, non-allergic, mechanism. In such cases it is difficult to determine whether the result is due to a weakness inherent in the test, such as lack of sensitivity, or whether the negative finding truly reflects the real situation.(12)
The ‘gold standard" to determine correct diagnosis of immediate hypersensitivity reactions (IHR) to drugs is a controlled drug provocation test (DPT) with the thought culprit compound(s). However, DPTs involve a risk of severe, life-threatening complications and can be contraindicated (e.g. patients having suffered from life-threatening reactions) or impossible (e.g., full-dose DPT in hypersensitivity to curarizing neuromuscular blocking agents (NMBA)). Moreover, the predictive value of DPTs is not known and DPTs might yield false negative results. Therefore, the diagnostic approach of anesthesia-related IgE-mediated IHR generally starts with patient’s history, thorough review of the anesthetic/surgical notes complemented with skin testing and/or in vitro quantification of specific IgE (sIgE) antibodies.(3)
Erroneous opiate allergy diagnosis might not only require unnecessary avoidance actions but also, most importantly, put patients at risk by overlooking alternative diagnoses such as an allergy to rocuronium or suxamethonium (3) or, possibly, other compounds as antibiotics or chlorhexidine.(4)
In cases of non-IgE-mediated IHR, diagnosis might benefit from cellular tests such as basophil mediator release tests and basophil activation tests (BAT).(3)
Cross-reactivity between different NMBA is common since they all share the quaternary ammonium ion allergenic epitope(2). However, the extent of cross-reactivity varies considerably between patients and it is unusual for an individual to be allergic to all NMBA(13). The explanation for this is that IgE antibody paratopes may not only recognize the quaternary ammonium ion; sometimes the molecular environment around the ammonium ion is also part of the allergenic epitope(14). The possibility of multiple allergies should therefore be considered.
Cross-inhibition results indicate that some patients have IgE not only against the ammonium ion but also against the hydrophobic part of the morphine molecule.(1)
Author: Dr. Fabio Iachetti
Reviewer: Dr. Christian Fischer
Last reviewed: August 2020
1. Armentia A, Ruiz-Munoz P, Quesada JM, Postigo I, Herrero M, Martin-Gil FJ, et al. Clinical value of morphine, pholcodine and poppy seed IgE assays in drug-abusers and allergic people. Allergol Immunopathol (Madr). 2013;41(1):37-44.
2. Mertes PM, Laxenaire MC, Alla F, Groupe d'Etudes des Reactions Anaphylactoides P. Anaphylactic and anaphylactoid reactions occurring during anesthesia in France in 1999-2000. Anesthesiology. 2003;99(3):536-45.
3. Ebo DG, Faber M, Elst J, Van Gasse AL, Bridts CH, Mertens C, et al. In Vitro Diagnosis of Immediate Drug Hypersensitivity During Anesthesia: A Review of the Literature. The Journal of Allergy and Clinical Immunology: In Practice. 2018;6(4):1176-84.
4. Meng J, Rotiroti G, Burdett E, Lukawska JJ. Anaphylaxis during general anaesthesia: experience from a drug allergy centre in the UK. Acta Anaesthesiol Scand. 2017;61(3):281-9.
5. Florvaag E, Johansson SG. The Pholcodine Case. Cough Medicines, IgE-Sensitization, and Anaphylaxis: A Devious Connection. World Allergy Organ J. 2012;5(7):73-8.
6. Baldo BA, Pham NH. Histamine-releasing and allergenic properties of opioid analgesic drugs: resolving the two. Anaesth Intensive Care. 2012;40(2):216-35.
7. Leysen J, Uyttebroek A, Sabato V, Bridts CH, De Clerck LS, Ebo DG. Predictive value of allergy tests for neuromuscular blocking agents: tackling an unmet need. Clin Exp Allergy. 2014;44(8):1069-75.
8. McAleer PT, McNicol L, Rose MA. Perioperative Anaphylaxis: Progress, Prevention and Pholcodine Policy. Anaesthesia and Intensive Care. 2017;45(2):147-50.
9. Pater GH, Florvaag E, Johansson SGO, Irgens Å, Petersen MNH, Guttormsen AB. Six years without pholcodine; Norwegians are significantly less IgE‐sensitized and clinically more tolerant to neuromuscular blocking agents. Allergy. 2017;72(5):813-9.
10. Reitter M, Petitpain N, Latarche C, Cottin J, Massy N, Demoly P, et al. Fatal anaphylaxis with neuromuscular blocking agents: a risk factor and management analysis. Allergy. 2014;69(7):954-9.
11. Brusch AM, Clarke RC, Platt PR, Phillips E. Response to letter regarding article 'Exploring the link between pholcodine exposure and neuromuscular blocking agent anaphylaxis'. Br J Clin Pharmacol. 2014;78(4):931-2.
12. Fisher MM, Baldo BA. Immunoassays in the diagnosis of anaphylaxis to neuromuscular blocking drugs: the value of morphine for the detection of IgE antibodies in allergic subjects. Anaesth Intensive Care. 2000;28(2):167-70.
13. Ebo DG, Fisher MM, Hagendorens MM, Bridts CH, Stevens WJ. Anaphylaxis during anaesthesia: diagnostic approach. Allergy. 2007;62(5):471-87.
14. Florvaag E, Johansson SG, Oman H, Venemalm L, Degerbeck F, Dybendal T, et al. Prevalence of IgE antibodies to morphine. Relation to the high and low incidences of NMBA anaphylaxis in Norway and Sweden, respectively. Acta Anaesthesiol Scand. 2005;49(4):437-44.