Table of Contents
Component
d202 Der p 1
d202 Der p 1 Scientific Information
Type:
Component
Name; WHO/IUIS:
Der p 1
Route of Exposure:
Airway (Inhalation)
Biological function:
Group 1 (Cysteine protease)
Allergen code:
d202
Molecular Weight:
25 kDa
Source Material:
Located in the mid-gut and faecal particles of house dust mite. Native sourced from the extracts of Dermatophagoides pteronyssinus or recombinant
Other Names :
Antigen P1, DP1, Gorup 1 Mites
Summary
Der p 1 is one of the major allergens of house dust mite (HDM), with a reported prevalence among Dermatophagoides pteronyssinus (Der p) sensitized patients to be between 70-100%. The exposure of Der p 1 is through inhalation of the mite excrement. Early exposure of dust mite antigen Der p 1 increases the asthma risk up to fivefold. Decrease of the Der p 1 allergen exposure in mite allergic patients have reported a reduction in symptoms of rhinitis and asthma. A high degree of cross-reactivity with Der f 1 (from Dermatophagoides farinae, Der f) is reported. Der p 1 sensitization evaluation is considered important for the selection of patients suitable for specific immunotherapy (SIT). 
Epidemiology
Worldwide distribution
Der p 1 is one of the major mite allergens of house dust mite (HDM) (1). The prevalence of Der p 1 among Dermatophagoides pteronyssinus sensitized patients is reported to be between 70-100%. Der p 1 is found to be present in the mite fecal particles and is strongly associated with asthma (2, 3).
Environmental Characteristics
Source and tissue
Der p 1 is the major allergen component extracted from the mite allergen crude extracts. It is found to be located in the mid-gut and fecal pellets of the Dermatophagoides pteronyssinus (4).
One of the novel methods used for purification or isolation of Der p 1 is Chromatography including chromatography on Resource Q column. The Fractions obtained from Resource Q column are usually confirmed and characterized by mass spectrometry. The molecule can also be generated by recombinant expression systems (5).
Risk factors
An elevation up to fivefold in the asthma risk is observed due to early exposure of the dust mite antigen Der p1 to individuals (6). According to a study conducted, the biological response to HDM allergen, Der p 1, is driven by targeting the sinonasal epithelium. Exposure of Der p 1 HDM antigen to sinonasal epithelium reduces the expression of tight junction proteins (TJPs). This expression in turn may escalate the permeability of antigens across the sinonasal epithelial layer, thus representing a potential mechanism for the exposure of the antigen in allergic rhinitis (7).
Clinical Relevance
Specific molecules
Der p 1 is a papain-like cysteine protease and is reported to be synthetized as a proenzyme. The proteolytic activity of Der p 1 is blocked due to the presence of a cysteine protease domain (222 residues) and an N-terminal pro-peptide (80 amino acids). The performance of pro-peptide of Der p 1, as an intramolecular chaperone, helps in its right folding and the extracellular secretion (8). Der p 1 in particular accounts for 22% of the total protease transcripts (9).
Cross-reactive molecules
A high degree of cross-reactivity pertaining to T-cell was observed between the group 1 and group 7 allergens of HDM denoted by the proliferative and cytokine response to the group 1 and group 7 allergens (10). Currently, in the literature, the structures of the Der p 1-specific epitopes for monoclonal antibodies (mAb) 5H8 and 10B9 are reported. The cross-reactivity between mAb 4C1 and Der p 1 was inhibited by the Der p 1-specific mAb 10B9, but not to Der f 1. In the study, most mAb produced against Der p 1 were directed to the identical epitope (mAb 5H8; 71.4%) and 4 were directed to a different epitope (mAb 10B9; 11.4%) (11).
Prevention and Therapy
Experimental trials
According to a meta-analysis of the five trials, major allergens including Der p 1 are responsible to sensitize a high percentage of HDM patients with allergy. The standardized quality (SQ) HDM sublingual immunotherapy (SLIT) tablet was reported to be superior in improving the symptoms in patients with allergic rhinitis and asthma outcomes. Furthermore, the effectiveness of the tablet was shown for a wide range of patients, and was explained by the presence of 1:1 ratio of the two major HDM allergens to which individuals were most frequently sensitized (12).
Molecular Aspects
Biochemistry
Der p 1 of HDM has a molecular weight of 25 kDa glycoprotein (13). The HDM allergens are reported to pass across the epithelial surface by disrupting the tight junction between the epithelial cells through the proteases present in the mites. The extracts of Der p 1 also demonstrate the desquamation of the epithelial cells in a protease-dependent manner which facilitates the entry of the allergen inside the airway wall. This process induces the release of pro-inflammatory cytokines (interleukin 6 and interleukin 8, through protease-activated receptors) in response to proteases present in HDM extract, leading to inflammatory responses (14).
Cross-reactivity
A high degree of cross-reactivity is noted between Dermatophagoides pteronyssinus and Dermatophagoides farinae extracts, however, the reactivity between Dermatophagoides and Blomia tropicalis is low. One of the main causes of cross-reactivity among mites, cockroaches, shellfish, helminths is due to Tropomyosin (Der p 10), however, glutathione transferase may also be included. In cases where genuine sensitization is unclear, specific allergen components can be useful to identify primary allergy (2).
The structural similarities between Der p 1 and Der f 1 were demonstrated with an X-ray crystal structure analysis. The analysis showed surface conservation of a crystal structure of natural Der f 1 with Der p 1 of 71% amino acid similarity along with an overlapping catalytic site. The structural similarities between Der p 1 and Der f 1 are commonly believed to be the basis for cross-reactivity among the 2 allergens (11, 15).
Diagnostic Relevance
Disease Severity
The measurement of IgE levels to native Der p 1 is a useful diagnostic tool for the assessment of genuine HDM allergy. A study was conducted in 55 DP-sensitized asthmatic patients, wherein, 89% positivity for IgE antibodies to native Der p 1was noted. The area under the receiver operating characteristic curves for IgE was found as a predictor of immediate asthmatic response (IAR) with a value of 0.913. The specificity of IgE to Der p 1 was higher than IgE to crude DP. IgE to Der p 1 was highly predictive of allergen-induced IAR (16).
AIT Prescription
Der p 1 sensitization evaluation is considered important for the selection of patients suitable for allergen immunotherapy using HDM extract (16).
Allergen components, including Der p 1 may be helpful in establishing the indication for AIT (17). According to a trial conducted, SLIT demonstrated its safety and effectiveness in primary house dust mite allergy-induced asthma in adolescents aged 14 years and in adults (18).
Exposure
The exposure is through inhalation of the mite fecal pellets. Exposure of Der p 1 is reported to exhibit cysteine protease activity that may result in epithelial cell desquamation, the release of cytokines, and in turn, leads to allergen expression across the epithelial cell layer. The desquamation further results in allergenic compound penetration deep into the airway wall (14).
Patients with mite allergy have reported a decrease in symptoms of rhinitis and asthma after an exposure reduction of the Der p 1 allergens from 13 μg/g to 0.2 μg/g (2).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: October  2020
References
  1. Calderon MA. House Dust Mite Respiratory Allergy: An Overview of Current Therapeutic Strategies. J Allergy Clin Immunol Pract. 2015;3(6):843-55.
  2. Matricardi PM. EAACI Molecular Allergology User's Guide. DUST MITE ALLERGY. Part B USING MOLECULAR ALLERGOLOGY IN THE CLINICAL PRACTICE. Chapter B04. Pediatr Allergy Immunol. 2016;27 Suppl 23:105-13.
  3. Platts-Mills TA, Tovey ER, Mitchell EB, Moszoro H, Nock P, Wilkins SR. Reduction of bronchial hyperreactivity during prolonged allergen avoidance. Lancet. 1982;2(8300):675-8.
  4. Herman J. Der p 1 is the primary activator of Der p 3, Der p 6 and Der p 9 the proteolytic allergens produced by the house dust mite Dermatophagoides pteronyssinus. Biochimica et Biophysica Acta 1840 (2014) 1117–1124. 2014.
  5. Zhang. Novel Method for the Purification of House Dust Mite Allergen Der p 1 and Its Use in Structure-Based Chemical Design of Novel Inhibitors. 2019:185-205.
  6. Thomson. ASTHMA | Extrinsic/Intrinsic. Encyclopedia of Respiratory Medicine 2006, Pages 206-215. 2006.
  7. Henriquez OA, Den Beste K, Hoddeson EK, Parkos CA, Nusrat A, Wise SK. House dust mite allergen Der p 1 effects on sinonasal epithelial tight junctions. Int Forum Allergy Rhinol. 2013;3(8):630-5.
  8. Gorines. Der p 1-based immunotoxin as potential tool for the treatment of dust mite respiratory allergy. 2020.
  9. Randall TA. Proteases of Dermatophagoides pteronyssinus. Int J Mol Sci. 2017;18(6).
  10. Hales BJ, Shen HD, Thomas WR. Cross-reactivity of T-cell responses to Dermatophagoides pteronyssinus and D. farinae. Studies with group 1 and 7 allergens. Clin Exp Allergy. 2000;30(7):927-33.
  11. Glesner J, Vailes LD, Schlachter C, Mank N, Minor W, Osinski T, et al. Antigenic Determinants of Der p 1: Specificity and Cross-Reactivity Associated with IgE Antibody Recognition. J Immunol. 2017;198(3):1334-44.
  12. Nolte, Plunkett G, Grosch K, Larsen JN, Lund K, Bollen M. Major allergen content consistency of SQ house dust mite sublingual immunotherapy tablets and relevance across geographic regions. Ann Allergy Asthma Immunol. 2016;117(3):298-303.
  13. Reithofer M, Jahn-Schmid B. Allergens with Protease Activity from House Dust Mites. Int J Mol Sci. 2017;18(7).
  14. Kauffman HF, Tamm M, Timmerman JA, Borger P. House dust mite major allergens Der p 1 and Der p 5 activate human airway-derived epithelial cells by protease-dependent and protease-independent mechanisms. Clin Mol Allergy. 2006;4:5.
  15. Chruszcz M, Chapman MD, Vailes LD, Stura EA, Saint-Remy JM, Minor W, et al. Crystal structures of mite allergens Der f 1 and Der p 1 reveal differences in surface-exposed residues that may influence antibody binding. J Mol Biol. 2009;386(2):520-30.
  16. Minami T, Fukutomi Y, Lidholm J, Yasueda H, Saito A, Sekiya K, et al. IgE Abs to Der p 1 and Der p 2 as diagnostic markers of house dust mite allergy as defined by a bronchoprovocation test. Allergol Int. 2015;64(1):90-5.
  17. Pfaar O, Bachert C, Bufe A, Buhl R, Ebner C, Eng P, et al. Guideline on allergen-specific immunotherapy in IgE-mediated allergic diseases: S2k Guideline of the German Society for Allergology and Clinical Immunology (DGAKI), the Society for Pediatric Allergy and Environmental Medicine (GPA), the Medical Association of German Allergologists (AeDA), the Austrian Society for Allergy and Immunology (OGAI), the Swiss Society for Allergy and Immunology (SGAI), the German Society of Dermatology (DDG), the German Society of Oto- Rhino-Laryngology, Head and Neck Surgery (DGHNO-KHC), the German Society of Pediatrics and Adolescent Medicine (DGKJ), the Society for Pediatric Pneumology (GPP), the German Respiratory Society (DGP), the German Association of ENT Surgeons (BV-HNO), the Professional Federation of Paediatricians and Youth Doctors (BVKJ), the Federal Association of Pulmonologists (BDP) and the German Dermatologists Association (BVDD). Allergo J Int. 2014;23(8):282-319.
  18. Mosbech H, Deckelmann R, de Blay F, Pastorello EA, Trebas-Pietras E, Andres LP, et al. Standardized quality (SQ) house dust mite sublingual immunotherapy tablet (ALK) reduces inhaled corticosteroid use while maintaining asthma control: a randomized, double-blind, placebo-controlled trial. J Allergy Clin Immunol. 2014;134(3):568-75 e7.