Whole Allergen
f13 Peanut
f13 Peanut Scientific Information
Type:
Whole Allergen
Display Name:
Peanut
Allergen code:
f13
Family:
Fabaceae
Genus:
Arachis
Species:
A. hypogaea
Route of Exposure:
Ingestion
Source Material:
Shelled nuts
Latin Name:
Arachis hypogaea
Other Names:
Groundnut, Monkeynut
WHO/ICD-11 code:
XM6Q82
(ICD-11 is currently under implementation and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Peanuts are consumed across the world, however, the form they are consumed in varies geographically and this can impact their allergenicity. Currently, 17 peanut allergens have been identified and the seed storage proteins being stable to cooking and digestion. Ara h 1, 2, 3 and 6 are considered to be the major peanut allergens and it is estimated that 97% of peanut allergy patients are sensitized to at least one of the allergens Ara h 1, 2 and 3. Peanut allergens belong to diverse protein families leading to immunochemical IgE-mediated cross-reactions among different members of the legume families, other plant foods such as tree nuts and also pollen. Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime, however, approximately 20% of young children develop tolerance. Typical clinical symptoms of peanut allergy range from angioedema, urticaria, nausea, abdominal pain, vomiting, wheezing, and breathlessness which usually occur soon after peanut ingestion. A significant proportion of sensitized individuals do not show clinical signs of peanut allergy. Sensitization to the storage proteins, Ara h 1, 2, 3, 6, and 7, carries an increased risk for more severe symptoms and anaphylactic reactions. Individuals with peanut allergy are of elevated risk of anaphylaxis compared to other food allergies, with over 90% of food-induced anaphylaxis-related fatalities occurring in peanut sensitized individuals. There are several prevention strategies including introducing peanuts to infants at an early stage, avoidance and ‘peanut-free’ schools have been proposed. However, the Food and Drug Administration (FDA) approved an oral immunotherapy for clinical use in the USA in 2020 which may be more widely available in the future. 
Allergen
Summary
Peanuts are consumed across the world, however, the form they are consumed in varies geographically and this can impact their allergenicity. Currently, 17 peanut allergens have been identified and the seed storage proteins being stable to cooking and digestion. Ara h 1, 2, 3 and 6 are considered to be the major peanut allergens and it is estimated that 97% of peanut allergy patients are sensitized to at least one of the allergens Ara h 1, 2 and 3. Peanut allergens belong to diverse protein families leading to immunochemical IgE-mediated cross-reactions among different members of the legume families, other plant foods such as tree nuts and also pollen. Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime however, approximately 20% of young children develop tolerance. Typical clinical symptoms of peanut allergy range from angioedema, urticaria, nausea, abdominal pain, vomiting, wheezing, and breathlessness which usually occur soon after peanut ingestion. A significant proportion of sensitized individuals do not show clinical signs of peanut allergy. Sensitization to the storage proteins, Ara h 1, 2, 3, 6, and 7, carries an increased risk for more severe symptoms and anaphylactic reactions. Individuals with peanut allergy are of elevated risk of anaphylaxis compared to other food allergies, with over 90% of food-induced anaphylaxis-related fatalities occurring in peanut sensitized individuals. There are several prevention strategies including introducing peanuts to infants at an early stage, avoidance and ‘peanut-free’ schools have been proposed. However, the Food and Drug Administration (FDA) approved an oral immunotherapy for clinical use in the USA in 2020 which may be more widely available in the future.
Nature
The budding ovary or “peg” grows down towards the soil. The peanut embryo burrows into the soil surface (geocarpy) and begins to mature which is atypical compared to other legume plants (1).
Peanut contains up to 32 different proteins, of which at least 18 have been identified as being capable of binding allergen specific IgE antibodies (2). Varieties of peanuts from different parts of the world contain similar proteins, including Ara h 1 and Ara h 2, and the IgE-binding properties have also been reported to be similar (3).
Taxonomy
The peanut (Arachis hypogaea) belongs to the legume family (Leguminosae) (4). Many different cultivars are known. Hybridization may not affect the allergenicity of peanuts, with high-oleic peanuts, known as the SunOleic type, showing no difference in allergenicity (5). Similarly, no difference in the allergic components of either raw or roasted extracts of Korean or American Peanuts could be demonstrated (6).
Cross-reactivity exists among legumes because they have structurally homologous proteins and share common epitopes. However, peanut allergy associated with other legumes is less frequently reported compared to the cross-reactivity demonstrated for other types of legumes (7). The taxonomic classification of peanut and tree nuts does not appear to predict allergenic cross-reactivity (8).
Tissue
Peanuts are consumed across the world, however, the form they are consumed in varies geographically. In addition to the global consumption of raw peanuts, a variety of peanut products are available such as roasted peanuts, peanut butter, oil, paste, sauce, flour, milk, peanut drinks, snacks and peanut cheese equivalent (9).
Epidemiology
Worldwide distribution
Studies have typically reported peanut allergy prevalence rates between 1-2% in Western nations. Peanut allergy appears to be less common in Asia and other global areas, although epidemiological studies in non-Western regions have been sparse (10). Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime however, approximately 20% of young children develop tolerance (11).
In 2002, the results from a USA survey indicated that about 0.8% of children had an allergy to peanuts (12) with the prevalence estimated to be 1.4% in 2008 (13). The prevalence of peanut allergy in a study of 7768 primary school children in Montreal, Canada, was 1.50%. When data concerning the peanut allergy status of non-responders were also included, the estimated prevalence was 1.34% (14).
According to a population-based study of children aged three to four years old in the United Kingdom, the prevalence of sensitization to peanuts increased three-fold from 1.1% in 1989 to 3.3% in 1994 to 1996. However, of 41 sensitized children in the study, 10 reported a convincing clinical reaction to peanut, and eight had positive oral challenge results, giving an overall estimate of peanut allergy of 1.5% (18/1246) (15). Similarly, the population-based study in Australia, HealthNuts, reported a challenge-confirmed peanut allergy prevalence of 1.9% in children aged four years (16). In European school children aged six to 10 years previously involved in the EuroPrevall cohort (except for the centers in Italy), three of 2097 children had positive double-blind, placebo-controlled oral food challenges (DBPCFC) to peanut equating to a prevalence of 0.1% (17). The EuroPrevall-INCO prevalence of ‘probable’ peanut allergy (defined as allergic symptoms within two hours of ingestion and positive IgE and/or skin prick test result) in Hong Kong, Guangzhou and Shaoguan (China), Tomsk (Russia) and India were, 0.10, 0.00, 0.00, 0.08 and 0.03%, respectively (18). 
Risk factors (WESTERN)
First sensitization has been attributed to the presence of peanut allergen in breast milk. In a study involving eight infants with immediate hypersensitivity reactions to foods, including peanut, occurring at the first-known exposure, the most likely route of sensitization was thought to be breast milk, and reactions were thought to be dose-dependent (19).
Differences in the way peanuts are prepared may contribute to the variations in the prevalence of peanut allergy in different parts of the world. Frying or boiling peanuts, commonly used in China, appears to reduce the allergenicity of peanuts compared with dry roasting, widely used in the US, which has shown to increase allergenicity. This could help to explain why despite a higher rate of peanut consumption in China, peanut allergy is lower than the United States (20).
A factor postulated to have contributed to peanut allergy in the UK is the cutaneous exposure to ultra-low doses of peanut antigens in peanut oil found in diaper rash emollients, which are applied to the skin of infants with eczema or diaper rash (21).
Peanut allergy has been reported to have been transferred through a combined liver and kidney transplant, bone marrow and peripheral blood stem cell transplantations (22-24).
There is evidence demonstrating that minute quantities of ingested peanut can cause allergic reactions, including fatal and very severe reactions, for example when ingesting undeclared peanut flour which was used in a food product as a flavoring. There are also reports highlighting that peanut-sensitive people can experience allergic reactions when in close proximity to peanuts (25).
Pediatric issues
Infants have reacted adversely to breast feeds after maternal consumption of peanut (26). Sensitization has been reported with use of products containing peanut oil such as vitamins and infant milk formula, and also topically in creams and lotions (21, 26).
Casual exposure to peanut butter (skin contact and inhalation of fumes) is unlikely to cause significant allergic reactions. Although the authors warn that the results cannot be generalized to larger exposures or to contact with peanut in other forms (flour and roasted peanuts) (27). However, the authors who published a series of five case reports concluded that severe food allergic reactions can occur from exposure to small amounts of allergens via skin contact or inhalation. Examples include a nine-month-old male who developed generalized urticaria after his brother ate a peanut butter sandwich and touched his bare leg. Similarly, a child developed wheezing when entering the classroom of a teacher who had just eaten peanuts (28). Although causal exposure (proximity to peanut/’airborne’ exposure, skin contact, kissing) poses a minimal risk, unintended ingestion could lead to a reaction (29).
Peanut allergy usually begins in childhood and persists throughout the affected individual’s lifetime however, approximately 20% of young children develop tolerance (11). Higher peanut-specific IgE levels are associated with both persistence of peanut allergy and more severe allergic reactions when peanuts are consumed (30).
Route of Exposure
Main
The main route of exposure is through consumption of peanuts or products containing peanuts and there is evidence that roasting increases the allergenicity of peanuts (5, 31). In fact, extracts of roasted peanuts where shown to bind to serum IgE from allergic people at approximately 90-fold higher levels compared to raw peanuts (31). The protein concentration is higher in raw peanuts (approx. 16.6 g per 100 g) than in roasted peanuts (approx. 2.6 g per 100 g), and the histamine concentration is higher in roasted peanuts. It has been suggested that an increased amount of histamine in roasted peanuts could worsen IgE-related allergies (32).
Frying or boiling peanuts, commonly used in China, appears to reduce the allergenicity of peanuts compared with dry roasting which has shown to increase the allergenicity of peanuts, widely used in the United States. This could help to explain why despite a higher rate of peanut consumption in China, peanut allergy is lower than that in the United States (20).
Secondary
A series of five case reports concluded that severe food allergic reactions can occur from exposure to small amounts of allergens via skin contact or inhalation (28). However, causal exposure (proximity to peanut/’airborne’ exposure, skin contact, kissing) poses a minimal risk of an allergic reaction but if unintended ingestion occurs, this could lead to an allergic reaction (29). A major concern has been exposure to peanut within airplanes and many airlines have decided to ban peanuts onboard because of risk of airborne exposure. Peanut proteins have been found in the ventilation filters of airplanes, however, these results have not been replicated (33). Peanut proteins have recently been detected on seat and tray tables of commercial irrespective of whether peanuts were served during the flight (34). The sample with the highest amount of peanut protein was taken after two bags of roasted peanuts were consumed and small peanut particles were present on the filter. Currently, the clinical relevance of these results are unknown (34).
Other topics
Peanut allergens have been shown in breast milk and may sensitize infants. In a study of 23 lactating women given 50 g of dry roasted peanuts to eat, peanut protein was detected in the breast milk of 11. It was detected in 10 participants within two hours of ingestion and in one participant within six hours. The median peak peanut protein concentration in breast milk was 200 ng/ml (range, 120-430 ng/ml). Both of the major peanut allergens, Ara h 1 and Ara h 2, were detected (35). Although peanut allergens in breast milk decline over time, a study showed the Ara h6 allergen was detected in breast milk 26 hours after ingestion (36).
Clinical Relevance
Typical clinical symptoms of peanut allergy range from angioedema, urticaria, nausea, abdominal pain, vomiting, wheezing, and breathlessness which usually occur soon after peanut ingestion (37). A significant proportion of sensitized individuals (have specific IgE) do not show clinical signs of peanut allergy (37).
Cutaneous
The most frequently encountered clinical signs of peanut allergy at initial presentation are cutaneous and typically involve urticaria, erythema and angioedema (38). The cutaneous clinical signs associated with peanut allergy are considered to be mild to moderate (39).
Respiratory
The second most frequently experienced clinical signs of peanut allergy at initial presentation involve the respiratory system. Patients can experience wheezing, stridor, coughing, dyspnea, tightness in the throat and/or chest, vocal changes and nasal congestion which are indicative of a more severe reaction to peanuts (38, 39).
Gastrointestinal
Vomiting, diarrhea and abdominal pain are estimated to be experienced by approximately a quarter of peanut-allergic patients on first presentation (38). In terms of severity, the GI signs are usually mild to moderate (39).
Anaphylaxis
Peanut allergy is of elevated risk compared to other food allergies, with over 90% of food-induced anaphylaxis-related fatalities occurring in peanut sensitized individuals (Ho, Wong et al. 2014). Asthma is a key risk factor for fatal and near-fatal food induced anaphylaxis with evidence to show that children over three years old with peanut allergy have a 2.3 times greater risk of hospitalization for asthma (40). A study on peanut or tree nut allergies demonstrated that patients with a history of severe asthma were at increased risk of life-threatening bronchospasm occurring after consuming nuts (41). In a study of children with peanut allergy, 9% (4/46) died from an exacerbation of asthma that indicating a higher fatality rate for people with asthma (41).
Atopic Dermatitis
Studies have shown that the prevalence of food allergy in patients with atopic dermatitis ranges from 20% to 80%. Peanuts are considered to be a common food trigger of atopic dermatitis and research has demonstrated that food allergy plays a major role in exacerbating symptoms of atopic dermatitis and diet exclusion will decrease the severity (42). In an Australian study it has been shown that infants with atopic dermatitis were 11-times more likely to develop peanut allergy than infants without atopic dermatitis (43). 
Other diseases
Peanuts are also one of the foods most commonly involved in food-dependent exercise-induced anaphylaxis (44). In addition, the association of food allergies and otitis media with effusion (OME) was investigated. One study reported that 78% of 104 children with recurrent OME were sensitized to one or more food allergens and 86% saw an improvement in clinical signs of OME when the suspected food was eliminated from the diet (45).
Rhinitis is not typically associated with food-induced allergic reactions (46, 47). However, peanut was found to be one of the most prevalent food allergies in adults with allergic rhinitis (48).
Diagnostics
The gold standard test for diagnosing peanut allergy and other food allergies is using an oral food challenge. However, it poses a risk of severe reactions and is time consuming (38). 
Prevention and Therapy
Allergen immunotherapy
In a multicenter, phase three, double-blind, placebo-controlled food challenge, participants with peanut allergy were randomized in a 3:1 ratio to receive a peanut-derived investigational biologic oral immunotherapy drug, AR101 or placebo in an increasing-dose protocol. The primary endpoint was the proportion of participants aged four to 17 who could ingest a challenge dose of 600 mg or more, without dose-limiting symptoms (49). Of the 496 participants who were four to 17 years old, 250 of 372 participants (67%) who received active treatment, as compared with five of 124 participants (4%) who received placebo, were able to ingest a dose of 600 mg or more of peanut protein, without dose-limiting symptoms (difference, 63.2 percentage points; 95% confidence interval, 53.0 to 73.3; P<0.001) (49). In 2020, oral immunotherapy for peanut allergy was approved by the Food and Drug Administration (FDA) for clinical use in the USA (50). The treatment will most likely be available within EU during 2021.
Prevention strategies
Introducing peanuts to infants early, significantly decreased the occurrence of the development of peanut allergy among children at increased risk for peanut allergy and modulated immune responses to peanuts (51). Egg allergy and/or severe eczema appear to be useful criteria for identifying high-risk infants (52). Avoiding and minimizing symptoms of food allergies are focused on avoidance and prompt emergency management (including the use of self-administered epinephrine) (47). Since individuals with peanut allergy can have serious reactions, banning peanut consumption within school premises has been suggested (53). Studies from different countries have evaluated ‘peanut-free’ schools as a possible prevention strategy although further research is required to evaluate the effect of allergen-restrictive policies on improving safety for students with food allergy (10, 54).
Molecular Aspects
Allergenic molecules
At the time of writing, 17 peanut allergens have been identified. The biochemical properties and molecular mass of peanut allergens are summarized below (55).
Peanut Allergen Biochemical property Molecular weight (kDa)
Ara h 1 Cupin (Vicilin-type, 7S globulin)  64
Ara h 2 Conglutin (2S albumin) 17
Ara h 3 Cupin (Legumin-type, 11S globulin, glycinin) 60 (37 – fragment)
Ara h 5 Profilin 15
Ara h 6 Conglutin (2S albumin) 15
Ara h 7 Conglutin (2S albumin) 15
Ara h 8 Pathogenesis-related protein, PR-10, Bet v 1 family member 17
Ara h 9 Non-specific lipid-transfer protein type 1 9.8
Ara h 10 Oleosin 16
Ara h 11 Oleosin 14
Ara h 12 Oleosin 8
Ara h 13 Oleosin 8
Ara h 14 Oleosin 17.5
Ara h 15 Oleosin 17
Ara h 16 Non-specific lipid-transfer protein type 2 8.5
Ara h 17 Non-specific lipid-transfer protein type 1 11
Ara h 18 Cyclophilin 18
To date, a number of peanut allergens have been identified. Many of them have protective functions or are seed storage proteins. Peanut allergens belong to diverse protein families leading to immunochemical IgE-mediated cross-reactions among different members of the legume families but also other plant foods such as tree nuts (Bublin and Breiteneder 2014). Peanut allergens can be grouped according to their components; cupins (Ara h 1 and 3), conglutins (Ara h 2, 6 and 7), profilin (Ara h 5), PR-10 protein (Ara h 8), oleosins (Ara h 10, 11, 14, and 15), non-specific lipid transfer proteins (Ara h 9, 16, 17), defensins (Ara h 12 and 13) and cyclophilins (Ara h 18) (56, 57).
Currently, Ara h 1, 2, 3 and 6 are considered to be the major peanut allergens (56, 58). It is estimated that 97% of peanut allergy patients are sensitized to at least one of the allergens Ara h 1, 2 and 3 and these components provide >30% of the total protein content of peanuts (56, 59).
Using sera from 40 peanut-allergic patients, 14 individual recognition patterns were identified and the following frequency of specific IgE binding emerged demonstrating that Ara h 2 had the highest frequency of recognition: Ara h 1 was recognized by 65%, Ara h 2 by 85%, Ara h 4 (which is now known to be an isoform of Ara h 3, Ara h 3.02 (56)) by 53%, Ara h 5 by 13%, Ara h 6 by 38% and Ara h 7 by 43% of the sera (60).
Another study involving 30 peanut-allergic individuals, the majority of patients with a positive SPT were sensitized to Ara h 2 (83%) and Ara h 6 (87%). Sixteen patients (53%) were sensitized to Ara h 1 and half of the patients to Ara h 3. In addition, patients with a positive SPT to Ara h 1 and/or Ara h 3 were also sensitized to Ara h 2 and/or Ara h 6 (61).
Biomarkers of severity
Individuals who are sensitized to the storage proteins, Ara h 1, 2, 3, 6, and 7, are at an increased risk for more severe symptoms and anaphylactic reactions (62). A study found specific IgE was positively correlated with clinical severity for Ara h 1, 2 and 3 in adult patients but this trend was not observed in children (63). In addition, another study found that sensitization to rAra h 2 and rAra h 1 and/or rAra h 3 appeared to be predictive of more severe reactions (64). Similarly, the results of a recent cross-sectional study involving 222 children in Australia found that polysensitization to Ara 1, 2 and 3 can help to predict the severity of reaction at challenge (65).
The results of a meta-analysis study on the diagnostic accuracy of peanut components demonstrated that specific IgE to Ara h 1, 2 and 3 are highly specific for peanut allergy in children, however, Ara h 8 and 9 are of low clinical relevance (66). In addition, the authors suggest that specific IgE to Ara h 2 can reduce the number of oral food challenges in unclear cases and has high diagnostic accuracy for peanut allergy in children across geographic locations (66).
Cross-reactivity
Peanut and tree nut show cross reactivity in 25 to 50% of peanut-allergic patients because Ara h 2 shares IgE-binding epitopes with almond and Brazil nut allergens (7). However, despite 2S seed albumin allergens sharing structural similarities, a major peanut allergen, Ara h 2, showed no structural homology with the corresponding regions of Walnut Jug r 1, Pecan Car i 1 or Brazil nut Ber e 1 (67). Although there is a theory of cross reactivity being dependent on allergens sharing a similar sequence and/or structure, there is experimental data highlighting a lack of relationship between the percentage of shared identity and the ability to bind IgE (68). Similarities in physicochemical properties to known IgE epitopes of 2S albumins could account for clinically observed cross-sensitivity between peanuts and tree nuts (68).
Patients with peanut allergy may demonstrate sensitization to other legumes, for example soybean, lupin, lentil or pea, although currently limited data are available (69). Cross-reactivity exists among legumes because they have structurally homologous proteins and share common epitopes. However, peanut allergy associated with other legumes is less frequently reported compared to the cross-reactivity demonstrated for other types of legumes (7) Clinically, peanut allergen cross-reactivity can be relevant but in some cases, irrelevant with approximately half of patients having positive SPTs to other legumes, however, less than 5% show clinical signs when consuming them (58).
The Bet v 1 homologous peanut allergen Ara h 8, is thought to be involved in the cross reactivity of peanut allergy patients. Twenty patients with peanut and birch pollen allergy, and a positive double-blind, placebo-controlled food challenge result to peanut, all experienced symptoms in the oral cavity, progressing to more severe symptoms in 40% of patients (70). Recombinant Ara h 8-specific IgE was demonstrated in 85%, IgE binding to Ara h 8 was inhibited by Bet v 1 and recombinant Ara h 8 inhibited IgE binding to peanut in four of seven tested patient sera. The study concludes that peanut allergy might be mediated in a subgroup of the patients by cross-reactivity of Bet v 1 with the homologous peanut allergen Ara h 8 (70). Similarly, examination of the sera of five patients in relation to four recombinant allergens led to the conclusion that IgE cross-reactivity existed between Bet v 1 (birch pollen) and its homologues Gly m 4 from soybean, Ara h 8, and Pru av 1 from Cherry but variation in IgE specificity between patients was observed (71). In addition, Ara h 8 is involved in grass pollen-associated food allergy (69).
Lupins are an emerging cause of food allergy because of recent large-scale introduction as an additive to wheat flour or the use of lupin flour in processed foods and is frequently used in Europe and Australia (72, 73). They are legumes of the Fabaceae family with PR-10 white lupin sharing significant sequence homology and molecular similarity between the peanut allergen Ara h 8 (72, 73). Sanz et al. (2010) explained that in patients with clinical sensitization to a legume, particularly peanut, greater caution is required depending on the population with lupin cross-reactivity of approximately 5% in British and Norwegian populations, 17% to 68% for French and Belgian populations, and 32% for Danish populations (73).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Magnus Borres & Dr. Eva Södergren
Last reviewed: July 2023
References
  1. Bertioli DJ, Seijo G, Freitas FO, Valls JFM, Leal-Bertioli SCM, Moretzsohn MC. An overview of peanut and its wild relatives. Plant Genetic Resources. 2011;9(1):134-49.
  2. Dean TP. Immunological responses in peanut allergy. Clin Exp Allergy. 1998;28(1):7-9.
  3. Koppelman SJ, Vlooswijk RA, Knippels LM, Hessing M, Knol EF, van Reijsen FC, et al. Quantification of major peanut allergens Ara h 1 and Ara h 2 in the peanut varieties Runner, Spanish, Virginia, and Valencia, bred in different parts of the world. Allergy. 2001;56(2):132-7.
  4. Gepts P, Beavis WD, Brummer EC, Shoemaker RC, Stalker HT, Weeden NF, et al. Legumes as a Model Plant Family. Genomics for Food and Feed Report of the Cross-Legume Advances through Genomics Conference. Plant Physiology. 2005;137(4):1228-35.
  5. Chung SY, Maleki S, Champagne ET, Buhr KL, Gorbet DW. High-oleic peanuts are not different from normal peanuts in allergenic properties. J Agric Food Chem. 2002;50(4):878-82.
  6. Park CW, Kim GI, Lee CH. A comparison study on allergen components between Korean (Arachis fastigiata Shinpung) and American peanut (Arachis hypogaea Runner). J Korean Med Sci. 2000;15(4):387-92.
  7. Verma AK, Kumar S, Das M, Dwivedi PD. A Comprehensive Review of Legume Allergy. Clinical Reviews in Allergy & Immunology. 2013;45(1):30-46.
  8. de Leon MP, Glaspole IN, Drew AC, Rolland JM, O'Hehir RE, Suphioglu C. Immunological analysis of allergenic cross-reactivity between peanut and tree nuts. Clin Exp Allergy. 2003;33(9):1273-80.
  9. Arya SS, Salve AR, Chauhan S. Peanuts as functional food: a review. J Food Sci Technol. 2016;53(1):31-41.
  10. Lieberman JA, Gupta R, Knibb RC, Haselkorn T, Tilles S, Mack DP, et al. The Global Burden of Illness of Peanut Allergy: A Comprehensive Literature Review. Allergy. 2021;76(5):1367–84.
  11. Sicherer SH. Clinical update on peanut allergy. Ann Allergy Asthma Immunol. 2002;88(4):350-61; quiz 61-2, 94.
  12. Sicherer SH, Muñoz-Furlong A, Sampson HA. Prevalence of peanut and tree nut allergy in the United States determined by means of a random digit dial telephone survey: A 5-year follow-up study. Journal of Allergy and Clinical Immunology. 2003;112(6):1203-7.
  13. Sicherer SH, Muñoz-Furlong A, Godbold JH, Sampson HA. US prevalence of self-reported peanut, tree nut, and sesame allergy: 11-year follow-up. Journal of Allergy and Clinical Immunology. 2010;125(6):1322-6.
  14. Kagan RS, Joseph L, Dufresne C, Gray-Donald K, Turnbull E, Pierre YS, et al. Prevalence of peanut allergy in primary-school children in Montreal, Canada. J Allergy Clin Immunol. 2003;112(6):1223-8.
  15. Grundy J, Matthews S, Bateman B, Dean T, Arshad SH. Rising prevalence of allergy to peanut in children: Data from 2 sequential cohorts. J Allergy Clin Immunol. 2002;110(5):784-9.
  16. Peters RL, Koplin JJ, Gurrin LC, Dharmage SC, Wake M, Ponsonby AL, et al. The prevalence of food allergy and other allergic diseases in early childhood in a population-based study: HealthNuts age 4-year follow-up. J Allergy Clin Immunol. 2017;140(1):145-53.e8.
  17. Grabenhenrich L, Trendelenburg V, Bellach J, Yürek S, Reich A, Fiandor A, et al. Frequency of food allergy in school-aged children in eight European countries-The EuroPrevall-iFAAM birth cohort. Allergy. 2020;75(9):2294-308.
  18. Li J, Ogorodova LM, Mahesh PA, Wang MH, Fedorova OS, Leung TF, et al. Comparative Study of Food Allergies in Children from China, India, and Russia: The EuroPrevall-INCO Surveys. J Allergy Clin Immunol Pract. 2020;8(4):1349-58.e16.
  19. van Asperen PP, Kemp AS, Mellis CM. Immediate food hypersensitivity reactions on the first known exposure to the food. Arch Dis Child. 1983;58(4):253-6.
  20. Beyer K, Morrow E, Li XM, Bardina L, Bannon GA, Burks AW, et al. Effects of cooking methods on peanut allergenicity. J Allergy Clin Immunol. 2001;107(6):1077-81.
  21. Lack G, Fox D, Northstone K, Golding J. Factors associated with the development of peanut allergy in childhood. N Engl J Med. 2003;348(11):977-85.
  22. Phan TG, Strasser SI, Koorey D, McCaughan GW, Rimmer J, Dunckley H, et al. Passive Transfer of Nut Allergy After Liver Transplantation. Archives of Internal Medicine. 2003;163(2):237-9.
  23. Brauns B, Schön MP, Wulf G, Mempel M. A lurking threat: transfer of peanut allergy through peripheral blood stem cell transplantation. World Allergy Organization Journal. 2016;9(1):3.
  24. Bellou A, Kanny G, Fremont S, Moneret-Vautrin DA. Transfer of atopy following bone marrow transplantation. Ann Allergy Asthma Immunol. 1997;78(5):513-6.
  25. McKenna C, Klontz KC. Systemic allergic reaction following ingestion of undeclared peanut flour in a peanut-sensitive woman. Ann Allergy Asthma Immunol. 1997;79(3):234-6.
  26. Hourihane JO. Peanut allergy--current status and future challenges. Clin Exp Allergy. 1997;27(11):1240-6.
  27. Simonte SJ, Ma S, Mofidi S, Sicherer SH. Relevance of casual contact with peanut butter in children with peanut allergy. J Allergy Clin Immunol. 2003;112(1):180-2.
  28. Tan BM, Sher MR, Good RA, Bahna SL. Severe food allergies by skin contact. Ann Allergy Asthma Immunol. 2001;86(5):583-6.
  29. Venter C, Sicherer SH, Greenhawt M. Management of Peanut Allergy. The Journal of Allergy and Clinical Immunology: In Practice. 2019;7(2):345-55.e2.
  30. Kattan J. The Prevalence and Natural History of Food Allergy. Curr Allergy Asthma Rep. 2016;16(7):47.
  31. Maleki SJ, Chung SY, Champagne ET, Raufman JP. The effects of roasting on the allergenic properties of peanut proteins. J Allergy Clin Immunol. 2000;106(4):763-8.
  32. Frémont S, Moneret-Vautrin DA, Zitouni N, Kanny G, Nicolas JP. Histamine content of peanuts. Allergy. 1999;54(5):528-9.
  33. Brough HA, Makinson K, Penagos M, Maleki SJ, Cheng H, Douiri A, et al. Distribution of peanut protein in the home environment. Journal of Allergy and Clinical Immunology. 2013;132(3):623-9.
  34. Jin JJ, Dorn JM, Yunginger J, Ott NL. Ara h 2 is detectable on surfaces of commercial airplanes. J Allergy Clin Immunol Pract. 2019;7(2):659-61.e2.
  35. Vadas P, Wai Y, Burks W, Perelman B. Detection of peanut allergens in breast milk of lactating women. Jama. 2001;285(13):1746-8.
  36. Foong RX, Brough H. The role of environmental exposure to peanut in the development of clinical allergy to peanut. Clinical & Experimental Allergy. 2017;47(10):1232-8.
  37. Anagnostou K, Clark A. The management of peanut allergy. Arch Dis Child. 2015;100(1):68-72.
  38. Al-Muhsen S, Clarke AE, Kagan RS. Peanut allergy: an overview. Cmaj. 2003;168(10):1279-85.
  39. Frith K, Katelaris CH. Current perspectives on peanut allergy. Internal Medicine Journal. 2019;49(12):1480-7.
  40. Simpson AB, Yousef E, Hossain J. Association Between Peanut Allergy and Asthma Morbidity. The Journal of Pediatrics. 2010;156(5):777-81.e1.
  41. Foong R-X, du Toit G, Fox AT. Asthma, Food Allergy, and How They Relate to Each Other. Frontiers in Pediatrics. 2017;5(89).
  42. Dhar S, Srinivas SM. Food Allergy in Atopic Dermatitis. Indian J Dermatol. 2016;61(6):645-8.
  43. Martin PE, Eckert JK, Koplin JJ, Lowe AJ, Gurrin LC, Dharmage SC, et al. Which infants with eczema are at risk of food allergy? Results from a population-based cohort. Clin Exp Allergy. 2015;45(1):255-64.
  44. Giannetti MP. Exercise-Induced Anaphylaxis: Literature Review and Recent Updates. Current Allergy and Asthma Reports. 2018;18(12):72.
  45. Zernotti ME, Pawankar R, Ansotegui I, Badellino H, Croce JS, Hossny E, et al. Otitis media with effusion and atopy: is there a causal relationship? World Allergy Organ J. 2017;10(1):37.
  46. Ho MH, Wong WH, Chang C. Clinical spectrum of food allergies: a comprehensive review. Clin Rev Allergy Immunol. 2014;46(3):225-40.
  47. Sicherer SH, Sampson HA. Food allergy: A review and update on epidemiology, pathogenesis, diagnosis, prevention, and management. J Allergy Clin Immunol. 2018;141(1):41-58.
  48. Sahin-Yilmaz A, Nocon CC, Corey JP. Immunoglobulin E-mediated food allergies among adults with allergic rhinitis. Otolaryngol Head Neck Surg. 2010;143(3):379-85.
  49. Vickery BP, Vereda A, Casale TB, Beyer K, du Toit G, Hourihane JO, et al. AR101 Oral Immunotherapy for Peanut Allergy. N Engl J Med. 2018;379(21):1991-2001.
  50. Dunlop JH. Oral immunotherapy for treatment of peanut allergy. Journal of Investigative Medicine. 2020;68(6):1152.
  51. Du Toit G, Roberts G, Sayre PH, Bahnson HT, Radulovic S, Santos AF, et al. Randomized trial of peanut consumption in infants at risk for peanut allergy. N Engl J Med. 2015;372(9):803-13.
  52. Du Toit G, Roberts G, Sayre PH, Plaut M, Bahnson HT, Mitchell H, et al. Identifying infants at high risk of peanut allergy: the Learning Early About Peanut Allergy (LEAP) screening study. J Allergy Clin Immunol. 2013;131(1):135-43.e1-12.
  53. Borres MP, Abrahamsson G, Andersson B, Andersson B, Bråkenhielm G, Fabricius T, et al. Asthma and allergies at school--a Swedish national position paper. Allergy. 2002;57(5):454-7.
  54. Pham MN, Wang J. Management of food allergies and asthma in schools. Ann Allergy Asthma Immunol. 2018;121(4):391-9.
  55. IUIS. WHO/IUIS Allergen Database 2020 [cited 2020 November]. Available from: http://www.allergen.org/.
  56. Hemmings O, Du Toit G, Radulovic S, Lack G, Santos AF. Ara h 2 is the dominant peanut allergen despite similarities with Ara h 6. J Allergy Clin Immunol. 2020;146(3):621-30.e5.
  57. Mattsson L, Valcour A, Holmqvist M, Larsson H, Lidholm J. Cyclophilin - A novel cross-reactive determinant in peanut. Clin Exp Allergy. 2021;51(4):620-2.
  58. Mueller GA, Maleki SJ, Pedersen LC. The molecular basis of peanut allergy. Curr Allergy Asthma Rep. 2014;14(5):429.
  59. Chassaigne H, Nørgaard JV, Hengel AJ. Proteomics-based approach to detect and identify major allergens in processed peanuts by capillary LC-Q-TOF (MS/MS). J Agric Food Chem. 2007;55(11):4461-73.
  60. Kleber-Janke T, Crameri R, Appenzeller U, Schlaak M, Becker WM. Selective Cloning of Peanut Allergens, Including Profilin and 2S Albumins, by Phage Display Technology. International Archives of Allergy and Immunology. 1999;119(4):265-74.
  61. Peeters KA, Koppelman SJ, van Hoffen E, van der Tas CW, den Hartog Jager CF, Penninks AH, et al. Does skin prick test reactivity to purified allergens correlate with clinical severity of peanut allergy? Clin Exp Allergy. 2007;37(1):108-15.
  62. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  63. Klemans RJ, Liu X, Knulst AC, Knol MJ, Gmelig-Meyling F, Borst E, et al. IgE binding to peanut components by four different techniques: Ara h 2 is the most relevant in peanut allergic children and adults. Clin Exp Allergy. 2013;43(8):967-74.
  64. Astier C, Morisset M, Roitel O, Codreanu F, Jacquenet S, Franck P, et al. Predictive value of skin prick tests using recombinant allergens for diagnosis of peanut allergy. J Allergy Clin Immunol. 2006;118(1):250-6.
  65. Kaur N, Mehr S, Katelaris C, Wainstein B, Altavilla B, Saad R, et al. Added Diagnostic Value of Peanut Component Testing: A Cross-Sectional Study in Australian Children. J Allergy Clin Immunol Pract. 2021 ;9(1):245–253.
  66. Nilsson C, Berthold M, Mascialino B, Orme ME, Sjölander S, Hamilton RG. Accuracy of component-resolved diagnostics in peanut allergy: Systematic literature review and meta-analysis. Pediatr Allergy Immunol. 2020;31(3):303-14.
  67. Barre A, Borges JP, Culerrier R, Rougé P. Homology modelling of the major peanut allergen Ara h 2 and surface mapping of IgE-binding epitopes. Immunol Lett. 2005;100(2):153-8.
  68. Nesbit JB, Schein CH, Braun BA, Gipson SAY, Cheng H, Hurlburt BK, et al. Epitopes with similar physicochemical properties contribute to cross reactivity between peanut and tree nuts. Mol Immunol. 2020;122:223-31.
  69. Bublin M, Breiteneder H. Cross-Reactivity of Peanut Allergens. Current Allergy and Asthma Reports. 2014;14(4):426.
  70. Mittag D, Akkerdaas J, Ballmer-Weber BK, Vogel L, Wensing M, Becker WM, et al. Ara h 8, a Bet v 1-homologous allergen from peanut, is a major allergen in patients with combined birch pollen and peanut allergy. J Allergy Clin Immunol. 2004;114(6):1410-7.
  71. Mittag D, Batori V, Neudecker P, Wiche R, Friis EP, Ballmer-Weber BK, et al. A novel approach for investigation of specific and cross-reactive IgE epitopes on Bet v 1 and homologous food allergens in individual patients. Mol Immunol. 2006;43(3):268-78.
  72. Guarneri F, Guarneri C, Benvenga S. Identification of potentially cross-reactive peanut-lupine proteins by computer-assisted search for amino acid sequence homology. Int Arch Allergy Immunol. 2005;138(4):273-7.
  73. Sanz ML, de Las Marinas MD, Fernández J, Gamboa PM. Lupin allergy: a hidden killer in the home. Clin Exp Allergy. 2010;40(10):1461-6.