Whole Allergen
f9 Rice
f9 Rice Scientific Information
Type:
Whole Allergen
Display Name:
Rice
Allergen code:
f9
Family:
Graminae (Poaceae)
Species:
Oryza sativa
Route of Exposure:
Ingestion
Source Material:
Unpolished rice
Latin Name:
Oryza sativa
Other Names:
Popped rice, Jasmine rice, Wild rice, Basmati rice, rice semolina
WHO/ICD-11 code:
XM3XQ1
(ICD-11 is currently under implementation and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Rice (Oryza sativa) is an essential food crop cultivated and routinely consumed globally and is regarded as a staple food for ~50% of the total world’s population. It is extensively cultivated in Asian countries, such as India, China, Japan, Indonesia, Myanmar, Cambodia, Nepal, the Philippines, Bangladesh, Pakistan, Thailand, the Republic of Korea, Srilanka, and Vietnam. Hence allergy to rice is predominantly reported from these countries. Oral ingestion of cooked/boiled rice, inhalation of uncooked, steamed rice, raw rice husk/dust, or direct skin contact with rice (sorting/handling rice or washing rice before cooking) have been considered as important routes of exposure causing rice allergy in individuals. Ingestion of rice may induce IgE-mediated food allergy symptoms, such as oral allergy syndrome or even anaphylaxis. Inhalation of rice husk/ raw rice powder could lead to respiratory symptoms (such as asthma, allergic rhinitis, and rhino-conjunctivitis), while direct contact with rice may cause skin symptoms (such as generalized urticaria/angioedema, and atopic dermatitis). Ory s 14 (a 14-kDa lipid transfer protein) is an important food allergen present in rice seed, that is found to be heat stable and resistant to pepsin digestion. Besides, Ory s 1 and Ory s 12 are rice pollen allergens responsible for causing respiratory allergic symptoms. Rice has been found to cross react with other cereals, such as barley, oats, wheat, rye, soybean, and corn, with fruit such as peach, and also with grass pollens.
Allergen
Nature
Oryza sativa, also referred as rice, is an essential food crop cultivated globally (1, 2). It is routinely consumed worldwide and is considered as a staple food for ~50% of the total world’s population (1, 3).
Rice is an annual grass plant, that may survive for several years in tropical areas. The characteristic feature of this plant species includes its erectness, with a height of about 1to 2 m, having extended blades, with dense and droopy panicles of approximately 15 to 40 cm in length. The spikelets measure about 7 to 10 mm in length and 3 to 4 mm in width, with a single flower grown. The flowering season is usually between the period from July to October (2).
Rice is considered as an essential member of grass family. During flowering season, the rice plants produce very large quantities of pollens in agricultural fields. Furthermore, it has been reported that the levels of rice pollens in the air are very high between the time period from 08:00 to 12:00 h and 14:00 to 16:00 h (4).
Habitat
Rice is considered as a type of grass that is semi-aquatic in nature. It can grow in a wide variety of soil, ranging from saline, alkaline, or acid–sulphate (1). Salinity, cold stress, irrigation water availability, and low temperature are certain important environmental limiting factors for the growth of these plants (5). The rice plants can grow either on highlands or wetlands (shallow water, swamps, and marshes) (2, 5). Rice is generally considered sensitive to drought condition, since it prefers either fully irrigated or rain-fed area. Furthermore, even though rice is found to be tolerant to floods; only a few varieties are able to tolerate prolonged submergence and majority of them die within 14 days of complete immersion (5).
Taxonomy
Rice belongs to the genus Oryza. This genus consists of approximately 22 species, among which 20 are considered wild. O. sativa and O. glaberrima are the only two varieties of cultivated rice that are widely consumed across the world (1).
Taxonomic tree of rice (O. sativa)  (6)
Domain Eukaryota
Kingdom Viridiplantae
Phylum Streptophyta
Class Magnoliopsida
Order Poales
Family Poaceae
Genus Oryza
Species
O. sativa
Tissue
The immunoglobulin (Ig)E-mediated allergic reactions might affect individuals, by inducing skin, respiratory or gastrointestinal (GI)/systemic symptoms, either through inhalation of raw rice dust or vapored rice while steaming, or by consumption of rice. Furthermore, reports of urticaria and angioedema caused by direct handling/sorting of rice has also been found (7-10).
Ory s 14, a 14-kDa lipid transfer protein (LTP) is present in the rice seeds (8). It has been found that, this LTP may be capable of inducing severe allergic reactions, such as anaphylaxis (11). It may be regarded as a true food allergen as it is seen to be extremely resistant to heat and pepsin digestion (12). Ory s 1 (35-kDa β-expansin) found in the rice pollen is a major allergen (8, 13). The protein microarray-IgE binding study revealed that β-expansin could be considered as one of the important allergens (13).
Epidemiology
Worldwide distribution
Despite high consumption of rice globally, prevalence of rice allergy is considered low (8, 14). It is a staple food, mostly in the Eastern Asia; hence allergy to rice is predominantly reported from Asian countries (8, 12).
In Western countries, rice allergy is rarely reported with varied prevalence, ranging between 0.7 and 2% in these population, although the true prevalence rate is unclear (14, 15). This is probably because in these regions rice is considered mainly as a cereal-related allergen source. Furthermore, rice allergy has also been found in countries, such as Russia, Denmark, France, Estonia, Lithuania, and Sweden (10).
In Thailand, the rice ranked 4th among the list of important food allergens, post shellfish, peanut, and soy, while in Japan it ranked 5th and in Indonesia 6th (10).
In Japan, the estimated prevalence of IgE-mediated rice allergy was found to be ~10% in atopic individuals (8).
A survey was conducted in India to determine the rice allergy in 1200 AR and bronchial-asthmatic patients aged between 5 and 60 years. Among these patients, 165 were found to have a prior history of rice allergy. It was reported that 12.1% of patients (20 out of 165) were found positive to skin prick test (SPT) result (n=20), 65% of patients (13 out of 20) showed increased-specific (s)-IgE for raw rice, whereas 60% of patients showed it against boiled rice. Furthermore, according to double blind placebo-controlled food challenge (DBPCFC), 60% of patients demonstrated allergy to rice (6 out of 10). Overall, rice was considered to be an essential allergen source, with IgE-mediated rice allergy affecting 0.8% of AR and bronchial-asthmatic patients (10). Furthermore, in a cross-sectional study conducted in India, allergy to rice was reported to be greater among patients of age group>40 years in comparison to patients<40 years (16).
In a study conducted in Malaysia, 148 adult patients (>17 years) having symptoms of AR (rhinorrhoea and nasal congestion) were compared to 113 subjects without AR, who acted as controls. The study revealed that, among 11 food items tested, 48% of AR patients (71 out of 148) showed positive SPT reactivity for food, in comparison to 4.4% (5 out of 113) among controls. Moreover, compared to other food allergens, rice (30.4%; 45 out of 148) and shrimp (47.9%; 71 out of 148) demonstrated greater reactivity to SPT among AR patients (17).
Risk factors
Risk of asthma and contact dermatitis might occur as a result of occupational exposure to rice, among food workers and bakers (12). Moreover, the LTP allergen of rice (Ory s 14) shares high-sequence similarity with major maize allergen, a 9 kDa LTP, attributing to high risk of co-sensitization in individuals having these allergies (18).
Environmental Characteristics
Worldwide distribution
Oryza sativa (cultivated rice) is widely distributed across Asia, the Middle East, North and South America, Africa and the European Union (1). Among the different varieties of cultivated rice species, the long-grain Indica rice is majorly found in tropical and the subtropical Asia, the short-to-medium-grained Japonica rice is grown in temperate areas of Japan and northern part of China, whereas the medium-grained Javonica rice are grown in the Philippines and the hilly terrains of Madagascar and Indonesia (1). In the US, O. sativa has been introduced in the regions, such as California, Florida, Louisiana, Mississippi, Texas, and Arkansas (2).
Rice is extensively cultivated in countries, such as India, China, Japan, Indonesia, Myanmar, Cambodia, Nepal, the Philippines, Bangladesh, Pakistan, Thailand, the Republic of Korea, Srilanka, and Vietnam, constituting of ~90% of the production of rice from these countries. India and China are the major contributors (~50%) of the total production of rice. Besides this, non-Asian countries such as the US, Madagascar, Brazil, and Nigeria produce 5% of rice across the globe. Furthermore, although rice has become the fastest growing staple diet in Africa, only about 54% of consumed rice is provided locally (1).
Route of Exposure
Main
Oral ingestion of cooked/boiled rice or inhalation of uncooked, steamed rice, raw rice husk/dust have been considered as important routes of exposure causing rice allergy in individuals (2, 7, 8, 10). It has been reported that oral ingestion of rice could lead to IgE-mediated and rapid hypersensitivity reactions, such as oral allergy syndrome (OAS), anaphylaxis, asthma, allergic rhinitis (AR), atopic dermatitis, and rhino-conjunctivitis (7, 8, 10, 19). Similarly, inhalation of rice dust, husk or flour as well as rice vapors during cooking could induce asthma, AR, rhino-conjunctivitis, anaphylaxis, and in some cases generalized urticaria/angioedema (7-10, 19).
Direct skin contact with rice (sorting/handling rice or washing rice before cooking) is another important route of exposure that may cause allergic reactions, such as contact urticaria and angioedema (7, 8, 10).
Clinical Relevance
According to published literatures, it has been reported that rice allergy may cause severe allergic reactions in individuals, leading to respiratory (asthma, AR, rhino-conjunctivitis), GI (vomiting, abdominal pain, diarrhea), skin (atopic dermatitis, angioedema, urticaria), and systemic symptoms (anaphylaxis) (2, 7, 11).
Oral allergy symptoms and Anaphylaxis
Ingestion of rice or inhalation of rice dust/flour may induce IgE-mediated food allergy symptoms, such as OAS or even anaphylaxis (10). This has been documented in several case reports, mentioned below.
In a case series involving three LTP-allergic patients (25-, 30-, and 50-year-old patients), anaphylaxis induced by rice was reported. All the three cases were presented with dyspnea, generalized urticaria or angioedema on consumption of risotto (cooked rice and mushrooms) and/or rice along with chicory. The results showed positive SPT for commercial rice extract with high rice sIgE (11).
A case of repeated anaphylactic episode, following oral ingestion of rice with symptoms, such as angioedema, pruritis, generalized urticaria, bronchospasm, and dysphagia was reported. The results revealed that IgE reactivity was confirmed (detected by SPT) for various rice extracts (both raw rice and cooked boiled rice) (8).
Gastrointestinal reactions, such as pain in the abdomen, nausea, vomiting and diarrhea on consumption of rice was reported in a 9-year-old girl. The SPT and prick-prick tests showed a noticeable wheal and flare response with rice. Furthermore, a double-blind, placebo-controlled, food challenge test performed on the patient reported GI symptoms within 10 mins of eating rice; however, it was observed that she didn’t show any allergic reaction while consuming other cereals (9).
Rice flour-induced anaphylaxis was reported in a 6-month-old girl who was admitted in the hospital thrice as a result of immediate onset of respiratory as well as GI symptoms, pale skin, and decreased responsiveness (20). Anaphylaxis has also been reported due to inhalation of rice proteins in a 11-year-old-boy with atopic dermatitis (15).
Allergic rhinitis, rhino-conjunctivitis, and asthma
Inhalation of rice dust/pollen may induce AR, and this could be considered as one of the factors for the development of asthma in individuals (19).
It has been reported that allergy to rice pollens may induce these respiratory symptoms among rice farmers and people living near rice farms during the flowering season of rice plants (4). A cross-sectional study conducted among 464 rice farmers (mean age 48.3± 15.2 years) in California (USA) reported AR (25.7%), persistent wheeze (8.8%), asthma and severe cough (7.1% each), and bronchitis (6.3%), due to burning of rice stubble in the agricultural fields (21).
In a study conducted in Taiwan, 9.3% of asthmatic children showed positive response to rice pollen extract (n=312), and it was reported that sensitization to rice pollen might be one of the important factors for eliciting IgE-specific responses in the patients (2, 22).
In a case series of four patients, it was reported that inhalation of raw rice (while throwing rice in wedding ceremony) may have led to respiratory-allergic symptoms (rhino-conjunctivitis and asthma) (23, 24) (23).
A case of AR was identified in a 31-year-old male who worked in a grain industry and showed a prior history of runny nose, blockage, sneezing and itchy/burning eyes at work, while being exposed to rice powder. His symptoms aggravated at workplace. The SPT was found positive for rice extracts, the serum-IgE level for rice was reported high, and the nasal provocation test with rice extract showed development of AR. The findings indicated that inhalation of rice flour at workplace may have led to AR (19).
Atopic Dermatitis
Allergic reaction induced by rice could be IgE mediated in cases of AD and bronchial asthma (10). Few studies have been reported, which suggest that exacerbation of AD may be caused by rice (7).
In a study conducted in Japan among 1006 patients with AD, it was found that rice may be associated with the severity of AD. Furthermore, excluding rice in the diet of 25 severe AD patients showed good improvement of AD in 9 patients, moderate improvement in 10 patients, and neither aggravation nor any effect in 6 patients (25)
A case of a 3-year-old boy (with AD since the age of 3 months) was reported, who showed worsening of AD symptoms (skin eruptions and rashes) after coming in contact with rice dust (3).
Urticaria and Angioedema
Direct touching/sorting of raw rice or the water used for washing rice may lead to contact urticaria in certain individuals (10).
Two case reports have been identified where handling of raw rice led to contact urticaria in hand. The first case was of a 27-year-old housewife who showed positive SPT with flour extract, positive rub test and challenge test with raw rice, and high rice-specific IgE in serum. (24). In the second case of a 17-year-old female, SPT with rice revealed urticaria suggesting raw rice could be the cause of cutaneous reaction in the patient (26). Furthermore, development of contact urticaria (erythema and itching) after washing rice in water was reported in a 30-year-old male who showed positive SPT with rice allergen, and avoidance of contact with water used for washing rice improved the skin conditions (27).
Furthermore, occurrence of urticaria and angioedema after inhalation of raw rice has rarely been reported. This was evident in a 9-year old girl who developed urticaria-angioedema after inhaling raw rice (9).
Urticaria and angioedema of the eyelids, tongue, and lips as a result of consumption of rice has also been reported in a case study of a 33-year-old German patient (8).
Other diseases
Rice millers’ syndrome
It has been reported that the rice millers’ syndrome might occur as a result of exposure to rice husk powder. The clinical manifestations may include irritation of eyes, skin, and respiratory tract; allergic reactions like nasal discharge, chest tightness, asthma, and high eosinophil counts, along with pulmonary infiltrates, which might represent early silicosis or hypersensitivity pneumonitis. This was evident in a Malaysian study conducted on 122 rice-mill workers (28).
Prevention and Therapy
Prevention strategies
Avoidance
Individuals who are allergic to rice should generally avoid consumption of this grain completely, along with foods that act as alternatives for rice. Furthermore, certain other foods (barley, oats, wheat, rye, soybean, corn, grass pollen and triticale) that belong to the same family as rice, should also be avoided (12).
Immediate medical care should be advised to an individual who is allergic to rice, in case if it is accidentally consumed (12). 
Molecular Aspects
Allergenic molecules
The raw rice, even though may be considered as more allergenic than the cooked rice, yet there are certain allergens found to be resistant to both heat and proteolysis (12). Globally, cultivated rice is found to be one of the important sources of airborne allergen among grass pollens (4). The World Health Organization (WHO) and International Union of Immunological Societies’ (IUIS) Allergen Nomenclature Subcommittee have identified and officially published two allergenic components, i.e., Ory s 1 and Ory s 12 from rice pollens, both of which are exposed through inhalation (8, 29). The table below provides the list of allergens.  
Allergens Molecular Weight (kDa) Biochemical name Allergenicity
Ory s 1  35 β–expansin 
  • Major allergen (8, 13).
  • Complete inhibition of IgE binding to Ory s 1 protein was reported when recombinant Ory s 1 was preincubated with the sera of patients exhibiting high sensitivity to rye grass pollen (29, 30).
  • Ory s 1 (rice pollen allergen) showed 66% sequence homology with Lol p 1 (rye grass pollen allergen) (8).
Ory s 12 14 Profilin A 
  • In a Japan study, sera of 10 patients (IgE-reactive with proteins of rice pollen) were detected positive to both β–expansin as well as polycalcin extensin, whereas low to polygalacturonase, with mostly negative towards profilin (13, 29).
Ig; Immunoglobulin
Apart from the ones included above, there are other putative rice pollen and rice-seed allergens that have been identified (8, 31).
Ory s 14 (14-kDa LTP) is an important food allergen present in the rice seeds (8, 11). Reports of anaphylaxis caused due to rice was identified in three patients who were found allergic to Ory s 14 LTP allergen (11). Furthermore, another LTP allergen i.e. 9-kDa LTP was a major allergen of rice and was found to have sequence similarity with Pru p 3 (7). Other important rice-seed food allergens include, Ory s aA/trypsin inhibitor (TI) (14–16 kDa α-amylase/ TI), Ory s glyoxalase I (33 kDa glyoxalase I activity protein) and α-globulin (25–28 kDa protein) (8).
Besides this, several rice-pollen allergens have been identified, which include Ory s 2 (12-13 kDa; calcium-binding protein), Ory s 7 (polcalcin), Ory s 11 (TI), Ory s 13 (polygalacturonase protein) and Ory s 23 (8),
Furthermore, allergens such as RAG2 and globulin (19 kDa) were identified in polished rice (present in the inside surface of rice grain); whereas a 52-kDa globulin protein was identified in the rice bran (present in outside surface of rice grain) (31).
In a study conducted in India among 1200 asthmatic and AR patients, 165 patients were found to be rice allergic, based on past history. The study identified major IgE-binding protein components (major allergens), such as 14-16, 33, 56, and 60 kDa from raw rice; whereas 16, 23, 33 and 53 kDa proteins from boiled rice (10).
Furthermore, a chitinase has also been identified at high levels in roots of the rice plant; however, its presence in the rice seed along with its allergenic potential has yet not been clearly studied (12).
Biomarkers of severity
Ory s 14, an LTP and an important rice allergen is extremely heat stable, pepsin resistant and may be responsible for inducing allergy, after consumption of cooked rice (12). Furthermore, due to its ability to resist heat and proteolysis, it may be considered an effective allergen (11).
Cross-reactivity
Individuals who are allergic to rice might also show cross-reactivity with other members of the same family, such as barley, oats, wheat, rye, soybean, corn, grass pollen and triticale (12, 18). The rice proteins with a molecular mass of 15.5, 16, 19, 25, 50, and 90 kDa have been identified as major allergens, developing cross-reactivity among cereal grains in family Poaceae (12).
Rice LTP is considered as a cross-reacting allergen, with reports of it cross-reacting with LTPs of peach (8, 11) In a study conducted on 148 peach-allergic individuals, 56.8% of patients were found sensitized to rice (IgE test). It was reported that all the patients with symptoms induced by rice were found to be Pru p 3 (a LTP from peach) positive. Furthermore, the major rice allergen identified was 9 kDa (LTP), that showed 53% sequence similarity with Pru p 3. This indicates that rice could lead to allergic reactions in peach-allergic patients (7).
Furthermore, rice LTP has also shown sequence identity with maize LTP, thus resulting into cross-reactivity between the two (18). A study revealed that IgE antibodies from 28.5% of patients with maize allergy (n=7) showed cross-reactivity with a 56 kDa rice allergen (32). In another study, rice extracts completely inhibited all the maize allergens. Moreover, maize LTP was found to have 79% identity and 92% similarity with rice LTP (18).
In a study it was observed that patients with allergies to food (mostly wheat) and pollens, may often display high levels of IgE-antibodies, particularly to rice (33).
In another study, based on the radioallergosorbent (RAST) test value (r= 0.56; p<0.01)), 16-kDa rice protein was found to be correlated with Italian millet, however this was not the case with either wheat or corn (34).
Cross-reactivity between rice and other cereal flour extracts have been noted. This evidence is supported by a case study of a 27-year-old female, who developed rhino-conjunctivitis, asthma as well as contact urticaria, while handling rice and certain other cereals (wheat, rye and corn). Based on the reverse enzyme-immunoassay (REIA) performed, cross-reactivity was reported between rice (75%) and other cereal flour extracts, such as rye (63%), corn (64%), and wheat (51%) (24). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: February 2021
References
  1. Muthayya S, Sugimoto JD, Montgomery S, Maberly GF. An overview of global rice production, supply, trade, and consumption. Ann N Y Acad Sci. 2014;1324:7-14.
  2. Weber RW. Allergen of the month--rice. Ann Allergy Asthma Immunol. 2012;108(2):A11.
  3. Nambu M, Shintaku N, Ohta S. Rice allergy. Pediatrics. 2006;117(6):2331-2.
  4. Sen MM, Adhikari A, Gupta-Bhattacharya S, Chanda S. Airborne rice pollen and pollen allergen in an agricultural field: aerobiological and immunochemical evidence. J Environ Monit. 2003;5(6):959-62.
  5. Menguer PK, Sperotto RA, Ricachenevsky FK. A walk on the wild side: Oryza species as source for rice abiotic stress tolerance. Genet Mol Biol. 2017;40(1 suppl 1):238-52.
  6. NCBI. Taxanomy Browser (Oryza sativa) 2020 [cited 26 September 2020]. Available from: https://www.ncbi.nlm.nih.gov/Taxonomy/Browser/wwwtax.cgi?id=4530.
  7. Pastorello EA, Scibilia J, Farioli L, Primavesi L, Giuffrida MG, Mascheri A, et al. Rice allergy demonstrated by double-blind placebo-controlled food challenge in peach-allergic patients is related to lipid transfer protein reactivity. Int Arch Allergy Immunol. 2013;161(3):265-73.
  8. Trcka J, Schad SG, Scheurer S, Conti A, Vieths S, Gross G, et al. Rice-induced anaphylaxis: IgE-mediated allergy against a 56-kDa glycoprotein. Int Arch Allergy Immunol. 2012;158(1):9-17.
  9. Orhan F, Sekerel BE. A case of isolated rice allergy. Allergy. 2003;58(5):456-7.
  10. Kumar R, Srivastava P, Kumari D, Fakhr H, Sridhara S, Arora N, et al. Rice (Oryza sativa) allergy in rhinitis and asthma patients: a clinico-immunological study. Immunobiology. 2007;212(2):141-7.
  11. Asero R, Amato S, Alfieri B, Folloni S, Mistrello G. Rice: another potential cause of food allergy in patients sensitized to lipid transfer protein. Int Arch Allergy Immunol. 2007;143(1):69-74.
  12. Suvarna B. Rice, the Allergen. Nepal Journal of Science and Technology. 2008;9.
  13. Hirano K, Hino S, Oshima K, Okajima T, Nadano D, Urisu A, et al. Allergenic potential of rice-pollen proteins: expression, immuno-cross reactivity and IgE-binding. J Biochem. 2013;154(2):195-205.
  14. Gilissen LJ, van der Meer IM, Smulders MJ. Reducing the incidence of allergy and intolerance to cereals. Journal of Cereal Science. 2014;59(3):337-53.
  15. Fiocchi A, Bouygue GR, Restani P, Gaiaschi A, Terracciano L, Martelli A. Anaphylaxis to rice by inhalation. J Allergy Clin Immunol. 2003;111(1):193-5.
  16. Dey D, Ghosh N, Pandey N, Gupta Bhattacharya S. A hospital-based survey on food allergy in the population of Kolkata, India. Int Arch Allergy Immunol. 2014;164(3):218-21.
  17. Gendeh BS, Murad S, Razi AM, Abdullah N, Mohamed AS, Kadir KA. Skin prick test reactivity to foods in adult Malaysians with rhinitis. Otolaryngol Head Neck Surg. 2000;122(5):758-62.
  18. Pastorello EA, Farioli L, Pravettoni V, Ispano M, Scibola E, Trambaioli C, et al. The maize major allergen, which is responsible for food-induced allergic reactions, is a lipid transfer protein. J Allergy Clin Immunol. 2000;106(4):744-51.
  19. Kim JH, Kim JE, Choi GS, Hwang EK, An S, Ye YM, et al. A case of occupational rhinitis caused by rice powder in the grain industry. Allergy Asthma Immunol Res. 2010;2(2):141-3.
  20. Klein SK, Kremers EM, Vreede WB. [Six-month old girl with an anaphylactic reaction to rice flour, a rare food allergy]. Ned Tijdschr Geneeskd. 2001;145(30):1471-3.
  21. McCurdy SA, Ferguson TJ, Goldsmith DF, Parker JE, Schenker MB. Respiratory health of California rice farmers. Am J Respir Crit Care Med. 1996;153(5):1553-9.
  22. Tsai YT, Chen SH, Lin KL, Hsieh KH. Rice pollen allergy in Taiwan. Ann Allergy. 1990;65(6):459-62.
  23. Enrique E, Ahrazem O, Bartra J, Latorre MD, Castello JV, de Mateo JA, et al. Lipid transfer protein is involved in rhinoconjunctivitis and asthma produced by rice inhalation. J Allergy Clin Immunol. 2005;116(4):926-8.
  24. Lezaun A, Igea JM, Quirce S, Cuevas M, Parra F, Alonso MD, et al. Asthma and contact urticaria caused by rice in a housewife. Allergy. 1994;49(2):92-5.
  25. Ikezawa Z, Miyakawa K, Komatsu H, Suga C, Miyakawa J, Sugiyama A, et al. A probable involvement of rice allergy in severe type of atopic dermatitis in Japan. Acta Derm Venereol Suppl (Stockh). 1992;176:103-7.
  26. di Lernia V, Albertini G, Bisighini G. Immunologic contact urticaria syndrome from raw rice. Contact Dermatitis. 1992;27(3):196.
  27. Yamakawa Y, Ohsuna H, Aihara M, Tsubaki K, Ikezawa Z. Contact urticaria from rice. Contact dermatitis. 2001;44:91-3.
  28. Lim HH, Domala Z, Joginder S, Lee SH, Lim CS, Abu Bakar CM. Rice millers' syndrome: a preliminary report. British journal of industrial medicine. 1984;41(4):445-9.
  29. WHO/IUIS. Oryza sativa (Rice) 2020 [cited 2020 29 Sep 2020]. Available from: http://www.allergen.org/search.php?allergensource=Oryza+sativa.
  30. Xu H, Theerakulpisut P, Goulding N, Suphioglu C, Singh MB, Bhalla PL. Cloning, expression and immunological characterization of Ory s 1, the major allergen of rice pollen. Gene. 1995;164(2):255-9.
  31. Satoh R, Tsuge I, Tokuda R, Teshima R. Analysis of the distribution of rice allergens in brown rice grains and of the allergenicity of products containing rice bran. Food Chem. 2019;276:761-7.
  32. Krishnan HB, Chen MH. Identification of an abundant 56 kDa protein implicated in food allergy as granule-bound starch synthase. J Agric Food Chem. 2013;61(22):5404-9.
  33. Golias J, Humlova Z, Halada P, Habova V, Janatkova I, Tuckova L. Identification of rice proteins recognized by the IgE antibodies of patients with food allergies. J Agric Food Chem. 2013;61(37):8851-60.
  34. Urisu A, Yamada K, Masuda S, Komada H, Wada E, Kondo Y, et al. 16-kilodalton rice protein is one of the major allergens in rice grain extract and responsible for cross-allergenicity between cereal grains in the Poaceae family. Int Arch Allergy Appl Immunol. 1991;96(3):244-52.