CMA is the leading cause of food allergy in children under 3 years of age but may be diagnosed in patients of all ages. As symptoms of CMA overlap with those of other gastrointestinal disorders, appropriate diagnostic workup is vital to avoid over- and under-diagnosis, and thus minimise the health burden to patients and their families.1
Read through this evidence-based guide to learn how to accurately diagnose CMA in clinical practice.
The symptoms of CMA can be diverse and of variable intensity, involving many different organ systems, but most commonly the skin and the gastrointestinal and respiratory tracts. The involvement of ≥2 systems increases the likelihood of CMA.1
Immediate and delayed reactions should be differentiated; however, combinations of these two types of reaction may be present in one patient.1
If any of the following signs or symptoms occur in an infant or child, and cannot be explained by another cause, CMA may be considered a potential diagnosis:1
*Anaphylaxis or shock-like symptoms are clear indications for a referral to an allergy specialist.
If the patient’s history indicates IgE-mediated allergy, specific IgE blood tests can assist in the diagnosis.2,3 Specific IgE blood tests can be performed at any age and use validated assays to quantify allergen-specific IgE levels in the serum.2 Firstly, a test for a specific IgE response to the whole allergen (cow’s milk) should be performed.3
Interpretation of whole-allergen specific IgE blood test results1
Whole-allergen tests predict the likelihood of CMA but are not predictive of reactivity to baked milk. After testing for a specific IgE response to whole allergen, cow’s milk allergen component tests can help evaluate reactivity to baked goods and provide further information on the likelihood of allergy persistence.3
Assessment of specific IgE responses to the following components (proteins) could help pinpoint the diagnosis and guide the management of patients with CMA:3
As caseins are more resistant than other proteins to heat denaturation, patients with high levels of casein-specific IgE are likely to react to baked milk. Such patients should therefore avoid consumption of all forms of milk. Patients who test negative for casein-specific IgE may be able to tolerate extensively heated milk, for example in baked goods.3
If relevant symptoms are present and CMA is likely, a diagnostic elimination diet should be initiated. Cow’s milk should be strictly excluded from the patient’s diet (or the mother’s diet in case of breastfeeding) for a limited period.1
The duration of the elimination diet should be kept as short as possible whilst being long enough to judge whether clinical symptoms resolve or stabilise. The required duration may be:1
If no improvement in symptoms is seen, then CMA is unlikely. However, exceptions may occur, especially in cases of multiple sensitisations. Infants with significant gastrointestinal symptoms that do not improve when using a hydrolysed or soy formula may benefit from a trial of an amino acid-based formula before CMA is excluded.1
If symptoms significantly improve during the diagnostic elimination diet, the diagnosis of CMA should be confirmed by a standardised oral food challenge (OFC) under medical supervision.1
In cases of anaphylactic/clear immediate reaction to cow’s milk and a positive whole-allergen specific IgE blood test, cow's milk should be strictly excluded from the diet (see step 5) and the OFC can be omitted. Cow's milk should be excluded from the diet for at least 1 year before an oral food challenge is considered by a specialist.1
If an immediate reaction to cow’s milk is apparent but the whole-allergen specific IgE blood test is negative, an OFC should be carried out in a hospital setting under strict medical supervision.1
Strict avoidance of cow's milk is the safest strategy in the management of CMA. Cow's milk should be eliminated from the diets of patients with:1
Based on allergen component blood test results, patients may be able to tolerate extensively heated (baked) milk. Dietary inclusion of baked products containing cow’s milk may accelerate development of tolerance to unheated cow’s milk.3
Patients should be re-evaluated at around 6-12 months, to avoid continuing the restrictive cow's milk exclusion diet for an unnecessarily long time.1,3 The nutritional benefits of cow’s milk are well known, and restrictions on its intake, particularly in childhood, may result in stunted growth.1 Periodic re-assessments with laboratory tests and oral food challenges are recommended.3
IgE: immunoglobulin E
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.