Whole Allergen
i1 Honeybee
i1 Honeybee Scientific Information
Type:
Whole Allergen
Display Name:
Honeybee
Route of Exposure:
Injection (sting)
Family:
Apidae; Order: Hymenoptera
Species:
Apis mellifera Linnaeus
Latin Name:
Apis mellifera
Other Names:
Western honeybee
Summary
The honeybee, a cosmopolitan insect, is the only stinging member of the order Hymenoptera that leaves its barbed stinger and venom sac in the victim. Hymenoptera stings cause 48% of severe anaphylactic reactions occurring in European adults, and 20% of those occurring in children. In the United States, the prevalence of Hymenoptera-induced anaphylaxis is estimated at 3% in adults and 1% in children, with 40 to 100 Hymenoptera sting-induced fatalities being documented annually. Anaphylaxis is more common in adults than in children. Systemic reactions usually occur within minutes of being stung. The risk of repeated anaphylaxis is 30% to 70%. An estimated 9 to 42% of the general population is sensitized to Hymenoptera venom. Beekeepers, greenhouse workers, and rural populations are at higher risk of developing bee sting allergy. Mast cell disorders including hereditary α-tryptasemia, elevated baseline serum tryptase, or a family history of honeybee allergy are associated with an increased risk of occurrence and severity of Hymenoptera sting-induced reactions. A history of Hymenoptera-induced anaphylaxis is a red flag for an underlying clonal mast cell disorder.
Honeybee venom (HBV) contains 12 characterized allergens, of which five are available for in vitro diagnosis: Api m 1, Api m 2, Api m 3, Api m 5, and Api m 10. Sensitization to Api m 1 and to Api m 10 is most prevalent, at 57%–97% and 51.5%–61.8%, respectively. When testing for specific IgE to venom components, inclusion of all five major allergens allows identification of the patient’s sensitization profile, facilitates precise venom immunotherapy (VIT), and helps to improve specificity when HBV-sensitized patients have also tested positive for Vespid allergen extracts. 
Allergen
Nature
HBV consists of a complex mixture of allergenic and non-allergenic molecules contained in the venom sac at the distal extremity of the insect’s abdomen.
Taxonomy
Taxonomic tree of Apis mellifera
Domain Eukaryota
Kingdom Metazoa (Animalia)
Phylum Arthropoda
Subphylum Hexapoda
Class Insecta
Order Hymenoptera
Suborder Apocrita
Family Apidae
Genus Apis
Species Apis mellifera
Within the order Hymenoptera, which includes bees, vespids (yellow jacket/wasps, hornets, and paper wasps) and ants, bees are important pollinating insects belonging to a monophyletic group of 16,000 species. The Apidae family comprises bees (genus Apis) and bumblebees (genus Bombus) [1-4].
The honeybee is distinct from other stinging Hymenoptera in that it nearly always leaves its barbed stinger and venom sac in its victim’s skin [4].
Tissue
Bee venom has a complex composition, containing enzymes, other proteins, biogenic amines, and phospholipids. To date, 12 HBV proteins have been characterized as allergens and included in the IUIS/WHO Allergen Nomenclature [5]. Allergenic proteins can also act as toxins, with melittin (Api m4) as the best known example [1, 6].
Epidemiology
Worldwide distribution 
Hymenoptera stings cause 48% of severe anaphylactic reactions occurring in European adults, and 20% of those occurring in children [4].
It is estimated that the worldwide annual incidence of immunologic reactions to Hymenoptera stings ranges from 0.3–3.0% which equates to almost 100 million cases per year. Severity ranges from  local wheal-and-flare reactions to death from anaphylactic shock [7].
In the US, Hymenoptera-induced systemic reactions are estimated to occur in 3% of adults and 1% of children, and approximately 40 to 100 fatalities are reported each year, although the figure is likely to be higher [8]. An Australian study reported honeybee to be the main cause of fatal insect venom anaphylaxis, however, there is a relatively high prevalence of honeybee allergy in Australia, and UK and European studies found that wasp was the most common cause of fatal and nonfatal venom anaphylaxis, respectively [9]. A consistent finding across studies in Australia, Canada, UK and US was that fatal insect venom anaphylaxis occurs at an approximate rate of 0.1 cases per million population [9]. In Europe, the prevalence of systemic reactions to Hymenoptera stings is 0.3% to 7.5% [10, 11].
Risk factors
Sensitization to Hymenoptera venom, estimated at 9.2% to 42% of the adult population, comprises a majority of asymptomatic individuals [4]. However, exposed populations, e.g. rural dwellers or beekeepers and their families, are at higher risk of developing HBV allergy [12]. In beekeepers, a 14% - 43% prevalence of systemic reactions to stings was reported [12, 13].
The prevalence and severity of Hymenoptera venom reactions are increased in patients with mast cell disorders including hereditary α-tryptasemia, with or without an elevated baseline serum tryptase concentration  [14-17]. Hymenoptera venom allergy was observed in 50% of patients with systemic mastocytosis without hereditary α-tryptasemia and in 82% of those with concurrent hereditary α-tryptasemia [15].
Cardiovascular risk factors, male gender and older age have also been associated with an increased risk of severe reactions to Hymenoptera venom [18]. Atopy and a family history of allergy to bee stings have been inconsistently associated with severe reactions [6, 18].
Pediatric issues
The prevalence of systemic reactions to Hymenoptera venoms is estimated at 3.4% in children [19]. In children younger than 16 years experiencing a cutaneous reaction to Hymenoptera venom, the chance of anaphylaxis if re-stung is lower than 3% [8]. 
Environmental Characteristics
Living environment
Worldwide distribution (of the species)
Apis mellifera (European, western or common honeybee), found worldwide as a result of human movements, and Apis cerana (eastern honeybee), found mostly in Asia, are the two domesticated species of honeybee, while bumblebees (Bombus spp) are currently employed as pollinators in greenhouses [4, 20]. Extensive cross-reactivity is found between these Apidae family members [4].
Wild honeybees make use of hollow trees, rotten logs and sometimes voids in buildings, in which to build their nests [21].
Most species of honeybee behave non-aggressively towards humans and sting only when they feel threatened. The accidental introduction of the Africanized bee hybrid to Brazil during the 1950s, however, has been associated with an increase in reported attacks from bees across most of the Americas [6].
Route of Exposure
Main
Exposure to HBV occurs through a honeybee sting, when the barbed stinger becomes embedded in the flesh and is pulled out of the abdomen of the insect along with the venom sac. Worker bees may survive for 18 to 114 hours after a stinging event but, unlike wasps, hornets, and yellow jackets, honeybees can only sting once [1, 6].
A single sting is capable of delivering an average of 140 µg to 150 µg of venom, although venom sacs may contain more than 300 µg of venom [1, 6]  Within the first 20 seconds after stinging, 90% or more of the contents of the venom sac will already have been delivered [6].
Secondary
Ingestion of bee products such as honey, royal jelly, propolis, or “bee pollen” are usually considered as unrelated to Hymenoptera venom allergy, however, among the three isoforms of Api m 11, two are described as venom allergens and one as a food allergen [5, 22, 23].
Clinical Relevance
Five types of reactions to Hymenoptera stings are recognized: normal local reactions, large local reactions (LLR), systemic anaphylactic reactions, systemic toxic reactions, and unusual reactions. Of these, LLR and systemic anaphylactic reactions are the most common [1]. It can be difficult to differentiate systemic anaphylactic reactions from local reactions and toxic reactions based on signs and symptoms [24].
Anaphylaxis
Systemic reactions limited to cutaneous signs only carry a risk of anaphylaxis to a future sting below 3%. Conversely, a history of systemic sting-induced reaction and detectable venom IgE put the risk of a second systemic reaction at 60% [25]. The patient’s prior sting history – the severity and pattern of reactions, baseline tryptase level, age and concurrent medications all influence future risk [2]. Hymenoptera sting-induced anaphylaxis must prompt investigations for an underlying mast cell disorder including hereditary α-tryptasemia [14-17, 26].
Local reactions
In the general population, the reported prevalence of LLR ranges between 2.4% and 26.4%. In children, this figure is lower, while in beekeepers, it is higher, at 38% [27]. If local inflammation is contiguous with the sting site, it may be considered and managed as a local reaction [2]. LLR are not dangerous unless they cause compression, and compartment syndrome develops, or if a patient is stung in the oropharynx, when airway obstruction becomes a risk [28], or in the context of an underlying condition [19].
LLR patients exhibit a 10% risk of systemic reactions and a 3% risk of severe anaphylaxis if re-stung  [2, 28].
Diagnostics Sensitization
In vitro diagnostics
Detection of HBV sensitization and a convincing clinical history are required for the diagnosis of HBV [4]. Current guidelines recommend skin testing with allergen extracts as the first step for Hymenoptera venom allergy, however, the adjunction of in vitro testing with allergen extracts and allergen components (recombinant, CCD-free molecular allergens) helps refine the diagnosis and the therapeutic management decisions [4, 29]. However, recent data showed that in vitro and skin tests yielded complementary rather than overlapping results, and that in vitro diagnosis was best performed prior to skin tests  [25].
As Hymenoptera venom IgE persist for extended periods, in vitro and skin testing can be done even a long time after the reported clinical reaction, however, it is recommended to observe a 2-week interval after the reaction before performing skin tests [4, 25]. In a prospective diagnostic study, the positive predictive value of ImmunoCAP in vitro testing of Hymenoptera extracts was 77% and the negative predictive value 59%, while skin tests yielded 87% and 55% respectively [25].
Besides allergen-extract specific IgE, in vitro investigation of Hymenoptera sting-induced reactions comprises allergen component IgE and CCD IgE determination to assess genuine versus cross-reactive sensitization, as double positivity to bee and wasp venom extracts during in vitro testing occurs in up to 50% of venom-allergic patients [4, 30, 31]. Total IgE measurements may be useful for calculating the specific-to-total IgE ratio, a proposed indicator for clinically relevant sensitization [4]. If, based on clinical history, the index of suspicion for anaphylactic reaction is high, but in vitro and skin tests are negative, testing should be repeated in three to six months [8].
Diagnostic investigation of Hymenoptera sting-induced systemic reactions also requires determination of baseline tryptase in search for a mast cell disorder, with levels at 8 µg/L or higher suggesting hereditary α-tryptasemia [4, 17]. Further investigations such as testing for the D816V c-kit mutation in peripheral blood may be considered [16].
In the approximately 5% of Hymenoptera venom-allergic patients with elevated baseline tryptase levels and/or mastocytosis, using molecular components and a decreased threshold sIgE level to 0.1 kUA/L may be needed, in order to enhance sensitivity [32].
The high prevalence of Hymenoptera venom sensitization in the general population (42%) explains why screening for Hymenoptera venom allergy is not recommended.
Skin prick tests
Diagnostic intradermal testing is usually carried out using venom extracts of concentrations of between 0.001 µg/mL and 1.0 µg/mL. The accuracy of the test is subject to proportionate representation of the relevant allergens in the extract, as false-negatives may be the result of underrepresented components and, conversely, irritant compounds may lead to false-positives [2].
Children have been shown to demonstrate lower intradermal testing sensitivity than adults. This could suggest that leaving out venom extract concentrations below 0.01 µg/L may facilitate less painful and less labor intensive testing without compromising accuracy [33].
Challenge tests
Live sting challenges are not a standard procedure in clinical practice [2, 4, 19].
Other topics
Prevention and Therapy
Allergen immunotherapy
Venom immunotherapy (VIT) is the only treatment that can prevent future sting-induced anaphylaxis in Hymenoptera venom-allergic patients and can induce tolerance in 75–98% of cases. The scale of success of VIT is attributable to its wide availability and to well-established protocols [2, 34].
VIT is most successful when treatment is selected based on specific IgE to venom allergens, i.e., bee and/or wasp molecular allergens [4, 19]. Candidates for VIT must have a documented history of a systemic reaction to a sting and evidence of IgE reactivity to a specific venom. Treatment with the incorrect venom, or with more than one venom without evidence of sensitization, can induce de novo sensitization, increase the likelihood of adverse effects, risk insufficient protection, and increase costs [34].
The usual duration of VIT is 3 to 5 years, although more prolonged or even lifelong VIT should be considered in patients with mast cell disorders [19]
Even in patients without diagnosed SM, an elevated baseline serum tryptase in a venom-allergic patient may be associated with very severe anaphylactic reactions [13] and may indicate the need for lifelong VIT [35].
Prevention strategies
Sting avoidance is difficult to achieve as it requires caution during outdoor activities [4].
Other topics
An emergency kit comprising autoinjectable epinephrin should be carried by HBV-allergic patients having experienced systemic reactions, including those having completed a successful VIT [4].
T cell epitope-bearing long and short peptides and mimotopes are being investigated with the aim of  to overcoming VIT adverse effects [10]. Alternative routes of vaccine administration have also been investigated: intralymphatic delivery of low-dose vaccine shows potential for an enhanced immune response and a markedly reduced treatment duration, from five years to just 12 weeks [10].
Molecular Aspects
Allergenic molecules
Api m 1, Api m 2, Api m 3, Api m 5, and Api m 10 are considered as HBV major allergens, albeit with great variations in their prevalence according to the patient population, geographical areas, and method of detection [4]. Seventy-four per cent of patients have been shown to be sensitized to multiple allergens within HBV. Testing for Api m 1–5 and Api m 10 was associated with 95% diagnostic sensitivity [36]. The greatest prevalence of clinical sensitization to components of HBV is demonstrated for the highly abundant allergens Api m 1 (57-97%) and Api m 10 (51.5-61.8%) [36]. However, IgE reactivity for Api m 1 or Api m 10 may lack in patients with primary HBV sensitization [37].
Using multiple bee venom components for in vitro testing improves diagnostic sensitivity and specificity, facilitating design of more effective VIT [4].  Other methods of in vitro testing include basophil activation with allergen extracts or components [4].
Table 1. The general properties of the 12 known HBV allergens. Adapted from Hoffmann-Sommergruber et al 2022, Elieh et al 2018, Blank et al 2020, and WHO/IUIS Allergen Nomenclature Sub-Committee [1, 4, 5, 38].
Allergen Common name(s) Molecular weight (kDa) as per WHO/IUIS Native allergen glycosylated? Properties Sensitization rate (%) in HBV-allergic patients
Api m 1 Phospholipase A2 16 Y
Marker allergen for HBV sensitization
Allows discrimination between HBV and yellow jacket venom (YJV)/paper wasp venom (PDV) sensitization
Binds directly to the CD206 mannose receptor
Secreted PLA2 (sPLA2) has a key role in a wide range of cellular responses, such as phospholipid metabolism, signal transduction and regulation of inflammatory and immune responses
Induces IL-4 release from murine mast cells and IgE response on low-dose immunization
47-97
Api m 2 Hyaluronidase 44 Y
Potential marker for HBV sensitization. Homology with Ves v 2 in YJV causes limited cross-reactivity with Ves v 2 and Pol d 2 in absence of CCDs
Catalyzes the hydrolysis of hyaluronan (HA)
Activity potentiates infiltration by dissolving the extracellular matrix
46.3-52.2
Api m 3 Acid phosphatase 43 Y
Marker allergen for HBV sensitization. Particularly valuable in Api m 1-negative patients
Allows discrimination between HBV and YJV/PDV sensitization
Present as both monomers of about 48 kDa and dimers
Can cause histamine release from sensitized human basophils, thus produces a wheal-and-flare reaction after intradermal injection
50
Api m 4 Melittin 3 N
Main lethal component
Promelittin during biosynthesis is converted to 22-amino-acid peptide, melittin
Possesses a predominantly hydrophobic N-terminal region and a hydrophilic C-terminal region
22.9-42.5
Api m 5
Allergen C/
dipeptidyl peptidase IV
100 Y Homology with Ves v 3 in yellow jacket causes high cross-reactivity, preventing its use as a marker allergen for HBV sensitization 58.3-60
Api m 6   8 N Unglycosylated allergen, exists as four isoforms of 7190, 7400, 7598, and 7808 Da  
Api m 7 CUB serine protease 1 39      
Api m 8 Carboxylesterase 70      
Api m 9 Serine carboxypeptidase 60      
Api m 10 Icarapin 50-55 Y
Marker allergen for HBV sensitization. Particularly valuable in Api m 1-negative patients
Allows discrimination between HBV and YJV/PDV sensitization
Dominant Api m 10 sensitization is a putative marker for risk of VIT failure
Phosphorylated allergen
51.5-61.8
Api m 11 Major royal jelly protein 9 50 (deglycosylated form)      
Api m 12 Vitellogenin 200 Y Peptide belonging to vitellogenin family  
Biomarkers of severity
Cross-reactivity
Double positivity to bee and wasp venom during in vitro testing, seen in 40% to 50% of venom-allergic patients, is solved by the use of marker allergens (Api m 1, Api m 3, Api m 4 and Api m 10 for HBV, Ves v 1 and Ves v 5 for Vespids) and CCD, allowing distinction between genuine bee-wasp cosensitization and sensitization to only bee or wasp with cross-reactivity through shared allergens or CCD [4].
Honeybee and bumblebee venoms are highly cross-reactive and differential diagnosis is not currently available [4].
Natural Api m 1 carries a core glycan structure that reacts with IgE directed at CCDs. Recombinant venom components, including  rApi m 1, lack all or part of that structure, which increases specificity since only IgE antibodies directed at the PLA2 protein epitopes will bind to rApi m 1 [31].
Compiled By
Author: Prof. Joana Vitte
Reviewer: Dr. Merima Mehic Chaveton
Last reviewed: May 2023
References
  1. Elieh Ali Komi D, Shafaghat F, Zwiener RD. Immunology of Bee Venom. Clin Rev Allergy Immunol. 2018;54(3):386-96.
  2. Tracy JM, Golden DBK. Hymenoptera Venom Extracts in Clinical Practice. J Allergy Clin Immunol Pract. 2018;6(6):1856-62.
  3. Michez D. The oldest fossil of a melittid bee (Hymenoptera: Apiformes) from the early Eocene of Oise (France). Zoological Journal of the Linnean Society. 2007;150:701-9.
  4. EAACI. MAUG 2.0 2022 [Available from: https://hub.eaaci.org/resources_guidelines/molecular-allergology-users-guide-2-0/.
  5. IUIS/WHO. IUIS/WHO Apis mellifera 2023 [Available from: http://allergen.org/search.php?allergenname=&allergensource=apis+mellifera&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
  6. Pucca MB, Cerni FA, Oliveira IS, Jenkins TP, Argemi L, Sorensen CV, et al. Bee Updated: Current Knowledge on Bee Venom and Bee Envenoming Therapy. Front Immunol. 2019;10:2090.
  7. Diaz JH. Recognition, management, and prevention of hymenopteran stings and allergic reactions in travelers. J Travel Med. 2009;16(5):357-64.
  8. Philipp A, Ferdman RM, Tam JS. Evaluation of venom allergy. Ann Allergy Asthma Immunol. 2016;117(4):344-7.
  9. Turner PJ, Jerschow E, Umasunthar T, Lin R, Campbell DE, Boyle RJ. Fatal Anaphylaxis: Mortality Rate and Risk Factors. J Allergy Clin Immunol Pract. 2017;5(5):1169-78.
  10. Zahirovic A, Luzar J, Molek P, Kruljec N, Lunder M. Bee Venom Immunotherapy: Current Status and Future Directions. Clin Rev Allergy Immunol. 2020;58(3):326-41.
  11. Bilo MB, Bonifazi F. The natural history and epidemiology of insect venom allergy: clinical implications. Clin Exp Allergy. 2009;39(10):1467-76.
  12. Muller UR. Bee venom allergy in beekeepers and their family members. Curr Opin Allergy Clin Immunol. 2005;5(4):343-7.
  13. Bilo BM, Rueff F, Mosbech H, Bonifazi F, Oude-Elberink JN, Hypersensitivity EIGoIV. Diagnosis of Hymenoptera venom allergy. Allergy. 2005;60(11):1339-49.
  14. Schuler CFt, Volertas S, Khokhar D, Yuce H, Chen L, Baser O, et al. Prevalence of mastocytosis and Hymenoptera venom allergy in the United States. J Allergy Clin Immunol. 2021;148(5):1316-23.
  15. Greiner G, Sprinzl B, Gorska A, Ratzinger F, Gurbisz M, Witzeneder N, et al. Hereditary alpha tryptasemia is a valid genetic biomarker for severe mediator-related symptoms in mastocytosis. Blood. 2021;137(2):238-47.
  16. Selb J, Rijavec M, Erzen R, Zidarn M, Kopac P, Skerget M, et al. Routine KIT p.D816V screening identifies clonal mast cell disease in patients with Hymenoptera allergy regularly missed using baseline tryptase levels alone. J Allergy Clin Immunol. 2021;148(2):621-6 e7.
  17. Selb J, Rijavec M, Kopac P, Lyons JJ, Korosec P. HalphaT is associated with increased risk for severe Hymenoptera venom-triggered anaphylaxis. J Allergy Clin Immunol. 2023;151(3):804-5.
  18. Pastorello EA, Borgonovo L, Preziosi D, Schroeder JW, Pravettoni V, Aversano MG, et al. Basal Tryptase High Levels Associated with a History of Arterial Hypertension and Hypercholesterolemia Represent Risk Factors for Severe Anaphylaxis in Hymenoptera Venom-Allergic Subjects over 50 Years Old. Int Arch Allergy Immunol. 2021;182(2):146-52.
  19. Sturm GJ, Varga EM, Roberts G, Mosbech H, Bilo MB, Akdis CA, et al. EAACI guidelines on allergen immunotherapy: Hymenoptera venom allergy. Allergy. 2018;73(4):744-64.
  20. Lindstrom I, Holtta P, Suuronen K, Suomela S, Suojalehto H. High prevalence of sensitization to bumblebee venom among greenhouse workers. J Allergy Clin Immunol Pract. 2022;10(2):637-9.
  21. Khurana T, Bridgewater JL, Rabin RL. Allergenic extracts to diagnose and treat sensitivity to insect venoms and inhaled allergens. Ann Allergy Asthma Immunol. 2017;118(5):531-6.
  22. Cifuentes L. Allergy to honeybee ... not only stings. Curr Opin Allergy Clin Immunol. 2015;15(4):364-8.
  23. Rodriguez-Perez R, Carretero P, Brigido C, Nin-Valencia A, Carpio-Hernandez D, Tomas M, et al. The new Api m 11.0301 Isoallergen From Apis mellifera Is a Food Allergen From Honey. J Investig Allergol Clin Immunol. 2022;32(6):492-3.
  24. Neugut AI, Ghatak AT, Miller RL. Anaphylaxis in the United States: an investigation into its epidemiology. Arch Intern Med. 2001;161(1):15-21.
  25. Park HJ, Brooks DI, Chavarria CS, Wu RL, Mikita CP, Beakes DE. Combining Discordant Serum IgE and Skin Testing Improves Diagnostic and Therapeutic Accuracy for Hymenoptera Venom Hypersensitivity Immunotherapy. J Allergy Clin Immunol Pract. 2022;10(3):837-43 e3.
  26. Kacar M, Rijavec M, Selb J, Korosec P. Clonal mast cell disorders and hereditary alpha-tryptasemia as risk factors for anaphylaxis. Clin Exp Allergy. 2023.
  27. Bilo MB, Martini M, Pravettoni V, Bignardi D, Bonadonna P, Cortellini G, et al. Large local reactions to Hymenoptera stings: Outcome of re-stings in real life. Allergy. 2019;74(10):1969-76.
  28. Golden DB. Large local reactions to insect stings. J Allergy Clin Immunol Pract. 2015;3(3):331-4.
  29. Ansotegui IJ, Melioli G, Canonica GW, Caraballo L, Villa E, Ebisawa M, et al. IgE allergy diagnostics and other relevant tests in allergy, a World Allergy Organization position paper. World Allergy Organ J. 2020;13(2):10008.
  30. Muller U, Schmid-Grendelmeier P, Hausmann O, Helbling A. IgE to recombinant allergens Api m 1, Ves v 1, and Ves v 5 distinguish double sensitization from crossreaction in venom allergy. Allergy. 2012;67(8):1069-73.
  31. Spillner E, Blank S, Jakob T. Hymenoptera allergens: from venom to "venome". Front Immunol. 2014;5:77.
  32. Michel J, Brockow K, Darsow U, Ring J, Schmidt-Weber CB, Grunwald T, et al. Added sensitivity of component-resolved diagnosis in hymenoptera venom-allergic patients with elevated serum tryptase and/or mastocytosis. Allergy. 2016;71(5):651-60.
  33. Cichocka Jarosz E, Stobiecki, M., Brzyski, P. et al. Simplification of intradermal skin testing in Hymenoptera venom allergic children. Ann Allergy Asthma Immunol. 2017;118:326-32.
  34. Ollert M, Blank S. Anaphylaxis to insect venom allergens: role of molecular diagnostics. Curr Allergy Asthma Rep. 2015;15(5):26.
  35. Simons FE, Ardusso LR, Bilo MB, Cardona V, Ebisawa M, El-Gamal YM, et al. International consensus on (ICON) anaphylaxis. World Allergy Organ J. 2014;7(1):9.
  36. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  37. Kohler J, Blank S, Muller S, Bantleon F, Frick M, Huss-Marp J, et al. Component resolution reveals additional major allergens in patients with honeybee venom allergy. J Allergy Clin Immunol. 2014;133(5):1383-9, 9 e1-6.
  38. Blank S. Marker allergens in Hymenoptera Venom Allergy - Characteristics and potential use in precision medicine. Allergo J Int. 2020.