Type:
Component
Component
Component
Api m 3
Acid phosphatase
i215
43 kDa
Recombinant protein
Honeybee (Apis mellifera) venom acid phosphatase
Api m 3, a glycoprotein of the acid phosphatase family, is a major allergen of Apis mellifera (honeybee) venom (HBV) and a marker allergen for genuine sensitization to this venom.
Api m 3 may be underrepresented in HBV extracts, which may affect the outcome of VIT if Api m 3 is the predominant sensitizer.
Api m 3 sensitization has a prevalence of 28% to 63% among HBV-allergic populations (reviewed in [1]). Thus, Api m 3 is considered a major allergen in HBV-allergic patients.
Apparent monosensitization to recombinant (r)Api m 3, i.e. without detectable sensitization to any other HBV allergen, was reported in 5% of a German cohort of 144 HBV-allergic patients displaying 50% overall prevalence of Api m 3-specific IgE [2].
Api m 3 is an enzyme secreted into the venom sac of Apis mellifera, where it contributes 1.7% ± 0.4% of venom dry weight [3].
Sensitization to Api m 3 occurs through injection (bee sting).
Api m 3 does not cross-react with Vespid venom allergens, therefore, Api m 3 is a marker allergen allowing to discriminate between HBV and Vespid venom sensitization and identifyf genuine HBV sensitization [1].
An acid phosphatase from bumblebee venom, not yet included in the IUIS/WHO database of allergens, was shown to display moderate cross-reactivity with Api m3 [1]. Acid phosphatases from other Hymenoptera venoms (bumblebees, ants, but not Vespids) exhibit sequence identity of 50% or higher with Api m 3 [1, 4].
Various patterns of sensitization to HBV allergens have been described, but no clinical correlate of severity has been identified so far [1].
Also known as acid phosphatase, Api m 3 is a secreted glycoprotein with a molecular weight of 43 kDa (388 aminoacids in the pro-form), displaying N-glycosylated side chains and disulfide bonds, found in HBV as monomers or homodimers [1, 4, 5]. The exact biological function of Api m 3 as an acid phosphatase is still elusive [1].
As of July 8, 2023, a unique isoallergen, Api m 3.0101, has been included in the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature [6].
Api m 3 displays aminoacid sequence similarity of 50% or higher restricted to venoms from bees, bumblebees, and ants [4].
The demonstration of specific IgE to Api m 3 confirms genuine sensitization to HBV or bumblebee venom [1]. With a prevalence of up to 63% in HBV-allergic patients, IgE to Api m 3 is a relevant clinical tool [1]. Indeed, in populations of HBV allergic patients with low figures of prevalence of Api m 1-specific IgE, using additional HBV marker allergens, mainly Api m 3, Api m 4 and Api m 10, increases the chance of demonstrating genuine sensitization to HBV [1, 2, 7].
In Hymenoptera IgE testing, the quantitative result of specific IgE to a molecular allergen or whole venom extract is neither predictive of, nor correlated to the severity of the reaction.
The prevalence of sensitization to individual HBV allergens, including Api m 3, in HBV-allergic patients varies depending on multiple factors such as geography, patient inclusion criteria, single or double positivity to HBV and Vespid venoms, underlying sensitization profile, use of a recombinant allergen versus a natural purified allergen, the assay format [1, 7]. Thus, the diagnostic sensitivity of rApi m 3-specific IgE ranges from 28 to 63% in HBV-allergic patients [1, 2, 7]. Using a panel of HBV marker allergens such as Api m 1, Api m 3, and Api m 10 improves the rate of confirmation of genuine HBV sensitization [1, 2, 7].
As an example, in a study addressing European HBV-allergic patients from the Czech Republic and Slovenia, the diagnostic sensitivity of rApi m 3-specific IgE was 28% in HBV-allergic patients devoid of Vespid sensitization and 49% in patients exhibiting HBV and Vespid venom sensitization, with an overall diagnostic sensitivity of 38% [7]. In this cohort, a panel comprising Api m 1, Api m 2, Api m 3, and Api m 10 increased the diagnostic sensitivity to 91% , in line with previous reports from other cohorts [2, 7].
The reported diagnostic specificity of IgE to rApi m 3 for HBV allergy is 100% [2, 7]. Api m 3 sensitization was not detected in subjects without HBV allergy, even in those with a history of bee stings or detectable IgE to HBV whole extract [2, 7]
The demonstration of Api m 3-specific IgE contributes to the identification of genuine HBV sensitization, thus supporting the choice of HBV AIT in eligible patients [1].
It has been suggested that Api m 3 might be underrepresented in therapeutic HBV extracts for VIT [2]. A lack of inhibition of IgE binding to Api m 3 following serum preincubation with HBV therapeutic extract, and failure to mount a specific IgG4 response to Api m 3 were observed, and might influence the outcome of immunotherapy [2].
Author: Prof. Joana Vitte
Reviewer: Dr. Merima Mehic Chaveton
Last reviewed: 2023-07-20