Anna and Oliver, both 8 years old, have a history of birch and soy allergy, both visit their healthcare provider after experiencing rhinitis during pollen season and dry skin.
Anna and OIiver's healthcare provider conducts a full clinical history and physical examination of them both and decides to test using soy components.
Two kids have a history of birch and soy allergy and visit their healthcare provider after experiencing dry skin and rhinitis during pollen season.
Test |
Type |
Anna's Results |
Oliver's Results |
Egg |
Whole Allergen |
0 |
0 |
Milk |
Whole Allergen |
0 |
0 |
Soy |
Whole Allergen |
+2 |
+2 |
Wheat |
Whole Allergen |
0 |
0 |
Fish |
Whole Allergen |
0 |
0 |
Birch |
Whole Allergen |
+2 |
+1 |
Timothy grass |
Whole Allergen |
0 |
0 |
Anna and Oliver's previous healthcare provider diagnosed them with birch and soy allergies. She recommended the use of oral antihistamines during birch pollen season and directed them to avoid soy and soy products.
These results together with this patient's case history and symptoms, help confirm the diagnosis.
Test |
Type |
Anna’s Results |
Oliver’s Results |
Soy |
Whole Allergen |
0.8 |
24 |
Gly m 4 |
Allergen Component |
15 |
2.5 |
Gly m 5 |
Allergen Component |
<0.1 |
16.2 |
Gly m 6 |
Allergen Component |
<0.1 |
6.7 |
Birch |
Whole Allergen |
58 |
20 |
Anna’s results indicate a clinically relevant cross-reaction between birch pollen and soy. Specific IgE antibodies to Gly m 4 are often associated with local reactions. However, systemic reactions may occur, particularly when consuming high amounts of low processed soy, e.g., soy milk. If clinical history is unclear, it may be advisable to proceed with a food challenge to confirm the diagnosis of soy allergy.1
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
Anna's healthcare provider advises her:
FOLLOW UP
Oliver’s results indicate his level of sensitization to Gly m 5 put him at an increased risk of severe reaction if soy is ingested, even small amounts.1
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
Oliver's healthcare provider advises him:
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.