Thomas, a 17-year-old boy with a history of egg allergy, visits his healthcare provider following two recent emergency room visits due to reaction to egg. Thomas has stopped carrying his epinephrine autoinjector and his mother is worried that he doesn’t take his allergy seriously.
Thomas’ healthcare provider conducts a full clinical history and physical examination and decides to test him for sensitization to egg white and components to help reinforce the seriousness of his egg allergy diagnosis.
Thomas has had severe reactions due to an egg allergy. Will specific IgE blood testing help him take his allergy seriously?
These results together with this patient's case history and symptoms, help confirm the diagnosis.
Test |
Type |
Thomas’ Results |
---|---|---|
Egg white |
Whole Allergen |
56.9 |
Gal d 1 (Ovomucoid) |
Allergen Component |
10.73 |
Gal d 2 (Ovalbumin) |
Allergen Component |
9.74 |
Test |
Type |
Thomas’ Results |
---|
Thomas’ test results revealed that he was highly sensitized to ovomucoid, which is associated with an increased risk of systemic reaction to all forms of egg.1
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.