As the gold standard for specific IgE blood tests,1 ImmunoCAP tests offer high performance and reliable results.2
Unlike traditional skin-prick tests, ImmunoCAP tests can be performed irrespective of age, disease activity, skin condition, medications, or pregnancy. They also pose no risk for anaphylaxis.3
for the relevant allergens (defined according to symptoms, age, and local sensitisation patterns) at your local pathology laboratory.4
(2.5 mL whole blood or 1 mL serum).
with the precise test results, for personalised patient management.
When you receive your patient’s specific IgE blood test results, they should be interpreted in conjunction with the clinical history and physical examination.5,6
ImmunoCAP blood test results will indicate whether the patient is sensitised (has IgE antibodies) to specific allergens. However, sensitisation to specific allergens does not always mean the patient has an allergy, as sensitisation does not always result in symptoms.7 Allergy is the symptomatic response to allergen(s) that an individual is sensitised to.3 The higher the specific IgE antibody concentration, the higher the risk of allergy.8
Specific IgE blood tests can help you diagnose or rule out an allergy, thereby directing your next steps in the management plan for your patient.6
Compared with a clinical history and physical examination, additional use of ImmunoCAP blood tests provides a more reliable diagnosis:10*
*Based on a prospective multicentre study of 380 children aged <6 years with symptoms of eczema and/or wheezing/asthma and/or rhinitis.10
Referral to specialist centres will be necessary in certain cases, such as when there is a discrepancy between the clinical history and specific IgE results.3,6
Where allergy has been confirmed, specific IgE can help pinpoint the causative allergen(s), thus allowing you to:4
Even in the absence of allergy, specific IgE antibodies could:7
As allergies change over time,11 regular follow-up tests of specific IgE could help you appropriately adapt the management of your patient by detecting:
IgE: immunoglobulin E; kUA/L: allergen-specific kilo units per litre