Talk with Us
ImmunoCAP Specific IgG4
An Aid During Allergen-Specific Immunotherapy
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Quantifying allergen-specific IgG4 antibodies in serum with ImmunoCAP™ Specific IgG4 tests1 offers proper evaluation of immunological response.2 During allergen-specific immunotherapy (AIT), increased levels of allergen-specific IgG4 indicate that the immune system is responding to therapy. IgG4 has the ability to modulate the immune response to allergen and thus the potential to influence the clinical response.3
With ImmunoCAP Specific IgG4, valuable clinical information in the fields of allergic disease and immunotherapy can be obtained. Measurement of specific IgG4 antibodies have also been used in clinical studies of different allergic diseases such as asthma, rhinitis, atopic dermatitis, and gastrointestinal disorders.4,5
Follow up allergen-specific immunotherapy (AIT)
During immunotherapy, increased levels of specific IgG4 may indicate:
  • A general, but not definitive, correlation with clinical outcome.5
  • Show that the immune system is responding to the therapy.5
There is no common cut-off value for above normal levels of circulating specific IgG4 antibodies, as these are markers for allergen exposure, which may not be directly related to the disease and will depend on the local environment and levels of exposure.5
ImmunoCAP Specific IgG4 tests
View Catalog
Article Number 14-4100-01
Product Type Test
Test Category Grass Pollens
Test Type Whole Allergen
Barcode 350
Size 16
Product Info
Article Number 14-4107-01
Product Type Test
Test Category Mites
Test Type Whole Allergen
Barcode 357
Size 16
Product Info
Article Number 10-9481-01
Product Type Reagent
Barcode C9B
Size 2 x 50
Product Info
Explore the full ImmunoCAP portfolio.
View Portfolio
Find an instrument to run ImmunoCAP tests.
View Phadia Laboratory Systems
Get scientific information on whole allergens, allergen mixes, and allergen components.
Visit Allergen Encyclopedia
References
  1. Jutel, M. and C.A. Akdis, Immunological mechanisms of allergen-specific immunotherapy. Allergy, 2011. 66(6): p. 725-32.
  2. Directions for use. ImmunoCAP Specific IgG4.
  3. Jutel, M., A. Kosowska, and S. Smolinska, Allergen Immunotherapy: Past, Present, and Future. Allergy Asthma Immunol Res, 2016. 8(3): p. 191-7.
  4. Smoldovskaya, O., et al., Allergen-specific IgE and IgG4 patterns among patients with different allergic diseases. World Allergy Organ J, 2018. 11(1): p. 35.
  5. Ito, K., et al., The usefulness of casein-specific IgE and IgG4 antibodies in cow's milk allergic children. Clin Mol Allergy, 2012. 10(1): p.1.