Accurate diagnosis of food allergy is essential for personalised patient management,1 including avoidance of unnecessary dietary restrictions and prevention of life-threatening reactions.2 Egg is one of the most common food allergy sources, particularly in infants and children.1 The prevalence of egg allergy in children is estimated to be around 0.5-2.5 percent.3
Read through this evidence-based guide to learn how to accurately diagnose egg allergy in clinical practice.
Sometimes the clinical history and specific IgE blood test results are strongly suggestive of an egg allergy, but often an OFC with egg is necessary to confirm the diagnosis. The presence or absence of a symptomatic response will demonstrate allergy or tolerance, respectively.5
OFCs should be performed in a specialist setting with rapid emergency support available; for patients deemed by their healthcare provider to be at medium-to-high risk of having a severe reaction, intensive care support must also be immediately available.5
Patients should be re-evaluated at around 6-12 months, to avoid continuing the restrictive exclusion diet for an unnecessarily long time. Repeated specific IgE blood tests can be helpful to:5
IgE: immunoglobulin E