Talk with Us
Baked Egg: Improving characterization and management of egg allergic patients
Andrea and Sylvia, 3-year-old girls both with a history of eczema and positive skin-prick test for egg white, are seen by their healthcare provider for additional insight into their egg allergies.
Andrea and Sylvia’s healthcare provider conducts a full clinical history and physical examination and decides to test using egg components.
Andrea and Sylvia were both diagnosed with egg allergies as infants. Will their test results change over time?
Patient History
Family History
  • None
Andrea and Sylvia Have the Same Personal History
  • Severe eczema since 6 to 7 months of age.
  • Gastrointestinal symptoms.
  • Urticaria and wheezing when accidentally eating egg.
Skin Prick Test
Test
Type
Andrea's Results
Sylvia's Results
Egg White
Whole Alergen
+5
+4
Andrea and Sylvia had a negative skin-prick test to milk.
ImmunoCAP™ Test Results (KUA/I)
Test
Type
Andrea's Results
Sylvia's Results
Egg white
Whole Allergen
20
22
Based on history, evaluation, and testing their previous healthcare provider recommended that Andrea and Sylvia remove egg entirely from their diets.
ImmunoCAP Test Results
These results, together with Andrea and Sylvia's case history and symptoms, may help their healthcare provider confirm the diagnosis.
ImmunoCAP Test Results (KUA/I)
Test
Type
Andrea's Results
Sylvia's Results
Egg white
Whole Allergen
16
19
Gal d 1 (Ovomucoid)
Allergen Component
0.4
16
Gal d 2 (Ovabumin)
Allergen Component
11.1
6.9
Gal d 3 (Conalbumin)
Allergen Component
0.4
0.2
Gal d 4 (Lysozyme)
Allergen Component
4.5
0.3
Differential Diagnosis
Andrea
Andrea’s low level of sensitization to Gal d 1 may be an indication of tolerance to baked egg.1,2 An open challenge in the clinic with baked egg may be performed to help you confirm the diagnosis. With components, Andrea’s quality of life could have been improved: No need of strict avoidance of egg and no cause of stress in the family due to the risk of eating products containing baked egg, such as cakes and cookies.
REFINED DIAGNOSIS
  • Likely tolerant to baked egg.
HEALTHCARE PROVIDER MANAGEMENT PLAN
  • Andrea’s healthcare provider advised that she undergo an oral food challenge with a specialist to confirm tolerance to baked egg.
FOLLOW UP
  • Andrea has successfully passed the oral food challenge for baked egg and has been eating foods containing extensively baked egg since with no issues.
Sylvia
Sylvia’s significant levels of specific IgE to Gal d 1 indicates that she has a risk for reaction to all forms of egg.1,2
REFINED DIAGNOSIS
  • Egg allergy.
HEALTHCARE PROVIDER MANAGEMENT PLAN
  • Sylvia’s healthcare provider advised a dietary restriction to all forms of egg remains.
FOLLOW UP
  • Sylvia is seen by her healthcare provider one year later. She has continued to avoid egg in all forms and is doing well. Her autoinjector technique is satisfactory.
    It may be wise to re-test her to see if she may have outgrown her egg allergy and ready to go through an oral challenge.
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
References
  1. Lemon-Mule H, et al. J Allergy Clin Immunol. 2008;122:977-83 e1.6.
  2. Ando H, et al. J Allergy Clin Immunol. 2008;122:583.