Alice, a 7-year-old who has experienced gastrointestinal distress, vomiting, and urticaria after consuming products with milk, visits her healthcare provider for testing.
The healthcare provider conducts a full clinical history and physical examination and decides to test using whole milk with reflex to components.
Specific IgE blood testing can help determine how likely Alice is to tolerate milk.
These results together with this patient's case history and symptoms, help confirm the diagnosis.
Test |
Type |
Alice’s Results |
---|---|---|
Milk |
Whole Allergen |
12 kU/L |
Bos d 4 (α-lactalbumin) |
Allergen Component |
1.22 |
Bos d 5 (β-lactoglobulin) |
Allergenr Component |
<0.10 |
Bos d 8 (casein) |
Allergen Component |
7.53 |
Test |
Type |
Alice’s Results |
---|
Alice’s sensitization to α-lactalbumin and significant levels of IgE to casein indicates a risk for reaction to all forms of milk. 1
REFINED DIAGNOSIS
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.