Recent searches Clear History
お問い合わせ
イムノキャップ総IgE
IgEの総濃度を定量化する
イムノキャップ総IgEは、ヒトの血清サンプルまたは血漿サンプルに含まれる遊離IgE抗体の総量(以下、総IgE抗体価)を測定します1。アレルギーを原因とする喘息、鼻炎、アトピー性皮膚炎を患う多くの患者さんでは、総IgE抗体価が上昇することがあります1-4
イムノキャップ総IgEは定量検査で、2~5000 kU/Lの幅広い測定範囲内で総IgE抗体価を測定します1
イムノキャップについて確認する
製品カタログをダウンロード
保険適用で測定可能なアレルゲン一覧をダウンロード
参考文献
  1. Directions for use.ImmunoCAP Total IgE.
  2. Cakanlar, F.N., E. Ceylan, and A. Kanbay, Relationship between serum total ige specific ige and serum total eosinophil counts in asthma bronchiale patients.World Allergy Organization Journal, 2015.8(Suppl 1): p. A25.
  3. Poleshuck, R.T.F., et al., Levels of total serum IgE and specific IgE sensitization profile in patients with atopic dermatitis.World Allergy Organization Journal, 2015.8(1): p. A142-A142.
  4. Poleshuck, Rafael Teixeira Figueredo et al.“Levels of total serum IgE and specific IgE sensitization profile in patients with atopic dermatitis” World Allergy Organization Journal, Volume 8, A142