+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Tests & Systems
COMPLETE SYSTEMS TO SUPPORT THE DIAGNOSIS OF ALLERGY, ASTHMA AND AUTOIMMUNE DISEASES
Proven performance in laboratories all over the world
Phadia™ Laboratory Systems, along with ImmunoCAP™ assays to support the diagnosis of allergic diseases and EliA™ assays for autoimmune diseases, are designed to meet the demands of clinicians—quickly and efficiently. With high sensitivity and specificity, these assays offer the ability to detect and quantitatively determine relevant antibodies in the blood.
Not only can the ImmunoCAP and EliA results produced by Phadia Laboratory Systems help clinicians guide patient care, but as your business grows our systems and software allow you to seamlessly increase your in-house testing capacity.
>4,750 systems in 70 countries across the world
Phadia Testing Systems
*Phadia 1000 is dedicated to running ImmunoCAP assays
Benefits include:
For your laboratory
In-house testing with diagnostic accuracy
Ability to run both ImmunoCAP and EliA assays on a single platform*
For your clinicians
Reliable results from assays with high predictive values which help aid diagnosis
Excellent consistency over time and between countries, systems, laboratories and users**
For your clinicians’ patients
Accurate results support efficient patient management, diagnoses and treatment decisions
*Phadia 1000 is dedicated to running ImmunoCAP assays
**Based on Quality Club data
Phadia Laboratory Systems can help your laboratory maximize operational efficiency
checkmark
Proven reagent quality Integrated QC concept, with calibrators and QC controls
specsens
Well-studied and documented sensitivity and specificity across assays
ribbon
In-house quality testing can help your facility achieve accreditation standards for allergy and autoimmunity
House
Your in-house solution for maximizing instrument performance and reducing operating costs
hand
Designed to be easy to use and run both ImmunoCAP and EliA assays*
watch
On-board storage for reagents, solutions, and waste—for less set-up time and increased efficiency
*Note: Phadia 1000  is an instrument dedicated to performing ImmunoCAP assays and does not run EliA autoimmune assays
ImmunoCAP and EliA assays on the same instrument*
For mid-size volume demands, the Phadia 250 instrument allows continuous sample loading for both ImmunoCAP and EliA assays with short switch over time in the assay run.
For higher-volume demands, the larger instruments—Phadia 2500  and Phadia 5000 instruments—offer multiple process lines that give your laboratory the ability to run ImmunoCAP or EliA assays simultaneously.
*Note: Phadia 1000  is an instrument dedicated to performing ImmunoCAP assays and does not run EliA autoimmune assays
system capabilities
Looking for specifics on Phadia
system capabilities?
autoimmune
Need detailed Information and data about individual assays
autoimmune
Keep up to date with the latest news and research