Designed to aid in the diagnosis of allergic diseases by providing test results with clinical relevance in a large, cost conscious laboratory
Part of the family of PhadiaTM Laboratory Systems, the Phadia 1000 instrument is a dedicated ImmunoCAPTM assay system for the detection of over 600 specific allergic sensitizations. It is capable of storing large volumes of reagents and ImmunoCAPTM carriers on board.
The Phadia 1000 instrument allows laboratories with high volumes of allergen sensitization testing to produce consistent and reproducible results.
Phadia 1000 instrument delivers a range of benefits, all with a size and capability that make sense for your laboratory:
Flexibility
Allergen panel detecting sensitizations to 600 allergens and 100 allergen components
Automatic start-up and shut-down function
User-configurable reflex testing functionality
Capacity
Throughput:
240 results/hour
Continuous random access
All reagents stored on board
28-day storage of calibration curves
Connectable to Laboratory Automation Systems
Clustered set-up for increased capacity
Reliability
Excellent consistency over time and between countries, systems, laboratories and users
Remote diagnostics via PhadiaTM LabCommunity
One of the largest quality assessment programs in the world
Not shown in picture:
Integrated loading area for stop solution bottles and container for used carriers
Stand-alone PC for Phadia™ Prime software, connectable to mainframe Laboratory Information Systems (LIS)
If your laboratory has been considering upgrading, or adding to your testing systems, Thermo Fisher Scientific can help you find the suitable Phadia Laboratory System for your needs.