A laboratory's investment in a Laboratory Information System (LIS) is key to streamline processing and track samples and test status through the laboratory and creates connectivity within your system. To ensure you can get the most of your PhadiaTM Laboratory Systems, Thermo Fisher Scientific puts great focus on ensuring that the PC based operator software unlocks the full potential of the Phadia instruments. The new PhadiaTM Prime software offers several valuable improvements compared to the previous PhadiaTM IDM software.
Phadia Prime | Feature | Phadia IDM |
---|---|---|
Yes | True server/client architecture |
Limited |
Yes | Sample QNS |
No |
Yes | Individualize adult and pediatric specimen processing |
No |
Yes | Enhanced LIS mainframe interfaces |
Limited |
Yes | True positive reflex testing |
Limited |
Fully Intergrated | LAS integration |
Requires external hardware |
Yes | Stock management | Limited |
Yes | Phadia™ LabCommunity |
Limited |
Enhanced | Patient report customization |
Limited |
Enhanced | Search functionality |
Limited |
Enhanced | Event log | Limited |
Yes | User role customization |
No |
In addition to the features listed above, Phadia Prime software offers several advanced LIS capabilities, such as:
Continuously improving workflow, such as through laboratory automation, is essential to the growth and development of clinical laboratories. Laboratory automation allows for integrated sample handling across various analyzers in the laboratory, and PhadiaTM 250 and PhadiaTM 1000 are capable of connecting to LAS tracks from leading providers. Establishing LAS connections require agreements between Thermo Fisher Scientific and the LAS provider.
Utilizing the LAS systems’ integrated preanalytical capabilities
Reducing operator time for sample handling for ImmunoCAPTM and EliATM assays
Reducing the need for aliquots and secondary sample tubes