+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
A TOOL TO EVALUATE AND PRESENT IMMUNOCAP ISAC RESULTS
Software Designed to Help Your Laboratory
PhadiaTM MIA is specially developed result evaluation software for ImmunoCAPTM ISAC microarray assays. Phadia MIA is required for your laboratory to handle the requests, results, calculations and statistics for dedicated in vitro diagnostic tests performed with ImmunoCAP ISAC.
Learn more about ImmunoCAP ISAC >
A Solution to Evaluate and Present Results
Phadia MIA is user-friendly making it easy for your laboratory to visualize scanned images from ImmunoCAP ISAC tests. In addition, the software allows you to translate the image intensities to antibody concentrations and present them on a result report that can be customized to meet the demands of your clinicians.
Request more info about Phadia MIA >
Key Phadia MIA functionality
Phadia MIA is PC software running on the Microsoft Windows operating system. The software supports USB communication with a connected LuxScanTM 10K Microarray Scanner.
Phadia MIA software is used for analyzing image files created by LuxscanTM 10k Microarray scanner from ImmunoCAP ISAC arrays and to transform these images into results. The image files can be stored and analyzed at a later stage, without the connection to the Luxscan microarray scanner.
Result data is stored in a database and can be exported to Phadia Prime/Phadia IDM or text-based file. It is also possible to print sample reports.