The PhadiaTM 200 instrument provides new levels of flexibility and automation in a benchtop instrument to aid in the diagnosis of allergic and autoimmune diseases for laboratories with limited space.
Confidence in numbers
Flexibility for the long term
Walk away automation
PERFORMANCE SPECIFICATIONS | ||||
Dimensions (H x W x D) |
55 cm x 80 cm x 60 cm |
Onboard Test Carrier Capacity | Carrier loading trays with 96 positions | |
Weight | 60 kg | Automation | Stand-alone operation with built-in touch screen |
|
Number of Samples | 42 sample tube positions/run | Software | Phadia Prime software |
|
Batch Processing | ImmunoCAP - 96 tests per batch EliA - 48 tests per batch |
Hands on set-up time | Approx. 20 minutes | |
Number of Methods | 4 in parallel (either EliA or ImmunoCAP) | Connectivity | LIS, Phadia LabCommunity, Phadia Prime cluster |
|
Batch Cycle Time | Up to 4 hours at max capacity |
If your laboratory has been considering upgrading, or adding to your testing systems, Thermo Fisher Scientific can help you
find the right Phadia Laboratory System for your needs.