+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
STREAMLINING YOUR DAILY ROUTINE
Software to support your laboratory's needs
PhadiaTM Information Data Manager (IDM) software helps your laboratory make the most of your PhadiaTM Laboratory Systems. Not only can Phadia IDM manage assay runs, test requests and results, but it also can help your laboratory ensure quality control of data and inventory.
Powerful technology to enhance your laboratory’s productivity
Phadia IDM, has all the key functionality required to operate Phadia Laboratory Systems. This software gives you the ability to seamlessly integrate up to 5 Phadia 100 instruments, 5 Phadia 250 instruments and 5 Phadia 1000 instruments in a single network.* In addition, Phadia IDM automates communication with other laboratory systems, such as Laboratory Information System (LIS) and Laboratory Automation System (LAS).
Offering several improvements to Phadia IDM, the new PhadiaTM Prime software provides better ease of use, new tests, test management and network connectivity.
See the differences >
*For laboratories using Phadia 2500 or Phadia 5000 instruments, only one instrument can be connected using Phadia IDM
Are you ready to take advantage of all Phadia Laboratory Systems can offer?
If your laboratory has been considering a Phadia Laboratory System or upgrading from Phadia IDM to Phadia Prime we can help.
Contact a sales representative >